Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2817967-2818369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
UniGene
Hs.446098
Gene Chromosome Location
Chr.12: 2812621-2832217 on Build GRCh38
Transcripts
NM_018463.3 NR_130744.1 XM_005253707.4 XM_005253709.4 XM_017019641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072064 c.406+1G>A p.? Chr.12: 2818278-2818278 on Build GRCh38
938525 c.405C>T p.I135I Chr.12: 2818276-2818276 on Build GRCh38
938526 c.406+9G>A p.? Chr.12: 2818286-2818286 on Build GRCh38
1361119 c.268T>C p.F90L Chr.12: 2818139-2818139 on Build GRCh38
6687692 c.333C>T p.I111I Chr.12: 2818204-2818204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780046954 Chr.12: 2818269-2818269 on Build GRCh38
rs754618933 Chr.12: 2818212-2818212 on Build GRCh38
rs767174094 Chr.12: 2818306-2818306 on Build GRCh38
rs371762049 Chr.12: 2818294-2818294 on Build GRCh38
rs747109095 Chr.12: 2818274-2818274 on Build GRCh38
rs886234920 Chr.12: 2818250-2818250 on Build GRCh38
rs775727717 Chr.12: 2818070-2818070 on Build GRCh38
rs376230301 Chr.12: 2818002-2818002 on Build GRCh38
rs761620739 Chr.12: 2818172-2818172 on Build GRCh38
rs754735307 Chr.12: 2818085-2818085 on Build GRCh38
rs759146836 Chr.12: 2818303-2818303 on Build GRCh38
rs368435841 Chr.12: 2818192-2818192 on Build GRCh38
rs1052032525 Chr.12: 2818190-2818190 on Build GRCh38
rs780564817 Chr.12: 2818344-2818344 on Build GRCh38
rs772993429 Chr.12: 2818193-2818193 on Build GRCh38
rs775608641 Chr.12: 2818123-2818123 on Build GRCh38
rs758777040 Chr.12: 2818337-2818337 on Build GRCh38
rs565413873 Chr.12: 2818218-2818218 on Build GRCh38
rs751233922 Chr.12: 2818083-2818083 on Build GRCh38
rs866146579 Chr.12: 2818029-2818029 on Build GRCh38
rs933183258 Chr.12: 2818143-2818143 on Build GRCh38
rs746343805 Chr.12: 2818256-2818256 on Build GRCh38
rs534541245 Chr.12: 2818277-2818277 on Build GRCh38
rs200862282 Chr.12: 2818109-2818109 on Build GRCh38
rs779380475 Chr.12: 2818240-2818240 on Build GRCh38
rs761669322 Chr.12: 2818063-2818063 on Build GRCh38
rs759006773 Chr.12: 2818261-2818261 on Build GRCh38
rs372931740 Chr.12: 2818016-2818016 on Build GRCh38
rs752372612 Chr.12: 2818314-2818314 on Build GRCh38
rs375089386 Chr.12: 2818297-2818297 on Build GRCh38
rs199535617 Chr.12: 2818163-2818163 on Build GRCh38
rs779321925 Chr.12: 2818097-2818097 on Build GRCh38
rs749353674 Chr.12: 2818281-2818281 on Build GRCh38
rs371530862 Chr.12: 2818204-2818204 on Build GRCh38
rs1005215879 Chr.12: 2818271-2818271 on Build GRCh38
rs111226449 Chr.12: 2817997-2817997 on Build GRCh38
rs576075732 Chr.12: 2818272-2818272 on Build GRCh38
rs767298903 Chr.12: 2818217-2818217 on Build GRCh38
rs531726711 Chr.12: 2818038-2818039 on Build GRCh38
rs755850400 Chr.12: 2818315-2818315 on Build GRCh38
rs757735011 Chr.12: 2818091-2818091 on Build GRCh38
rs772026053 Chr.12: 2818118-2818118 on Build GRCh38
rs368815825 Chr.12: 2818151-2818151 on Build GRCh38
rs757875504 Chr.12: 2818078-2818078 on Build GRCh38
rs750800905 Chr.12: 2818332-2818332 on Build GRCh38
rs768821581 Chr.12: 2818316-2818316 on Build GRCh38
rs773949675 Chr.12: 2818300-2818300 on Build GRCh38
rs551874254 Chr.12: 2818205-2818205 on Build GRCh38
rs779653696 Chr.12: 2818081-2818081 on Build GRCh38
rs59252735 Chr.12: 2818089-2818089 on Build GRCh38
rs200177526 Chr.12: 2818313-2818313 on Build GRCh38
rs749897172 Chr.12: 2818072-2818072 on Build GRCh38
rs201588528 Chr.12: 2818164-2818164 on Build GRCh38
rs370306985 Chr.12: 2818057-2818057 on Build GRCh38
rs773118411 Chr.12: 2818291-2818291 on Build GRCh38
rs374998892 Chr.12: 2818276-2818276 on Build GRCh38
rs778216834 Chr.12: 2818086-2818086 on Build GRCh38
rs748443998 Chr.12: 2818280-2818280 on Build GRCh38
rs200519806 Chr.12: 2818284-2818284 on Build GRCh38

View Full Product Details