Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4336653-4337164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGAR
Entrez Gene ID
57103
Gene Name
TP53 induced glycolysis regulatory phosphatase
Gene Aliases
C12orf5, FR2BP
UniGene
Hs.504545
Gene Chromosome Location
Chr.12: 4321193-4360028 on Build GRCh38
Transcripts
NM_020375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4041879 c.158A>G p.H53R Chr.12: 4337126-4337126 on Build GRCh38
4645852 c.179T>C p.M60T Chr.12: 4337147-4337147 on Build GRCh38
228567 c.131T>C p.I44T Chr.12: 4337099-4337099 on Build GRCh38
2067323 c.150G>C p.K50N Chr.12: 4337118-4337118 on Build GRCh38
6614204 c.87A>C p.E29D Chr.12: 4337055-4337055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988161706 Chr.12: 4336770-4336770 on Build GRCh38
rs769255664 Chr.12: 4336797-4336799 on Build GRCh38
rs1049574364 Chr.12: 4337003-4337003 on Build GRCh38
rs904311650 Chr.12: 4337012-4337012 on Build GRCh38
rs748449031 Chr.12: 4337098-4337098 on Build GRCh38
rs367654834 Chr.12: 4336700-4336700 on Build GRCh38
rs200518489 Chr.12: 4337009-4337009 on Build GRCh38
rs1028186684 Chr.12: 4336867-4336867 on Build GRCh38
rs779823420 Chr.12: 4337089-4337089 on Build GRCh38
rs148681725 Chr.12: 4336964-4336964 on Build GRCh38
rs7296163 Chr.12: 4336950-4336950 on Build GRCh38
rs4765778 Chr.12: 4336871-4336871 on Build GRCh38
rs749970757 Chr.12: 4337073-4337073 on Build GRCh38
rs964208940 Chr.12: 4336734-4336734 on Build GRCh38
rs144640944 Chr.12: 4337004-4337004 on Build GRCh38
rs149505187 Chr.12: 4336743-4336743 on Build GRCh38
rs576364014 Chr.12: 4336725-4336725 on Build GRCh38
rs988400837 Chr.12: 4337083-4337083 on Build GRCh38
rs569054643 Chr.12: 4336891-4336891 on Build GRCh38
rs757965275 Chr.12: 4337086-4337086 on Build GRCh38
rs778288561 Chr.12: 4337112-4337112 on Build GRCh38
rs757189604 Chr.12: 4337054-4337054 on Build GRCh38
rs778201803 Chr.12: 4337132-4337132 on Build GRCh38
rs749775941 Chr.12: 4337113-4337113 on Build GRCh38
rs986559490 Chr.12: 4336717-4336717 on Build GRCh38
rs935543893 Chr.12: 4336760-4336760 on Build GRCh38
rs778354019 Chr.12: 4337057-4337057 on Build GRCh38
rs764003646 Chr.12: 4337031-4337031 on Build GRCh38
rs59385818 Chr.12: 4336873-4336873 on Build GRCh38
rs11609492 Chr.12: 4336845-4336845 on Build GRCh38
rs1018030853 Chr.12: 4336996-4336996 on Build GRCh38
rs373191225 Chr.12: 4337090-4337090 on Build GRCh38
rs527685145 Chr.12: 4336768-4336768 on Build GRCh38
rs753714896 Chr.12: 4337032-4337032 on Build GRCh38
rs1025761630 Chr.12: 4336917-4336917 on Build GRCh38
rs377039834 Chr.12: 4336683-4336683 on Build GRCh38
rs1000304076 Chr.12: 4336711-4336711 on Build GRCh38
rs866352651 Chr.12: 4336890-4336890 on Build GRCh38
rs370764273 Chr.12: 4337055-4337055 on Build GRCh38
rs144179559 Chr.12: 4336771-4336771 on Build GRCh38
rs949536488 Chr.12: 4336807-4336807 on Build GRCh38
rs139173144 Chr.12: 4336710-4336710 on Build GRCh38
rs767833226 Chr.12: 4337000-4337000 on Build GRCh38
rs970654648 Chr.12: 4336843-4336843 on Build GRCh38
rs4765777 Chr.12: 4336756-4336756 on Build GRCh38
rs561451137 Chr.12: 4336853-4336853 on Build GRCh38
rs976657483 Chr.12: 4336931-4336931 on Build GRCh38
rs375097199 Chr.12: 4337019-4337019 on Build GRCh38
rs749947980 Chr.12: 4336910-4336910 on Build GRCh38
rs7306862 Chr.12: 4336911-4336911 on Build GRCh38
rs771591594 Chr.12: 4337115-4337115 on Build GRCh38
rs533791943 Chr.12: 4336971-4336971 on Build GRCh38
rs866793687 Chr.12: 4337059-4337059 on Build GRCh38
rs1049966493 Chr.12: 4336702-4336702 on Build GRCh38
rs756491476 Chr.12: 4337110-4337110 on Build GRCh38
rs553453481 Chr.12: 4336986-4336986 on Build GRCh38
rs956969334 Chr.12: 4337056-4337056 on Build GRCh38
rs979612864 Chr.12: 4337021-4337021 on Build GRCh38

View Full Product Details