Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4662341-4662765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA9
Entrez Gene ID
4704
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Gene Aliases
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1
UniGene
Hs.75227
Gene Chromosome Location
Chr.12: 4649098-4687554 on Build GRCh38
Transcripts
NM_005002.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
288561 c.628G>A p.E210K Chr.12: 4662608-4662608 on Build GRCh38
939667 c.633T>A p.D211E Chr.12: 4662613-4662613 on Build GRCh38
321974 c.574A>T p.R192* Chr.12: 4662554-4662554 on Build GRCh38
6368431 c.581C>A p.A194E Chr.12: 4662561-4662561 on Build GRCh38
6723696 c.608C>T p.P203L Chr.12: 4662588-4662588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766697773 Chr.12: 4662588-4662588 on Build GRCh38
rs138112681 Chr.12: 4662592-4662592 on Build GRCh38
rs2267549 Chr.12: 4662516-4662516 on Build GRCh38
rs138434279 Chr.12: 4662568-4662568 on Build GRCh38
rs536515528 Chr.12: 4662702-4662702 on Build GRCh38
rs143710385 Chr.12: 4662558-4662558 on Build GRCh38
rs763787589 Chr.12: 4662506-4662506 on Build GRCh38
rs149534005 Chr.12: 4662652-4662652 on Build GRCh38
rs2302245 Chr.12: 4662479-4662479 on Build GRCh38
rs759983004 Chr.12: 4662591-4662591 on Build GRCh38
rs760201093 Chr.12: 4662569-4662569 on Build GRCh38
rs768425350 Chr.12: 4662574-4662574 on Build GRCh38
rs776252587 Chr.12: 4662641-4662641 on Build GRCh38
rs760279506 Chr.12: 4662695-4662695 on Build GRCh38
rs569362628 Chr.12: 4662737-4662737 on Build GRCh38
rs181781262 Chr.12: 4662466-4662466 on Build GRCh38
rs150765610 Chr.12: 4662524-4662524 on Build GRCh38
rs767443077 Chr.12: 4662643-4662644 on Build GRCh38
rs766755488 Chr.12: 4662513-4662513 on Build GRCh38
rs778745677 Chr.12: 4662567-4662567 on Build GRCh38
rs961082748 Chr.12: 4662467-4662467 on Build GRCh38
rs913189015 Chr.12: 4662713-4662713 on Build GRCh38
rs752780525 Chr.12: 4662596-4662596 on Build GRCh38
rs990215436 Chr.12: 4662627-4662627 on Build GRCh38
rs749509666 Chr.12: 4662536-4662536 on Build GRCh38
rs863224085 Chr.12: 4662560-4662560 on Build GRCh38
rs756255858 Chr.12: 4662602-4662602 on Build GRCh38
rs878917965 Chr.12: 4662456-4662456 on Build GRCh38
rs769443764 Chr.12: 4662668-4662668 on Build GRCh38
rs755482936 Chr.12: 4662521-4662521 on Build GRCh38
rs758278017 Chr.12: 4662624-4662624 on Build GRCh38
rs768333416 Chr.12: 4662640-4662640 on Build GRCh38
rs1005122740 Chr.12: 4662561-4662561 on Build GRCh38
rs370125773 Chr.12: 4662484-4662484 on Build GRCh38
rs75878894 Chr.12: 4662539-4662539 on Build GRCh38
rs777815546 Chr.12: 4662609-4662609 on Build GRCh38
rs566828763 Chr.12: 4662435-4662435 on Build GRCh38
rs745623102 Chr.12: 4662620-4662620 on Build GRCh38
rs777906100 Chr.12: 4662531-4662531 on Build GRCh38
rs939180172 Chr.12: 4662473-4662473 on Build GRCh38
rs866347845 Chr.12: 4662434-4662434 on Build GRCh38
rs771652214 Chr.12: 4662580-4662580 on Build GRCh38
rs946639431 Chr.12: 4662414-4662414 on Build GRCh38
rs372645808 Chr.12: 4662502-4662502 on Build GRCh38
rs199935545 Chr.12: 4662551-4662551 on Build GRCh38
rs185126760 Chr.12: 4662610-4662610 on Build GRCh38
rs976380237 Chr.12: 4662475-4662475 on Build GRCh38
rs751916214 Chr.12: 4662589-4662589 on Build GRCh38
rs201865179 Chr.12: 4662581-4662581 on Build GRCh38
rs940360863 Chr.12: 4662745-4662745 on Build GRCh38
rs915179758 Chr.12: 4662375-4662375 on Build GRCh38
rs923815707 Chr.12: 4662659-4662659 on Build GRCh38
rs201326871 Chr.12: 4662639-4662639 on Build GRCh38
rs923830681 Chr.12: 4662437-4662437 on Build GRCh38
rs953107888 Chr.12: 4662408-4662408 on Build GRCh38
rs201029973 Chr.12: 4662669-4662669 on Build GRCh38
rs757468784 Chr.12: 4662614-4662614 on Build GRCh38
rs750828601 Chr.12: 4662511-4662511 on Build GRCh38
rs947832905 Chr.12: 4662731-4662731 on Build GRCh38
rs767347095 Chr.12: 4662635-4662635 on Build GRCh38
rs766040100 Chr.12: 4662735-4662735 on Build GRCh38
rs200660398 Chr.12: 4662619-4662619 on Build GRCh38
rs763340151 Chr.12: 4662512-4662512 on Build GRCh38
rs775034211 Chr.12: 4662493-4662493 on Build GRCh38
rs756343672 Chr.12: 4662529-4662529 on Build GRCh38
rs992372663 Chr.12: 4662689-4662689 on Build GRCh38

View Full Product Details