Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7717328-7717801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA3
Entrez Gene ID
359787
Gene Name
developmental pluripotency associated 3
Gene Aliases
STELLA
UniGene
Hs.131358
Gene Chromosome Location
Chr.12: 7711454-7717559 on Build GRCh38
Transcripts
NM_199286.3

SNP Detail

SNP ID Chromosome Location
rs997255664 Chr.12: 7717427-7717427 on Build GRCh38
rs928972286 Chr.12: 7717713-7717713 on Build GRCh38
rs1021592305 Chr.12: 7717466-7717466 on Build GRCh38
rs533764585 Chr.12: 7717492-7717492 on Build GRCh38
rs1014912465 Chr.12: 7717627-7717627 on Build GRCh38
rs1043757192 Chr.12: 7717495-7717495 on Build GRCh38
rs796297332 Chr.12: 7717740-7717740 on Build GRCh38
rs1008716155 Chr.12: 7717379-7717379 on Build GRCh38
rs910923426 Chr.12: 7717476-7717476 on Build GRCh38
rs985664085 Chr.12: 7717726-7717726 on Build GRCh38
rs769000287 Chr.12: 7717586-7717589 on Build GRCh38
rs1038844420 Chr.12: 7717750-7717750 on Build GRCh38
rs887827110 Chr.12: 7717608-7717608 on Build GRCh38
rs994075995 Chr.12: 7717651-7717651 on Build GRCh38
rs553389025 Chr.12: 7717517-7717517 on Build GRCh38
rs890326911 Chr.12: 7717370-7717370 on Build GRCh38
rs796947611 Chr.12: 7717375-7717375 on Build GRCh38
rs962066141 Chr.12: 7717639-7717639 on Build GRCh38
rs1023810142 Chr.12: 7717593-7717593 on Build GRCh38
rs2286947 Chr.12: 7717621-7717621 on Build GRCh38
rs955275370 Chr.12: 7717681-7717681 on Build GRCh38
rs985223727 Chr.12: 7717422-7717422 on Build GRCh38
rs896652809 Chr.12: 7717539-7717539 on Build GRCh38
rs776799035 Chr.12: 7717516-7717516 on Build GRCh38
rs1029623668 Chr.12: 7717666-7717666 on Build GRCh38
rs34349468 Chr.12: 7717667-7717668 on Build GRCh38
rs545694828 Chr.12: 7717729-7717729 on Build GRCh38
rs574473896 Chr.12: 7717485-7717485 on Build GRCh38
rs972666610 Chr.12: 7717391-7717391 on Build GRCh38
rs920295765 Chr.12: 7717694-7717694 on Build GRCh38
rs796123395 Chr.12: 7717460-7717460 on Build GRCh38
rs375579301 Chr.12: 7717722-7717722 on Build GRCh38
rs922415258 Chr.12: 7717396-7717396 on Build GRCh38
rs890192702 Chr.12: 7717735-7717735 on Build GRCh38
rs964631129 Chr.12: 7717415-7717415 on Build GRCh38
rs902206549 Chr.12: 7717505-7717505 on Build GRCh38
rs749655337 Chr.12: 7717547-7717547 on Build GRCh38
rs961258569 Chr.12: 7717368-7717368 on Build GRCh38
rs911070199 Chr.12: 7717739-7717739 on Build GRCh38
rs748640331 Chr.12: 7717775-7717775 on Build GRCh38
rs531890926 Chr.12: 7717721-7717721 on Build GRCh38
rs1012870785 Chr.12: 7717554-7717554 on Build GRCh38
rs933845839 Chr.12: 7717399-7717399 on Build GRCh38
rs1056671689 Chr.12: 7717520-7717520 on Build GRCh38
rs948960004 Chr.12: 7717575-7717577 on Build GRCh38
rs200516222 Chr.12: 7717402-7717402 on Build GRCh38
rs992981043 Chr.12: 7717517-7717518 on Build GRCh38

View Full Product Details