Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10185209-10185626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM52B
Entrez Gene ID
120939
Gene Name
transmembrane protein 52B
Gene Aliases
C12orf59
UniGene
Hs.226422
Gene Chromosome Location
Chr.12: 10170599-10191804 on Build GRCh38
Transcripts
NM_001079815.1 NM_153022.3 XM_006719029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934123 c.52G>T p.D18Y Chr.12: 10185343-10185343 on Build GRCh38
6519970 c.118G>A p.V40I Chr.12: 10185349-10185349 on Build GRCh38
6135538 c.62A>G p.H21R Chr.12: 10185353-10185353 on Build GRCh38
691578 c.76T>C p.W26R Chr.12: 10185367-10185367 on Build GRCh38
6135539 c.122A>G p.H41R Chr.12: 10185353-10185353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748463489 Chr.12: 10185451-10185451 on Build GRCh38
rs993748769 Chr.12: 10185487-10185487 on Build GRCh38
rs752127498 Chr.12: 10185279-10185279 on Build GRCh38
rs897223714 Chr.12: 10185442-10185442 on Build GRCh38
rs762477021 Chr.12: 10185289-10185289 on Build GRCh38
rs925545839 Chr.12: 10185382-10185382 on Build GRCh38
rs369785406 Chr.12: 10185303-10185303 on Build GRCh38
rs559455566 Chr.12: 10185524-10185524 on Build GRCh38
rs919599786 Chr.12: 10185387-10185387 on Build GRCh38
rs1000750203 Chr.12: 10185498-10185498 on Build GRCh38
rs774264160 Chr.12: 10185407-10185407 on Build GRCh38
rs779988880 Chr.12: 10185401-10185401 on Build GRCh38
rs367931481 Chr.12: 10185590-10185590 on Build GRCh38
rs1015173860 Chr.12: 10185564-10185564 on Build GRCh38
rs563350587 Chr.12: 10185266-10185266 on Build GRCh38
rs1012297183 Chr.12: 10185532-10185532 on Build GRCh38
rs1021137897 Chr.12: 10185585-10185585 on Build GRCh38
rs183338554 Chr.12: 10185274-10185274 on Build GRCh38
rs770437566 Chr.12: 10185401-10185402 on Build GRCh38
rs143277263 Chr.12: 10185595-10185595 on Build GRCh38
rs913634026 Chr.12: 10185355-10185355 on Build GRCh38
rs1032319030 Chr.12: 10185526-10185526 on Build GRCh38
rs878919778 Chr.12: 10185542-10185542 on Build GRCh38
rs939650654 Chr.12: 10185247-10185247 on Build GRCh38
rs756958883 Chr.12: 10185340-10185340 on Build GRCh38
rs979596923 Chr.12: 10185368-10185368 on Build GRCh38
rs902467138 Chr.12: 10185488-10185488 on Build GRCh38
rs751148460 Chr.12: 10185326-10185326 on Build GRCh38
rs1008054326 Chr.12: 10185431-10185431 on Build GRCh38
rs780959435 Chr.12: 10185344-10185344 on Build GRCh38
rs983645208 Chr.12: 10185546-10185546 on Build GRCh38
rs770137858 Chr.12: 10185565-10185565 on Build GRCh38
rs749211331 Chr.12: 10185408-10185408 on Build GRCh38
rs896522430 Chr.12: 10185425-10185425 on Build GRCh38
rs780856456 Chr.12: 10185367-10185367 on Build GRCh38
rs1049945337 Chr.12: 10185416-10185416 on Build GRCh38
rs938228740 Chr.12: 10185383-10185383 on Build GRCh38
rs970919880 Chr.12: 10185545-10185545 on Build GRCh38
rs973670995 Chr.12: 10185584-10185584 on Build GRCh38
rs1027465655 Chr.12: 10185503-10185503 on Build GRCh38
rs528565898 Chr.12: 10185531-10185531 on Build GRCh38
rs750271310 Chr.12: 10185353-10185353 on Build GRCh38
rs558866666 Chr.12: 10185411-10185411 on Build GRCh38
rs890927812 Chr.12: 10185420-10185420 on Build GRCh38

View Full Product Details