Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22472052-22472565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD5
Entrez Gene ID
9847
Gene Name
C2 calcium dependent domain containing 5
Gene Aliases
CDP138, KIAA0528
UniGene
Hs.271014
Gene Chromosome Location
Chr.12: 22448546-22544546 on Build GRCh38
Transcripts
NM_001286173.1 NM_001286174.1 NM_001286175.1 NM_001286176.1 NM_001286177.1 NM_014802.2 XM_005253538.2 XM_005253539.3 XM_006719170.2 XM_017020264.1 XM_017020265.1 XM_017020266.1 XM_017020267.1 XM_017020268.1 XM_017020269.1 XM_017020270.1 XM_017020271.1 XM_017020272.1 XM_017020273.1 XM_017020274.1 XM_017020275.1 XM_017020276.1 XM_017020277.1 XM_017020278.1 XM_017020279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4477479 c.1602C>T p.T534T Chr.12: 22472298-22472298 on Build GRCh38
5170012 c.1553-7T>G p.? Chr.12: 22472354-22472354 on Build GRCh38
4477478 c.2157C>T p.T719T Chr.12: 22472298-22472298 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535220853 Chr.12: 22472158-22472158 on Build GRCh38
rs910798408 Chr.12: 22472233-22472233 on Build GRCh38
rs977823698 Chr.12: 22472162-22472162 on Build GRCh38
rs570843029 Chr.12: 22472516-22472516 on Build GRCh38
rs749283585 Chr.12: 22472388-22472388 on Build GRCh38
rs766527258 Chr.12: 22472302-22472302 on Build GRCh38
rs554996750 Chr.12: 22472168-22472168 on Build GRCh38
rs756981827 Chr.12: 22472350-22472350 on Build GRCh38
rs368955461 Chr.12: 22472245-22472245 on Build GRCh38
rs760692248 Chr.12: 22472273-22472273 on Build GRCh38
rs937138570 Chr.12: 22472196-22472196 on Build GRCh38
rs562201710 Chr.12: 22472467-22472467 on Build GRCh38
rs1014830341 Chr.12: 22472419-22472419 on Build GRCh38
rs755622091 Chr.12: 22472375-22472375 on Build GRCh38
rs16924983 Chr.12: 22472304-22472304 on Build GRCh38
rs375777831 Chr.12: 22472380-22472380 on Build GRCh38
rs548400337 Chr.12: 22472461-22472461 on Build GRCh38
rs765762043 Chr.12: 22472317-22472317 on Build GRCh38
rs879225408 Chr.12: 22472202-22472202 on Build GRCh38
rs933660533 Chr.12: 22472187-22472187 on Build GRCh38
rs760064122 Chr.12: 22472305-22472305 on Build GRCh38
rs927020200 Chr.12: 22472185-22472185 on Build GRCh38
rs775321843 Chr.12: 22472224-22472224 on Build GRCh38
rs747844140 Chr.12: 22472148-22472148 on Build GRCh38
rs548237076 Chr.12: 22472271-22472271 on Build GRCh38
rs751671527 Chr.12: 22472247-22472248 on Build GRCh38
rs373390040 Chr.12: 22472201-22472202 on Build GRCh38
rs556512775 Chr.12: 22472123-22472123 on Build GRCh38
rs946225006 Chr.12: 22472234-22472235 on Build GRCh38
rs779994120 Chr.12: 22472088-22472088 on Build GRCh38
rs771586547 Chr.12: 22472228-22472228 on Build GRCh38
rs780727140 Chr.12: 22472363-22472363 on Build GRCh38
rs1057482901 Chr.12: 22472200-22472201 on Build GRCh38
rs16924981 Chr.12: 22472132-22472132 on Build GRCh38
rs926200821 Chr.12: 22472516-22472517 on Build GRCh38
rs772156840 Chr.12: 22472252-22472252 on Build GRCh38
rs878994072 Chr.12: 22472093-22472093 on Build GRCh38
rs752971808 Chr.12: 22472346-22472346 on Build GRCh38
rs771476875 Chr.12: 22472528-22472528 on Build GRCh38
rs768579011 Chr.12: 22472398-22472398 on Build GRCh38
rs992555072 Chr.12: 22472476-22472476 on Build GRCh38
rs927017384 Chr.12: 22472454-22472454 on Build GRCh38
rs895043622 Chr.12: 22472447-22472447 on Build GRCh38
rs1047864100 Chr.12: 22472321-22472321 on Build GRCh38
rs374464335 Chr.12: 22472086-22472086 on Build GRCh38
rs189750753 Chr.12: 22472406-22472406 on Build GRCh38
rs202165258 Chr.12: 22472341-22472341 on Build GRCh38
rs973468366 Chr.12: 22472453-22472453 on Build GRCh38
rs899800418 Chr.12: 22472539-22472539 on Build GRCh38
rs1022098916 Chr.12: 22472509-22472509 on Build GRCh38
rs199759626 Chr.12: 22472258-22472258 on Build GRCh38
rs954501292 Chr.12: 22472402-22472402 on Build GRCh38
rs989323765 Chr.12: 22472126-22472126 on Build GRCh38
rs749154483 Chr.12: 22472110-22472110 on Build GRCh38
rs958617882 Chr.12: 22472455-22472455 on Build GRCh38
rs16924984 Chr.12: 22472481-22472481 on Build GRCh38
rs1040364784 Chr.12: 22472526-22472526 on Build GRCh38
rs963316502 Chr.12: 22472453-22472454 on Build GRCh38
rs183913793 Chr.12: 22472237-22472237 on Build GRCh38
rs540668234 Chr.12: 22472249-22472249 on Build GRCh38
rs750048468 Chr.12: 22472371-22472371 on Build GRCh38
rs529036598 Chr.12: 22472420-22472420 on Build GRCh38
rs879368102 Chr.12: 22472517-22472517 on Build GRCh38
rs11046438 Chr.12: 22472513-22472513 on Build GRCh38
rs906496896 Chr.12: 22472330-22472330 on Build GRCh38
rs899914013 Chr.12: 22472323-22472323 on Build GRCh38
rs766626717 Chr.12: 22472080-22472084 on Build GRCh38

View Full Product Details