Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31096577-31097083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX11
Entrez Gene ID
1663
Gene Name
DEAD/H-box helicase 11
Gene Aliases
CHL1, CHLR1, KRG2, WABS
UniGene
Hs.443960
Gene Chromosome Location
Chr.12: 31073574-31104799 on Build GRCh38
Transcripts
NM_001257144.1 NM_001257145.1 NM_004399.2 NM_030653.3 NM_152438.1 XM_005253331.3 XM_005253333.3 XM_006719050.3 XM_011520592.2 XM_011520595.2 XM_011520597.2 XM_017018918.1 XM_017018919.1 XM_017018920.1 XM_017018921.1 XM_017018922.1 XM_017018923.1 XM_017018924.1 XM_017018925.1 XM_017018926.1 XM_017018927.1 XM_017018928.1 XM_017018929.1 XM_017018930.1 XM_017018931.1 XM_017018932.1 XM_017018933.1 XM_017018934.1 XM_017018935.1 XM_017018936.1 XM_017018937.1 XM_017018938.1 XM_017018939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871472 c.1528G>A p.G510R Chr.12: 31096643-31096643 on Build GRCh38
6072503 c.1542_1543GT>TA p.Y515N Chr.12: 31096657-31096658 on Build GRCh38
2205327 c.1565G>A p.R522Q Chr.12: 31096680-31096680 on Build GRCh38
6641534 c.1545C>T p.Y515Y Chr.12: 31096660-31096660 on Build GRCh38
4041380 c.1546G>A p.G516R Chr.12: 31096661-31096661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781327101 Chr.12: 31096610-31096610 on Build GRCh38
rs752847188 Chr.12: 31096953-31096953 on Build GRCh38
rs769266160 Chr.12: 31096874-31096874 on Build GRCh38
rs765545968 Chr.12: 31096651-31096651 on Build GRCh38
rs1001464794 Chr.12: 31096721-31096721 on Build GRCh38
rs71455622 Chr.12: 31096774-31096774 on Build GRCh38
rs776815284 Chr.12: 31096947-31096947 on Build GRCh38
rs747023303 Chr.12: 31096607-31096607 on Build GRCh38
rs745784326 Chr.12: 31096901-31096901 on Build GRCh38
rs768373123 Chr.12: 31096615-31096615 on Build GRCh38
rs781301437 Chr.12: 31096686-31096686 on Build GRCh38
rs918502766 Chr.12: 31096917-31096917 on Build GRCh38
rs777925425 Chr.12: 31096703-31096703 on Build GRCh38
rs780382531 Chr.12: 31096808-31096808 on Build GRCh38
rs764433669 Chr.12: 31096770-31096770 on Build GRCh38
rs373961029 Chr.12: 31096691-31096691 on Build GRCh38
rs772865522 Chr.12: 31096625-31096625 on Build GRCh38
rs771980266 Chr.12: 31096694-31096694 on Build GRCh38
rs746969480 Chr.12: 31096821-31096821 on Build GRCh38
rs377608793 Chr.12: 31096642-31096642 on Build GRCh38
rs375363924 Chr.12: 31097039-31097039 on Build GRCh38
rs142312636 Chr.12: 31096924-31096924 on Build GRCh38
rs979818680 Chr.12: 31096801-31096801 on Build GRCh38
rs758308408 Chr.12: 31096811-31096811 on Build GRCh38
rs752044834 Chr.12: 31096674-31096674 on Build GRCh38
rs117287945 Chr.12: 31096839-31096839 on Build GRCh38
rs759146055 Chr.12: 31096737-31096737 on Build GRCh38
rs754109777 Chr.12: 31096809-31096809 on Build GRCh38
rs746094785 Chr.12: 31096614-31096614 on Build GRCh38
rs776024108 Chr.12: 31096745-31096746 on Build GRCh38
rs755996490 Chr.12: 31096965-31096965 on Build GRCh38
rs372110755 Chr.12: 31096843-31096843 on Build GRCh38
rs778605700 Chr.12: 31096985-31096985 on Build GRCh38
rs574295109 Chr.12: 31097047-31097047 on Build GRCh38
rs772176887 Chr.12: 31096680-31096680 on Build GRCh38
rs145547443 Chr.12: 31096963-31096963 on Build GRCh38
rs377010461 Chr.12: 31096602-31096602 on Build GRCh38
rs755206505 Chr.12: 31096880-31096880 on Build GRCh38
rs772537025 Chr.12: 31096997-31096997 on Build GRCh38
rs758908151 Chr.12: 31096661-31096661 on Build GRCh38
rs147653293 Chr.12: 31096943-31096943 on Build GRCh38
rs756008353 Chr.12: 31096888-31096888 on Build GRCh38
rs775843579 Chr.12: 31096745-31096745 on Build GRCh38
rs762331360 Chr.12: 31096879-31096880 on Build GRCh38
rs564015937 Chr.12: 31096782-31096782 on Build GRCh38
rs776970202 Chr.12: 31096955-31096956 on Build GRCh38
rs547161084 Chr.12: 31096840-31096840 on Build GRCh38
rs370140116 Chr.12: 31096775-31096775 on Build GRCh38
rs760876221 Chr.12: 31096871-31096871 on Build GRCh38
rs765606145 Chr.12: 31096599-31096599 on Build GRCh38
rs145344014 Chr.12: 31096930-31096930 on Build GRCh38
rs769004603 Chr.12: 31096774-31096775 on Build GRCh38
rs73290434 Chr.12: 31096608-31096608 on Build GRCh38
rs758855146 Chr.12: 31096603-31096603 on Build GRCh38
rs769419349 Chr.12: 31096738-31096738 on Build GRCh38
rs748728528 Chr.12: 31096982-31096982 on Build GRCh38
rs780574130 Chr.12: 31096826-31096826 on Build GRCh38
rs144315069 Chr.12: 31096680-31096680 on Build GRCh38
rs773809963 Chr.12: 31096935-31096935 on Build GRCh38
rs776860164 Chr.12: 31096833-31096835 on Build GRCh38
rs747490271 Chr.12: 31096773-31096774 on Build GRCh38
rs370811400 Chr.12: 31096679-31096679 on Build GRCh38
rs992432538 Chr.12: 31096764-31096764 on Build GRCh38
rs200507494 Chr.12: 31096621-31096621 on Build GRCh38
rs759745463 Chr.12: 31097035-31097035 on Build GRCh38
rs374221282 Chr.12: 31096837-31096837 on Build GRCh38
rs147386779 Chr.12: 31096984-31096984 on Build GRCh38
rs754162876 Chr.12: 31096878-31096878 on Build GRCh38
rs759191828 Chr.12: 31097054-31097054 on Build GRCh38
rs759207736 Chr.12: 31096650-31096650 on Build GRCh38
rs2075322 Chr.12: 31096927-31096927 on Build GRCh38
rs771501792 Chr.12: 31097033-31097033 on Build GRCh38
rs578019303 Chr.12: 31096951-31096951 on Build GRCh38
rs765892801 Chr.12: 31096973-31096973 on Build GRCh38
rs201180239 Chr.12: 31096900-31096900 on Build GRCh38
rs1001036793 Chr.12: 31096950-31096950 on Build GRCh38
rs185072266 Chr.12: 31096814-31096814 on Build GRCh38
rs375197179 Chr.12: 31096941-31096941 on Build GRCh38
rs3168411 Chr.12: 31096926-31096926 on Build GRCh38
rs756306472 Chr.12: 31096786-31096786 on Build GRCh38
rs746912368 Chr.12: 31096723-31096723 on Build GRCh38
rs770400902 Chr.12: 31096724-31096724 on Build GRCh38
rs149627310 Chr.12: 31096660-31096660 on Build GRCh38
rs543298119 Chr.12: 31097051-31097051 on Build GRCh38
rs772070125 Chr.12: 31096623-31096623 on Build GRCh38
rs770865342 Chr.12: 31096823-31096823 on Build GRCh38
rs757152452 Chr.12: 31096890-31096890 on Build GRCh38
rs751029831 Chr.12: 31097002-31097002 on Build GRCh38
rs747726783 Chr.12: 31096986-31096986 on Build GRCh38
rs767998920 Chr.12: 31096855-31096855 on Build GRCh38
rs371449048 Chr.12: 31096966-31096966 on Build GRCh38
rs17857386 Chr.12: 31096952-31096952 on Build GRCh38
rs368696054 Chr.12: 31096842-31096842 on Build GRCh38
rs756309950 Chr.12: 31096692-31096692 on Build GRCh38
rs11545334 Chr.12: 31096864-31096864 on Build GRCh38
rs150142916 Chr.12: 31096958-31096958 on Build GRCh38
rs774170602 Chr.12: 31096618-31096618 on Build GRCh38
rs201232228 Chr.12: 31096634-31096634 on Build GRCh38
rs747587361 Chr.12: 31097014-31097014 on Build GRCh38
rs745323011 Chr.12: 31096834-31096834 on Build GRCh38
rs764523679 Chr.12: 31096672-31096672 on Build GRCh38
rs561628163 Chr.12: 31096644-31096644 on Build GRCh38
rs763362966 Chr.12: 31096746-31096746 on Build GRCh38
rs140857856 Chr.12: 31096954-31096954 on Build GRCh38
rs973855740 Chr.12: 31097059-31097059 on Build GRCh38
rs535328861 Chr.12: 31096885-31096885 on Build GRCh38
rs755685762 Chr.12: 31096920-31096920 on Build GRCh38
rs12322122 Chr.12: 31096921-31096921 on Build GRCh38
rs962810711 Chr.12: 31097050-31097050 on Build GRCh38
rs776161116 Chr.12: 31096728-31096728 on Build GRCh38

View Full Product Details