Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48823596-48824088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB3
Entrez Gene ID
784
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
Gene Aliases
CAB3, CACNLB3
UniGene
Hs.250712
Gene Chromosome Location
Chr.12: 48814432-48828943 on Build GRCh38
Transcripts
NM_000725.3 NM_001206915.1 NM_001206916.1 NM_001206917.1 XM_011538727.2 XM_011538728.2 XM_017019957.1 XM_017019958.1 XM_017019959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1361914 c.271G>T p.D91Y Chr.12: 48823783-48823783 on Build GRCh38
939896 c.215T>C p.V72A Chr.12: 48823730-48823730 on Build GRCh38
3396075 c.199G>A p.V67I Chr.12: 48823714-48823714 on Build GRCh38
6279168 c.217C>A p.L73M Chr.12: 48823732-48823732 on Build GRCh38
939898 c.230G>A p.C77Y Chr.12: 48823745-48823745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748191475 Chr.12: 48823755-48823755 on Build GRCh38
rs564818980 Chr.12: 48823641-48823641 on Build GRCh38
rs778984960 Chr.12: 48823746-48823746 on Build GRCh38
rs780219348 Chr.12: 48823948-48823948 on Build GRCh38
rs760172329 Chr.12: 48823662-48823662 on Build GRCh38
rs1053532414 Chr.12: 48823661-48823661 on Build GRCh38
rs750225184 Chr.12: 48823845-48823845 on Build GRCh38
rs889726378 Chr.12: 48823663-48823663 on Build GRCh38
rs981669039 Chr.12: 48823741-48823741 on Build GRCh38
rs536052641 Chr.12: 48823809-48823809 on Build GRCh38
rs763762133 Chr.12: 48823810-48823810 on Build GRCh38
rs746290663 Chr.12: 48823646-48823646 on Build GRCh38
rs147257060 Chr.12: 48823794-48823794 on Build GRCh38
rs770350566 Chr.12: 48823929-48823929 on Build GRCh38
rs771528377 Chr.12: 48823651-48823651 on Build GRCh38
rs1040298157 Chr.12: 48824004-48824004 on Build GRCh38
rs773568601 Chr.12: 48823642-48823642 on Build GRCh38
rs765207919 Chr.12: 48823817-48823818 on Build GRCh38
rs993057978 Chr.12: 48823943-48823943 on Build GRCh38
rs750349650 Chr.12: 48823728-48823728 on Build GRCh38
rs531589814 Chr.12: 48823780-48823780 on Build GRCh38
rs915032996 Chr.12: 48823918-48823918 on Build GRCh38
rs766251447 Chr.12: 48823852-48823852 on Build GRCh38
rs762597329 Chr.12: 48823702-48823702 on Build GRCh38
rs777892321 Chr.12: 48823778-48823778 on Build GRCh38
rs562938723 Chr.12: 48823659-48823659 on Build GRCh38
rs1128340 Chr.12: 48823691-48823691 on Build GRCh38
rs756112767 Chr.12: 48823732-48823732 on Build GRCh38
rs970832487 Chr.12: 48823995-48823995 on Build GRCh38
rs900410130 Chr.12: 48824045-48824045 on Build GRCh38
rs542061443 Chr.12: 48823898-48823898 on Build GRCh38
rs773959698 Chr.12: 48823819-48823819 on Build GRCh38
rs760603175 Chr.12: 48823850-48823850 on Build GRCh38
rs771403497 Chr.12: 48823786-48823786 on Build GRCh38
rs377133522 Chr.12: 48823804-48823804 on Build GRCh38
rs546343163 Chr.12: 48824044-48824044 on Build GRCh38
rs767296329 Chr.12: 48823829-48823829 on Build GRCh38
rs944639617 Chr.12: 48823975-48823975 on Build GRCh38
rs373224801 Chr.12: 48823721-48823721 on Build GRCh38
rs200536434 Chr.12: 48823633-48823633 on Build GRCh38
rs923568361 Chr.12: 48823966-48823966 on Build GRCh38
rs777191452 Chr.12: 48823792-48823792 on Build GRCh38
rs140514453 Chr.12: 48823797-48823797 on Build GRCh38
rs767338902 Chr.12: 48823722-48823722 on Build GRCh38
rs764788635 Chr.12: 48823682-48823682 on Build GRCh38
rs958958146 Chr.12: 48823940-48823940 on Build GRCh38
rs761501706 Chr.12: 48823826-48823826 on Build GRCh38
rs936547240 Chr.12: 48823654-48823654 on Build GRCh38
rs746315237 Chr.12: 48823708-48823708 on Build GRCh38
rs917594556 Chr.12: 48823957-48823957 on Build GRCh38
rs748398980 Chr.12: 48823632-48823632 on Build GRCh38
rs751306110 Chr.12: 48823712-48823712 on Build GRCh38
rs376386774 Chr.12: 48823743-48823743 on Build GRCh38
rs774926593 Chr.12: 48823700-48823700 on Build GRCh38
rs901128138 Chr.12: 48823729-48823729 on Build GRCh38
rs758602315 Chr.12: 48823777-48823777 on Build GRCh38
rs777250733 Chr.12: 48823660-48823660 on Build GRCh38
rs145046377 Chr.12: 48823718-48823718 on Build GRCh38
rs202047654 Chr.12: 48823647-48823647 on Build GRCh38

View Full Product Details