Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50085385-50085925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCD1
Entrez Gene ID
6602
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Gene Aliases
BAF60A, CRACD1, Rsc6p
UniGene
Hs.79335
Gene Chromosome Location
Chr.12: 50085200-50100711 on Build GRCh38
Transcripts
NM_003076.4 NM_139071.2 XM_005269107.3 XM_011538695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6909874 c.136C>G p.Q46E Chr.12: 50085505-50085505 on Build GRCh38
6929995 c.65_66insG p.A24fs*107 Chr.12: 50085434-50085435 on Build GRCh38
6913694 c.60+1G>A p.? Chr.12: 50085547-50085547 on Build GRCh38
6922737 c.26C>A p.S9Y Chr.12: 50085395-50085395 on Build GRCh38
6920340 c.116G>A p.R39Q Chr.12: 50085485-50085485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040041634 Chr.12: 50085682-50085682 on Build GRCh38
rs553744546 Chr.12: 50085659-50085659 on Build GRCh38
rs71464994 Chr.12: 50085473-50085473 on Build GRCh38
rs761283519 Chr.12: 50085569-50085569 on Build GRCh38
rs773406919 Chr.12: 50085568-50085568 on Build GRCh38
rs774710402 Chr.12: 50085475-50085475 on Build GRCh38
rs780226982 Chr.12: 50085423-50085423 on Build GRCh38
rs997929554 Chr.12: 50085831-50085831 on Build GRCh38
rs551623873 Chr.12: 50085618-50085618 on Build GRCh38
rs976962586 Chr.12: 50085500-50085500 on Build GRCh38
rs921169724 Chr.12: 50085690-50085693 on Build GRCh38
rs780138983 Chr.12: 50085705-50085705 on Build GRCh38
rs566006926 Chr.12: 50085601-50085601 on Build GRCh38
rs548902421 Chr.12: 50085511-50085511 on Build GRCh38
rs770736273 Chr.12: 50085420-50085420 on Build GRCh38
rs904193455 Chr.12: 50085772-50085772 on Build GRCh38
rs972167194 Chr.12: 50085617-50085617 on Build GRCh38
rs1013624066 Chr.12: 50085723-50085723 on Build GRCh38
rs749508764 Chr.12: 50085718-50085718 on Build GRCh38
rs746992940 Chr.12: 50085418-50085418 on Build GRCh38
rs971989793 Chr.12: 50085513-50085513 on Build GRCh38
rs985342530 Chr.12: 50085528-50085528 on Build GRCh38
rs189195538 Chr.12: 50085794-50085794 on Build GRCh38
rs749309509 Chr.12: 50085457-50085457 on Build GRCh38
rs772579523 Chr.12: 50085553-50085553 on Build GRCh38
rs956172058 Chr.12: 50085816-50085816 on Build GRCh38
rs386762920 Chr.12: 50085792-50085794 on Build GRCh38
rs924697497 Chr.12: 50085573-50085573 on Build GRCh38
rs956140872 Chr.12: 50085585-50085585 on Build GRCh38
rs570526816 Chr.12: 50085661-50085661 on Build GRCh38
rs1038106526 Chr.12: 50085702-50085702 on Build GRCh38
rs74090049 Chr.12: 50085715-50085715 on Build GRCh38
rs910587993 Chr.12: 50085540-50085540 on Build GRCh38
rs1037751284 Chr.12: 50085699-50085699 on Build GRCh38
rs941999997 Chr.12: 50085557-50085557 on Build GRCh38
rs78073502 Chr.12: 50085792-50085792 on Build GRCh38
rs556264672 Chr.12: 50085740-50085740 on Build GRCh38
rs769058585 Chr.12: 50085471-50085471 on Build GRCh38
rs990261320 Chr.12: 50085823-50085823 on Build GRCh38
rs566928546 Chr.12: 50085674-50085698 on Build GRCh38
rs555690370 Chr.12: 50085826-50085826 on Build GRCh38
rs528387075 Chr.12: 50085616-50085616 on Build GRCh38
rs1023640969 Chr.12: 50085720-50085720 on Build GRCh38
rs990228655 Chr.12: 50085606-50085606 on Build GRCh38
rs1013610690 Chr.12: 50085478-50085478 on Build GRCh38
rs943369400 Chr.12: 50085631-50085631 on Build GRCh38
rs572555543 Chr.12: 50085862-50085862 on Build GRCh38
rs919317813 Chr.12: 50085700-50085700 on Build GRCh38
rs954124678 Chr.12: 50085517-50085517 on Build GRCh38

View Full Product Details