Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52285825-52286319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT81
Entrez Gene ID
3887
Gene Name
keratin 81
Gene Aliases
HB1, Hb-1, KRTHB1, MLN137, ghHkb1, hHAKB2-1
UniGene
Hs.658118
Gene Chromosome Location
Chr.12: 52285913-52291515 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Gene Symbol
KRT86
Entrez Gene ID
3892
Gene Name
keratin 86
Gene Aliases
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX
UniGene
Hs.278658
Gene Chromosome Location
Chr.12: 52270507-52309163 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533565 c.1475C>T p.S492F Chr.12: 52286298-52286298 on Build GRCh38
1362452 c.1494C>T p.C498C Chr.12: 52286279-52286279 on Build GRCh38
5375387 c.1470C>T p.I490I Chr.12: 52286303-52286303 on Build GRCh38
6450410 c.1495G>A p.G499S Chr.12: 52286278-52286278 on Build GRCh38
4945528 c.1472G>A p.S491N Chr.12: 52286301-52286301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932814933 Chr.12: 52285985-52285985 on Build GRCh38
rs921550298 Chr.12: 52286227-52286227 on Build GRCh38
rs577321031 Chr.12: 52286069-52286069 on Build GRCh38
rs764882574 Chr.12: 52286089-52286089 on Build GRCh38
rs377741143 Chr.12: 52286294-52286294 on Build GRCh38
rs997329513 Chr.12: 52285872-52285872 on Build GRCh38
rs192117741 Chr.12: 52286119-52286119 on Build GRCh38
rs200814913 Chr.12: 52286279-52286279 on Build GRCh38
rs549423860 Chr.12: 52285853-52285853 on Build GRCh38
rs774184185 Chr.12: 52286230-52286230 on Build GRCh38
rs745373993 Chr.12: 52286219-52286219 on Build GRCh38
rs546389524 Chr.12: 52286072-52286072 on Build GRCh38
rs768552191 Chr.12: 52286229-52286229 on Build GRCh38
rs376061576 Chr.12: 52286266-52286266 on Build GRCh38
rs897508957 Chr.12: 52286076-52286076 on Build GRCh38
rs3657 Chr.12: 52286224-52286224 on Build GRCh38
rs757018912 Chr.12: 52286209-52286209 on Build GRCh38
rs546731827 Chr.12: 52286154-52286156 on Build GRCh38
rs201146279 Chr.12: 52286248-52286248 on Build GRCh38
rs79978767 Chr.12: 52285868-52285868 on Build GRCh38
rs1030681747 Chr.12: 52285885-52285885 on Build GRCh38
rs894391102 Chr.12: 52286130-52286130 on Build GRCh38
rs759047304 Chr.12: 52285952-52285952 on Build GRCh38
rs1021447718 Chr.12: 52286176-52286176 on Build GRCh38
rs935577474 Chr.12: 52286025-52286025 on Build GRCh38
rs138935547 Chr.12: 52286265-52286265 on Build GRCh38
rs542225476 Chr.12: 52286212-52286212 on Build GRCh38
rs1005913345 Chr.12: 52286045-52286045 on Build GRCh38
rs77315594 Chr.12: 52285867-52285867 on Build GRCh38
rs924586709 Chr.12: 52285929-52285929 on Build GRCh38
rs568720615 Chr.12: 52285930-52285930 on Build GRCh38
rs534191489 Chr.12: 52285953-52285953 on Build GRCh38
rs971324002 Chr.12: 52285948-52285948 on Build GRCh38
rs1048552722 Chr.12: 52286010-52286010 on Build GRCh38
rs899175230 Chr.12: 52286283-52286283 on Build GRCh38
rs780313674 Chr.12: 52286014-52286015 on Build GRCh38
rs887562214 Chr.12: 52286034-52286034 on Build GRCh38
rs371934116 Chr.12: 52286015-52286015 on Build GRCh38
rs76411635 Chr.12: 52285865-52285865 on Build GRCh38
rs968811428 Chr.12: 52286183-52286183 on Build GRCh38
rs554972437 Chr.12: 52285928-52285928 on Build GRCh38
rs776111472 Chr.12: 52285947-52285947 on Build GRCh38
rs968783655 Chr.12: 52285917-52285917 on Build GRCh38
rs377645057 Chr.12: 52286221-52286221 on Build GRCh38
rs113743804 Chr.12: 52285852-52285852 on Build GRCh38
rs932911400 Chr.12: 52286246-52286246 on Build GRCh38
rs977518732 Chr.12: 52286184-52286184 on Build GRCh38
rs77442466 Chr.12: 52285864-52285864 on Build GRCh38
rs187344718 Chr.12: 52285871-52285871 on Build GRCh38
rs540942003 Chr.12: 52286257-52286257 on Build GRCh38
rs540946161 Chr.12: 52285879-52285879 on Build GRCh38
rs112103702 Chr.12: 52285900-52285900 on Build GRCh38
rs763763828 Chr.12: 52286222-52286222 on Build GRCh38
rs1002460753 Chr.12: 52286101-52286101 on Build GRCh38
rs3660 Chr.12: 52286153-52286153 on Build GRCh38
rs747494104 Chr.12: 52285912-52285912 on Build GRCh38
rs554468774 Chr.12: 52286046-52286046 on Build GRCh38
rs773668893 Chr.12: 52285898-52285898 on Build GRCh38
rs921490495 Chr.12: 52285961-52285961 on Build GRCh38
rs759545377 Chr.12: 52286267-52286267 on Build GRCh38
rs914683224 Chr.12: 52285903-52285903 on Build GRCh38

View Full Product Details