Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52776655-52777143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT76
Entrez Gene ID
51350
Gene Name
keratin 76
Gene Aliases
HUMCYT2A, KRT2B, KRT2P
UniGene
Hs.654392
Gene Chromosome Location
Chr.12: 52768155-52777345 on Build GRCh38
Transcripts
NM_015848.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042922 c.517G>A p.D173N Chr.12: 52776775-52776775 on Build GRCh38
4575526 c.495G>A p.L165L Chr.12: 52776797-52776797 on Build GRCh38
4549259 c.468G>A p.Q156Q Chr.12: 52776824-52776824 on Build GRCh38
3462459 c.516C>T p.I172I Chr.12: 52776776-52776776 on Build GRCh38
1322423 c.459G>A p.G153G Chr.12: 52776833-52776833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761121693 Chr.12: 52776761-52776761 on Build GRCh38
rs1000072395 Chr.12: 52776846-52776846 on Build GRCh38
rs754470139 Chr.12: 52776947-52776947 on Build GRCh38
rs764340078 Chr.12: 52776933-52776933 on Build GRCh38
rs200858751 Chr.12: 52776979-52776979 on Build GRCh38
rs763134108 Chr.12: 52776920-52776920 on Build GRCh38
rs61730599 Chr.12: 52776814-52776814 on Build GRCh38
rs148421734 Chr.12: 52776881-52776881 on Build GRCh38
rs753751656 Chr.12: 52777029-52777029 on Build GRCh38
rs745396930 Chr.12: 52776737-52776737 on Build GRCh38
rs369803234 Chr.12: 52777117-52777117 on Build GRCh38
rs989223897 Chr.12: 52776847-52776847 on Build GRCh38
rs201649328 Chr.12: 52776680-52776680 on Build GRCh38
rs748806174 Chr.12: 52776879-52776879 on Build GRCh38
rs780616622 Chr.12: 52777061-52777061 on Build GRCh38
rs952432860 Chr.12: 52776760-52776760 on Build GRCh38
rs1019501318 Chr.12: 52776713-52776713 on Build GRCh38
rs756017618 Chr.12: 52776942-52776942 on Build GRCh38
rs775200433 Chr.12: 52777121-52777121 on Build GRCh38
rs779467774 Chr.12: 52776796-52776796 on Build GRCh38
rs757202648 Chr.12: 52776788-52776788 on Build GRCh38
rs755641756 Chr.12: 52776818-52776820 on Build GRCh38
rs760383184 Chr.12: 52777017-52777017 on Build GRCh38
rs752943841 Chr.12: 52776775-52776775 on Build GRCh38
rs536959890 Chr.12: 52776943-52776943 on Build GRCh38
rs935891850 Chr.12: 52776994-52776994 on Build GRCh38
rs1026907404 Chr.12: 52776932-52776932 on Build GRCh38
rs764688635 Chr.12: 52777038-52777038 on Build GRCh38
rs776902809 Chr.12: 52776895-52776895 on Build GRCh38
rs753839418 Chr.12: 52776946-52776946 on Build GRCh38
rs140231882 Chr.12: 52776954-52776954 on Build GRCh38
rs771302954 Chr.12: 52776893-52776893 on Build GRCh38
rs755078136 Chr.12: 52776769-52776769 on Build GRCh38
rs778442055 Chr.12: 52776949-52776949 on Build GRCh38
rs181600647 Chr.12: 52776794-52776794 on Build GRCh38
rs769264668 Chr.12: 52776738-52776738 on Build GRCh38
rs149286740 Chr.12: 52776820-52776820 on Build GRCh38
rs1042625698 Chr.12: 52776698-52776698 on Build GRCh38
rs755093620 Chr.12: 52776697-52776697 on Build GRCh38
rs781573301 Chr.12: 52776958-52776958 on Build GRCh38
rs536179360 Chr.12: 52777026-52777026 on Build GRCh38
rs770268869 Chr.12: 52776970-52776970 on Build GRCh38
rs762654173 Chr.12: 52776742-52776742 on Build GRCh38
rs756034287 Chr.12: 52776875-52776875 on Build GRCh38
rs759422989 Chr.12: 52777118-52777118 on Build GRCh38
rs777674254 Chr.12: 52776780-52776780 on Build GRCh38
rs748177156 Chr.12: 52777069-52777069 on Build GRCh38
rs116352440 Chr.12: 52776974-52776974 on Build GRCh38
rs377202243 Chr.12: 52776973-52776973 on Build GRCh38
rs775959284 Chr.12: 52776736-52776736 on Build GRCh38
rs552095996 Chr.12: 52776957-52776957 on Build GRCh38
rs778532065 Chr.12: 52776883-52776883 on Build GRCh38
rs773503735 Chr.12: 52776808-52776808 on Build GRCh38
rs746826620 Chr.12: 52776785-52776785 on Build GRCh38
rs61730593 Chr.12: 52777063-52777063 on Build GRCh38
rs752172537 Chr.12: 52777039-52777039 on Build GRCh38
rs575967584 Chr.12: 52777125-52777125 on Build GRCh38
rs753973524 Chr.12: 52776685-52776685 on Build GRCh38
rs761005852 Chr.12: 52777097-52777097 on Build GRCh38
rs758012608 Chr.12: 52777045-52777045 on Build GRCh38
rs746916248 Chr.12: 52776722-52776722 on Build GRCh38
rs777443354 Chr.12: 52776883-52776884 on Build GRCh38
rs767301204 Chr.12: 52776939-52776939 on Build GRCh38
rs372499307 Chr.12: 52776940-52776940 on Build GRCh38
rs371597584 Chr.12: 52777081-52777081 on Build GRCh38
rs749784108 Chr.12: 52777062-52777062 on Build GRCh38
rs781194470 Chr.12: 52776790-52776790 on Build GRCh38
rs747855077 Chr.12: 52776888-52776888 on Build GRCh38
rs368252592 Chr.12: 52776907-52776907 on Build GRCh38
rs780909422 Chr.12: 52776863-52776863 on Build GRCh38
rs771265763 Chr.12: 52776813-52776813 on Build GRCh38
rs745863620 Chr.12: 52776791-52776791 on Build GRCh38
rs148675815 Chr.12: 52776995-52776995 on Build GRCh38
rs138159446 Chr.12: 52776705-52776705 on Build GRCh38
rs750354939 Chr.12: 52776865-52776865 on Build GRCh38
rs142655432 Chr.12: 52776750-52776750 on Build GRCh38
rs751336286 Chr.12: 52776836-52776836 on Build GRCh38
rs146014128 Chr.12: 52776776-52776776 on Build GRCh38
rs966227657 Chr.12: 52776900-52776900 on Build GRCh38
rs770783035 Chr.12: 52776984-52777007 on Build GRCh38
rs766130047 Chr.12: 52777099-52777099 on Build GRCh38
rs199823023 Chr.12: 52777047-52777047 on Build GRCh38
rs371593117 Chr.12: 52776802-52776802 on Build GRCh38
rs746233114 Chr.12: 52776969-52776969 on Build GRCh38
rs780753874 Chr.12: 52776804-52776809 on Build GRCh38
rs771027498 Chr.12: 52776726-52776726 on Build GRCh38
rs889054165 Chr.12: 52777003-52777003 on Build GRCh38
rs757116734 Chr.12: 52776843-52776843 on Build GRCh38
rs775205869 Chr.12: 52776741-52776741 on Build GRCh38
rs1028167059 Chr.12: 52776744-52776744 on Build GRCh38
rs767834474 Chr.12: 52776743-52776743 on Build GRCh38
rs866786798 Chr.12: 52776877-52776877 on Build GRCh38
rs575273672 Chr.12: 52777082-52777082 on Build GRCh38
rs974183696 Chr.12: 52776867-52776884 on Build GRCh38
rs752221152 Chr.12: 52776814-52776815 on Build GRCh38
rs758251215 Chr.12: 52776708-52776708 on Build GRCh38
rs779565129 Chr.12: 52776876-52776876 on Build GRCh38
rs762044056 Chr.12: 52776983-52776983 on Build GRCh38
rs765174471 Chr.12: 52777119-52777119 on Build GRCh38
rs760049315 Chr.12: 52776897-52776897 on Build GRCh38
rs776440364 Chr.12: 52777102-52777102 on Build GRCh38
rs959198082 Chr.12: 52777044-52777044 on Build GRCh38
rs753789537 Chr.12: 52776767-52776767 on Build GRCh38
rs748896169 Chr.12: 52776797-52776797 on Build GRCh38
rs145056985 Chr.12: 52776938-52776938 on Build GRCh38
rs760007522 Chr.12: 52776816-52776816 on Build GRCh38
rs553174610 Chr.12: 52777007-52777007 on Build GRCh38
rs777762141 Chr.12: 52776716-52776716 on Build GRCh38
rs761993409 Chr.12: 52776936-52776936 on Build GRCh38
rs774511905 Chr.12: 52777067-52777067 on Build GRCh38
rs1040479320 Chr.12: 52777034-52777034 on Build GRCh38
rs749183675 Chr.12: 52776903-52776920 on Build GRCh38
rs752856882 Chr.12: 52776821-52776821 on Build GRCh38
rs764424088 Chr.12: 52776826-52776826 on Build GRCh38
rs878964906 Chr.12: 52777093-52777093 on Build GRCh38
rs775653286 Chr.12: 52776911-52776911 on Build GRCh38
rs761031890 Chr.12: 52776812-52776812 on Build GRCh38
rs779196533 Chr.12: 52776700-52776700 on Build GRCh38
rs772989693 Chr.12: 52777005-52777005 on Build GRCh38
rs901559448 Chr.12: 52777053-52777053 on Build GRCh38
rs992605418 Chr.12: 52776953-52776953 on Build GRCh38
rs1039219017 Chr.12: 52776724-52776724 on Build GRCh38

View Full Product Details