Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54648140-54648643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
UniGene
Hs.350570
Gene Chromosome Location
Chr.12: 54644591-54648493 on Build GRCh38
Transcripts
NM_001300854.1 NM_053283.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721793 c.24C>T p.F8F Chr.12: 54648280-54648280 on Build GRCh38
1747120 c.58T>A p.Y20N Chr.12: 54648246-54648246 on Build GRCh38
694623 c.57C>T p.A19A Chr.12: 54648247-54648247 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893667582 Chr.12: 54648403-54648403 on Build GRCh38
rs568656494 Chr.12: 54648489-54648489 on Build GRCh38
rs537035571 Chr.12: 54648188-54648188 on Build GRCh38
rs147479363 Chr.12: 54648255-54648255 on Build GRCh38
rs778304023 Chr.12: 54648316-54648316 on Build GRCh38
rs540831671 Chr.12: 54648221-54648221 on Build GRCh38
rs768989743 Chr.12: 54648216-54648216 on Build GRCh38
rs573293524 Chr.12: 54648167-54648167 on Build GRCh38
rs1032189057 Chr.12: 54648169-54648169 on Build GRCh38
rs975964995 Chr.12: 54648317-54648317 on Build GRCh38
rs182051287 Chr.12: 54648288-54648288 on Build GRCh38
rs772711180 Chr.12: 54648600-54648600 on Build GRCh38
rs1008542333 Chr.12: 54648451-54648451 on Build GRCh38
rs750067609 Chr.12: 54648298-54648298 on Build GRCh38
rs970610420 Chr.12: 54648597-54648597 on Build GRCh38
rs756880289 Chr.12: 54648204-54648204 on Build GRCh38
rs1029960739 Chr.12: 54648163-54648163 on Build GRCh38
rs143140702 Chr.12: 54648226-54648226 on Build GRCh38
rs1017301726 Chr.12: 54648458-54648458 on Build GRCh38
rs569577343 Chr.12: 54648520-54648520 on Build GRCh38
rs139941471 Chr.12: 54648286-54648286 on Build GRCh38
rs765009749 Chr.12: 54648413-54648413 on Build GRCh38
rs1012376403 Chr.12: 54648574-54648574 on Build GRCh38
rs949485161 Chr.12: 54648564-54648564 on Build GRCh38
rs78394705 Chr.12: 54648334-54648334 on Build GRCh38
rs144435466 Chr.12: 54648613-54648613 on Build GRCh38
rs61956600 Chr.12: 54648510-54648510 on Build GRCh38
rs1009362085 Chr.12: 54648622-54648622 on Build GRCh38
rs1054009235 Chr.12: 54648391-54648391 on Build GRCh38
rs767998531 Chr.12: 54648318-54648318 on Build GRCh38
rs60529685 Chr.12: 54648425-54648425 on Build GRCh38
rs200521753 Chr.12: 54648336-54648336 on Build GRCh38
rs761473082 Chr.12: 54648422-54648422 on Build GRCh38
rs975721708 Chr.12: 54648494-54648494 on Build GRCh38
rs529553936 Chr.12: 54648509-54648509 on Build GRCh38
rs776009938 Chr.12: 54648191-54648191 on Build GRCh38
rs771090043 Chr.12: 54648254-54648254 on Build GRCh38
rs112219304 Chr.12: 54648571-54648571 on Build GRCh38
rs752472469 Chr.12: 54648272-54648272 on Build GRCh38
rs370745463 Chr.12: 54648187-54648187 on Build GRCh38
rs1033618072 Chr.12: 54648528-54648528 on Build GRCh38
rs368995848 Chr.12: 54648227-54648227 on Build GRCh38
rs937912113 Chr.12: 54648599-54648599 on Build GRCh38
rs779257950 Chr.12: 54648305-54648305 on Build GRCh38
rs866939156 Chr.12: 54648341-54648341 on Build GRCh38
rs756756691 Chr.12: 54648291-54648291 on Build GRCh38
rs905121212 Chr.12: 54648165-54648165 on Build GRCh38
rs947453715 Chr.12: 54648437-54648439 on Build GRCh38
rs376739076 Chr.12: 54648330-54648330 on Build GRCh38
rs764041088 Chr.12: 54648284-54648284 on Build GRCh38
rs746091503 Chr.12: 54648338-54648338 on Build GRCh38
rs749748187 Chr.12: 54648276-54648276 on Build GRCh38
rs954444247 Chr.12: 54648446-54648446 on Build GRCh38
rs755405602 Chr.12: 54648300-54648300 on Build GRCh38
rs538886246 Chr.12: 54648593-54648593 on Build GRCh38
rs202162814 Chr.12: 54648565-54648565 on Build GRCh38
rs926667597 Chr.12: 54648585-54648585 on Build GRCh38
rs780852752 Chr.12: 54648211-54648211 on Build GRCh38
rs371654950 Chr.12: 54648293-54648293 on Build GRCh38
rs920082220 Chr.12: 54648507-54648507 on Build GRCh38
rs949349136 Chr.12: 54648340-54648340 on Build GRCh38
rs964341262 Chr.12: 54648315-54648315 on Build GRCh38
rs201094516 Chr.12: 54648278-54648278 on Build GRCh38
rs760759546 Chr.12: 54648244-54648244 on Build GRCh38
rs1002144556 Chr.12: 54648389-54648389 on Build GRCh38
rs372901613 Chr.12: 54648213-54648213 on Build GRCh38
rs11171000 Chr.12: 54648576-54648576 on Build GRCh38
rs931376575 Chr.12: 54648333-54648333 on Build GRCh38
rs898917475 Chr.12: 54648548-54648548 on Build GRCh38
rs368602620 Chr.12: 54648362-54648362 on Build GRCh38
rs759462379 Chr.12: 54648259-54648259 on Build GRCh38
rs1038867293 Chr.12: 54648473-54648473 on Build GRCh38
rs758854232 Chr.12: 54648311-54648311 on Build GRCh38
rs148716199 Chr.12: 54648164-54648164 on Build GRCh38
rs748456447 Chr.12: 54648309-54648309 on Build GRCh38
rs1037327260 Chr.12: 54648537-54648537 on Build GRCh38
rs1002139151 Chr.12: 54648505-54648505 on Build GRCh38
rs1053741298 Chr.12: 54648612-54648612 on Build GRCh38
rs186646965 Chr.12: 54648519-54648519 on Build GRCh38
rs981664263 Chr.12: 54648285-54648285 on Build GRCh38
rs528213283 Chr.12: 54648409-54648409 on Build GRCh38
rs554784879 Chr.12: 54648604-54648604 on Build GRCh38
rs747099940 Chr.12: 54648324-54648324 on Build GRCh38

View Full Product Details