Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57057153-57057657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEMP1
Entrez Gene ID
23306
Gene Name
nuclear envelope integral membrane protein 1
Gene Aliases
TMEM194, TMEM194A
UniGene
Hs.591040
Gene Chromosome Location
Chr.12: 57055643-57088703 on Build GRCh38
Transcripts
NM_001130963.1 NM_015257.2 XM_011538056.2 XM_011538057.2 XM_011538058.2 XM_011538059.1 XM_017019077.1 XM_017019078.1 XM_017019079.1

SNP Detail

SNP ID Chromosome Location
rs990892703 Chr.12: 57057283-57057283 on Build GRCh38
rs898836501 Chr.12: 57057534-57057534 on Build GRCh38
rs899119100 Chr.12: 57057264-57057264 on Build GRCh38
rs1004578711 Chr.12: 57057258-57057258 on Build GRCh38
rs1047570125 Chr.12: 57057374-57057374 on Build GRCh38
rs900096587 Chr.12: 57057433-57057433 on Build GRCh38
rs894297839 Chr.12: 57057566-57057566 on Build GRCh38
rs891321529 Chr.12: 57057417-57057417 on Build GRCh38
rs180727027 Chr.12: 57057470-57057470 on Build GRCh38
rs766725703 Chr.12: 57057488-57057488 on Build GRCh38
rs147326879 Chr.12: 57057180-57057180 on Build GRCh38
rs932154754 Chr.12: 57057435-57057435 on Build GRCh38
rs959901284 Chr.12: 57057568-57057568 on Build GRCh38
rs960132660 Chr.12: 57057235-57057235 on Build GRCh38
rs186105993 Chr.12: 57057536-57057536 on Build GRCh38
rs1011748797 Chr.12: 57057440-57057440 on Build GRCh38
rs143257630 Chr.12: 57057484-57057484 on Build GRCh38
rs542592830 Chr.12: 57057507-57057507 on Build GRCh38
rs944200577 Chr.12: 57057420-57057420 on Build GRCh38
rs533605639 Chr.12: 57057591-57057591 on Build GRCh38
rs140881728 Chr.12: 57057354-57057354 on Build GRCh38
rs919894132 Chr.12: 57057337-57057337 on Build GRCh38
rs970684217 Chr.12: 57057303-57057303 on Build GRCh38
rs1025509784 Chr.12: 57057292-57057292 on Build GRCh38
rs1022773552 Chr.12: 57057621-57057621 on Build GRCh38
rs529111559 Chr.12: 57057581-57057581 on Build GRCh38
rs1051507391 Chr.12: 57057452-57057452 on Build GRCh38
rs941889497 Chr.12: 57057277-57057277 on Build GRCh38
rs556771622 Chr.12: 57057312-57057312 on Build GRCh38
rs958248285 Chr.12: 57057336-57057336 on Build GRCh38
rs190704525 Chr.12: 57057242-57057242 on Build GRCh38
rs1042741948 Chr.12: 57057451-57057451 on Build GRCh38
rs931709838 Chr.12: 57057200-57057200 on Build GRCh38
rs911665435 Chr.12: 57057455-57057455 on Build GRCh38
rs561820927 Chr.12: 57057555-57057555 on Build GRCh38
rs1012759470 Chr.12: 57057596-57057596 on Build GRCh38
rs545459967 Chr.12: 57057447-57057447 on Build GRCh38
rs995172385 Chr.12: 57057265-57057265 on Build GRCh38
rs900675332 Chr.12: 57057179-57057179 on Build GRCh38
rs1034149434 Chr.12: 57057278-57057278 on Build GRCh38
rs1036062003 Chr.12: 57057481-57057481 on Build GRCh38
rs1014828870 Chr.12: 57057259-57057259 on Build GRCh38
rs760962137 Chr.12: 57057478-57057478 on Build GRCh38
rs754213496 Chr.12: 57057514-57057514 on Build GRCh38
rs1035553556 Chr.12: 57057202-57057202 on Build GRCh38
rs1039971728 Chr.12: 57057432-57057432 on Build GRCh38
rs922100074 Chr.12: 57057413-57057413 on Build GRCh38
rs958807692 Chr.12: 57057279-57057279 on Build GRCh38
rs977545952 Chr.12: 57057183-57057183 on Build GRCh38
rs1051968206 Chr.12: 57057234-57057234 on Build GRCh38
rs924187664 Chr.12: 57057329-57057329 on Build GRCh38

View Full Product Details