Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70638744-70639251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRR
Entrez Gene ID
5801
Gene Name
protein tyrosine phosphatase, receptor type R
Gene Aliases
EC-PTP, PCPTP1, PTP-SL, PTPBR7, PTPRQ
UniGene
Hs.506076
Gene Chromosome Location
Chr.12: 70638073-70920804 on Build GRCh38
Transcripts
NM_001207015.1 NM_001207016.1 NM_002849.3 NM_130846.2 NR_073474.1 XM_011538615.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955125 c.1912G>T p.E638* Chr.12: 70639246-70639246 on Build GRCh38
468875 c.1934G>A p.C645Y Chr.12: 70639224-70639224 on Build GRCh38
3772719 c.1971G>A p.Q657Q Chr.12: 70639187-70639187 on Build GRCh38
3464937 c.1951C>T p.L651F Chr.12: 70639207-70639207 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1043824405 Chr.12: 70638844-70638844 on Build GRCh38
rs755867495 Chr.12: 70639169-70639169 on Build GRCh38
rs376215111 Chr.12: 70639133-70639133 on Build GRCh38
rs916216746 Chr.12: 70638807-70638807 on Build GRCh38
rs985761646 Chr.12: 70638961-70638961 on Build GRCh38
rs991833631 Chr.12: 70639119-70639119 on Build GRCh38
rs951958082 Chr.12: 70638948-70638948 on Build GRCh38
rs1053262228 Chr.12: 70638920-70638920 on Build GRCh38
rs564197501 Chr.12: 70638939-70638939 on Build GRCh38
rs915459521 Chr.12: 70639066-70639066 on Build GRCh38
rs947750564 Chr.12: 70638825-70638825 on Build GRCh38
rs991885983 Chr.12: 70638780-70638780 on Build GRCh38
rs145029505 Chr.12: 70639008-70639008 on Build GRCh38
rs763595138 Chr.12: 70639123-70639123 on Build GRCh38
rs779044270 Chr.12: 70639195-70639195 on Build GRCh38
rs962174137 Chr.12: 70638872-70638872 on Build GRCh38
rs762418905 Chr.12: 70639144-70639144 on Build GRCh38
rs747400938 Chr.12: 70639229-70639229 on Build GRCh38
rs373469188 Chr.12: 70639157-70639157 on Build GRCh38
rs769143248 Chr.12: 70638805-70638805 on Build GRCh38
rs774846843 Chr.12: 70639001-70639001 on Build GRCh38
rs145800470 Chr.12: 70639211-70639211 on Build GRCh38
rs1064942 Chr.12: 70638775-70638775 on Build GRCh38
rs760171639 Chr.12: 70639024-70639024 on Build GRCh38
rs1035147000 Chr.12: 70638811-70638811 on Build GRCh38
rs969578854 Chr.12: 70639134-70639134 on Build GRCh38
rs1004032536 Chr.12: 70638785-70638785 on Build GRCh38
rs763903014 Chr.12: 70639226-70639227 on Build GRCh38
rs965780845 Chr.12: 70639012-70639012 on Build GRCh38
rs780640797 Chr.12: 70639226-70639226 on Build GRCh38
rs1053378446 Chr.12: 70639196-70639196 on Build GRCh38
rs1052564868 Chr.12: 70639038-70639038 on Build GRCh38
rs754066021 Chr.12: 70639185-70639185 on Build GRCh38
rs759459837 Chr.12: 70639152-70639152 on Build GRCh38
rs897928179 Chr.12: 70639164-70639164 on Build GRCh38
rs202121137 Chr.12: 70639147-70639147 on Build GRCh38
rs946971383 Chr.12: 70639073-70639073 on Build GRCh38
rs936280811 Chr.12: 70638901-70638901 on Build GRCh38
rs926219443 Chr.12: 70638896-70638897 on Build GRCh38
rs35935373 Chr.12: 70639057-70639058 on Build GRCh38
rs1045328019 Chr.12: 70639076-70639076 on Build GRCh38
rs376367611 Chr.12: 70639179-70639179 on Build GRCh38
rs976222958 Chr.12: 70639020-70639020 on Build GRCh38
rs771826412 Chr.12: 70638979-70638979 on Build GRCh38
rs759082873 Chr.12: 70638903-70638903 on Build GRCh38
rs1025405668 Chr.12: 70639068-70639068 on Build GRCh38
rs757318753 Chr.12: 70639194-70639194 on Build GRCh38
rs544263161 Chr.12: 70639098-70639098 on Build GRCh38
rs935472298 Chr.12: 70639026-70639026 on Build GRCh38
rs994067444 Chr.12: 70639067-70639067 on Build GRCh38
rs548236977 Chr.12: 70639204-70639204 on Build GRCh38
rs993367895 Chr.12: 70638889-70638889 on Build GRCh38
rs998242278 Chr.12: 70638952-70638952 on Build GRCh38

View Full Product Details