Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71439940-71440427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR5
Entrez Gene ID
8549
Gene Name
leucine rich repeat containing G protein-coupled receptor 5
Gene Aliases
FEX, GPR49, GPR67, GRP49, HG38
UniGene
Hs.658889
Gene Chromosome Location
Chr.12: 71439770-71586310 on Build GRCh38
Transcripts
NM_001277226.1 NM_001277227.1 NM_003667.3 NR_110596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243809 c.3G>T p.M1I Chr.12: 71440083-71440083 on Build GRCh38
6399847 c.119G>T p.C40F Chr.12: 71440199-71440199 on Build GRCh38
6398829 c.182C>T p.P61L Chr.12: 71440262-71440262 on Build GRCh38
4641772 c.99C>A p.G33G Chr.12: 71440179-71440179 on Build GRCh38
4488922 c.33C>T p.S11S Chr.12: 71440113-71440113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010293944 Chr.12: 71440003-71440003 on Build GRCh38
rs568020422 Chr.12: 71440206-71440206 on Build GRCh38
rs756666957 Chr.12: 71440172-71440172 on Build GRCh38
rs879109460 Chr.12: 71440177-71440177 on Build GRCh38
rs369629768 Chr.12: 71440328-71440328 on Build GRCh38
rs998978114 Chr.12: 71440377-71440377 on Build GRCh38
rs765147377 Chr.12: 71440099-71440099 on Build GRCh38
rs374681906 Chr.12: 71440033-71440033 on Build GRCh38
rs188717405 Chr.12: 71440016-71440016 on Build GRCh38
rs748575318 Chr.12: 71440053-71440053 on Build GRCh38
rs748663178 Chr.12: 71440113-71440113 on Build GRCh38
rs921516135 Chr.12: 71440093-71440093 on Build GRCh38
rs181180635 Chr.12: 71440245-71440245 on Build GRCh38
rs992680229 Chr.12: 71440058-71440058 on Build GRCh38
rs369292496 Chr.12: 71440334-71440334 on Build GRCh38
rs1036506346 Chr.12: 71440240-71440240 on Build GRCh38
rs375258674 Chr.12: 71440020-71440020 on Build GRCh38
rs775004100 Chr.12: 71440136-71440136 on Build GRCh38
rs1002817076 Chr.12: 71439989-71439989 on Build GRCh38
rs34598750 Chr.12: 71440302-71440303 on Build GRCh38
rs201521164 Chr.12: 71440042-71440042 on Build GRCh38
rs577202350 Chr.12: 71440002-71440002 on Build GRCh38
rs767050416 Chr.12: 71440067-71440067 on Build GRCh38
rs778241955 Chr.12: 71440178-71440178 on Build GRCh38
rs758686494 Chr.12: 71440102-71440102 on Build GRCh38
rs375498019 Chr.12: 71440373-71440373 on Build GRCh38
rs767796832 Chr.12: 71440343-71440343 on Build GRCh38
rs375163459 Chr.12: 71440255-71440255 on Build GRCh38
rs941746616 Chr.12: 71439960-71439960 on Build GRCh38
rs1024525695 Chr.12: 71440064-71440064 on Build GRCh38
rs140805486 Chr.12: 71440153-71440153 on Build GRCh38
rs951201050 Chr.12: 71440393-71440393 on Build GRCh38
rs768384590 Chr.12: 71440205-71440205 on Build GRCh38
rs1034264127 Chr.12: 71440044-71440044 on Build GRCh38
rs757247944 Chr.12: 71440074-71440074 on Build GRCh38
rs531257187 Chr.12: 71440167-71440167 on Build GRCh38
rs775117943 Chr.12: 71440068-71440068 on Build GRCh38
rs79836431 Chr.12: 71440043-71440043 on Build GRCh38
rs1000200685 Chr.12: 71440275-71440275 on Build GRCh38
rs894400841 Chr.12: 71440348-71440348 on Build GRCh38
rs768059416 Chr.12: 71440311-71440311 on Build GRCh38
rs768296709 Chr.12: 71440140-71440140 on Build GRCh38
rs773017722 Chr.12: 71440210-71440210 on Build GRCh38
rs1014558982 Chr.12: 71440387-71440387 on Build GRCh38
rs759781148 Chr.12: 71440223-71440223 on Build GRCh38
rs780115142 Chr.12: 71440105-71440105 on Build GRCh38
rs890551624 Chr.12: 71439973-71439973 on Build GRCh38
rs750279449 Chr.12: 71440148-71440148 on Build GRCh38
rs770857568 Chr.12: 71440330-71440330 on Build GRCh38
rs1035596936 Chr.12: 71439993-71439993 on Build GRCh38
rs753832960 Chr.12: 71440076-71440076 on Build GRCh38
rs781084434 Chr.12: 71440096-71440096 on Build GRCh38
rs201324217 Chr.12: 71440316-71440316 on Build GRCh38
rs746682823 Chr.12: 71440061-71440063 on Build GRCh38
rs73349178 Chr.12: 71440263-71440263 on Build GRCh38
rs765235346 Chr.12: 71440145-71440145 on Build GRCh38
rs1036216969 Chr.12: 71439974-71439974 on Build GRCh38
rs151098233 Chr.12: 71440325-71440325 on Build GRCh38
rs1031710940 Chr.12: 71440398-71440398 on Build GRCh38
rs373453153 Chr.12: 71440331-71440331 on Build GRCh38
rs757735108 Chr.12: 71440261-71440261 on Build GRCh38
rs139369662 Chr.12: 71440207-71440207 on Build GRCh38
rs770233037 Chr.12: 71440117-71440117 on Build GRCh38
rs558974898 Chr.12: 71439999-71439999 on Build GRCh38
rs768334189 Chr.12: 71440325-71440334 on Build GRCh38
rs544572456 Chr.12: 71440011-71440011 on Build GRCh38
rs769908352 Chr.12: 71440209-71440209 on Build GRCh38
rs1021648485 Chr.12: 71440360-71440360 on Build GRCh38
rs758499497 Chr.12: 71440034-71440034 on Build GRCh38
rs527854331 Chr.12: 71440059-71440059 on Build GRCh38
rs781268462 Chr.12: 71440169-71440169 on Build GRCh38
rs775049810 Chr.12: 71440239-71440239 on Build GRCh38
rs897641474 Chr.12: 71439982-71439982 on Build GRCh38
rs751732638 Chr.12: 71440154-71440154 on Build GRCh38
rs779176913 Chr.12: 71440265-71440265 on Build GRCh38
rs759317042 Chr.12: 71440339-71440339 on Build GRCh38
rs779282038 Chr.12: 71440191-71440191 on Build GRCh38
rs776232102 Chr.12: 71440141-71440141 on Build GRCh38
rs117324318 Chr.12: 71440257-71440257 on Build GRCh38
rs965557176 Chr.12: 71440404-71440404 on Build GRCh38
rs894517710 Chr.12: 71440019-71440019 on Build GRCh38
rs552023923 Chr.12: 71440151-71440151 on Build GRCh38
rs868578636 Chr.12: 71440238-71440238 on Build GRCh38
rs771817911 Chr.12: 71440132-71440132 on Build GRCh38
rs150864769 Chr.12: 71440201-71440201 on Build GRCh38
rs746213778 Chr.12: 71440284-71440284 on Build GRCh38
rs766098378 Chr.12: 71440211-71440211 on Build GRCh38
rs755005549 Chr.12: 71440111-71440111 on Build GRCh38
rs781180319 Chr.12: 71440112-71440112 on Build GRCh38
rs749598507 Chr.12: 71440131-71440131 on Build GRCh38
rs780976282 Chr.12: 71440303-71440303 on Build GRCh38
rs769253623 Chr.12: 71440046-71440046 on Build GRCh38
rs761384871 Chr.12: 71440208-71440208 on Build GRCh38
rs751316503 Chr.12: 71440219-71440219 on Build GRCh38
rs1024162108 Chr.12: 71440389-71440389 on Build GRCh38
rs759268679 Chr.12: 71440065-71440065 on Build GRCh38
rs760093215 Chr.12: 71440073-71440073 on Build GRCh38
rs141472970 Chr.12: 71440190-71440190 on Build GRCh38
rs747169147 Chr.12: 71440108-71440108 on Build GRCh38
rs376041503 Chr.12: 71440047-71440047 on Build GRCh38
rs951147297 Chr.12: 71440088-71440088 on Build GRCh38
rs147257938 Chr.12: 71440137-71440137 on Build GRCh38
rs371327811 Chr.12: 71440030-71440030 on Build GRCh38
rs1003951973 Chr.12: 71440403-71440403 on Build GRCh38
rs752746094 Chr.12: 71440171-71440171 on Build GRCh38
rs369732552 Chr.12: 71440055-71440055 on Build GRCh38
rs767708854 Chr.12: 71440228-71440228 on Build GRCh38

View Full Product Details