Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71566441-71566965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR5
Entrez Gene ID
8549
Gene Name
leucine rich repeat containing G protein-coupled receptor 5
Gene Aliases
FEX, GPR49, GPR67, GRP49, HG38
UniGene
Hs.658889
Gene Chromosome Location
Chr.12: 71439770-71586310 on Build GRCh38
Transcripts
NM_001277226.1 NM_001277227.1 NM_003667.3 NR_110596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166703 c.993G>A p.E331E Chr.12: 71566695-71566695 on Build GRCh38
7161221 c.1039G>T p.V347F Chr.12: 71566881-71566881 on Build GRCh38
6234847 c.950delA p.I318fs*1 Chr.12: 71566652-71566652 on Build GRCh38
1299851 c.1070+1G>A p.? Chr.12: 71566913-71566913 on Build GRCh38
7178677 c.1022C>T p.S341L Chr.12: 71566864-71566864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025305487 Chr.12: 71566568-71566568 on Build GRCh38
rs35021570 Chr.12: 71566677-71566678 on Build GRCh38
rs148218887 Chr.12: 71566509-71566509 on Build GRCh38
rs757464715 Chr.12: 71566740-71566740 on Build GRCh38
rs200459468 Chr.12: 71566507-71566507 on Build GRCh38
rs113031894 Chr.12: 71566763-71566763 on Build GRCh38
rs763925049 Chr.12: 71566727-71566727 on Build GRCh38
rs1797373 Chr.12: 71566549-71566549 on Build GRCh38
rs778633348 Chr.12: 71566942-71566942 on Build GRCh38
rs934314492 Chr.12: 71566874-71566874 on Build GRCh38
rs765346055 Chr.12: 71566873-71566873 on Build GRCh38
rs772016226 Chr.12: 71566911-71566911 on Build GRCh38
rs77632104 Chr.12: 71566621-71566621 on Build GRCh38
rs745462563 Chr.12: 71566943-71566943 on Build GRCh38
rs961356618 Chr.12: 71566616-71566616 on Build GRCh38
rs116452232 Chr.12: 71566878-71566878 on Build GRCh38
rs772703165 Chr.12: 71566940-71566942 on Build GRCh38
rs1002717343 Chr.12: 71566583-71566583 on Build GRCh38
rs773780730 Chr.12: 71566846-71566846 on Build GRCh38
rs781449893 Chr.12: 71566923-71566923 on Build GRCh38
rs761993414 Chr.12: 71566887-71566887 on Build GRCh38
rs780757046 Chr.12: 71566527-71566528 on Build GRCh38
rs747669913 Chr.12: 71566917-71566917 on Build GRCh38
rs746197354 Chr.12: 71566476-71566477 on Build GRCh38
rs752079669 Chr.12: 71566919-71566919 on Build GRCh38
rs754027007 Chr.12: 71566611-71566611 on Build GRCh38
rs748521529 Chr.12: 71566924-71566924 on Build GRCh38
rs754029658 Chr.12: 71566732-71566732 on Build GRCh38
rs969362950 Chr.12: 71566597-71566597 on Build GRCh38
rs781567446 Chr.12: 71566825-71566825 on Build GRCh38
rs770370252 Chr.12: 71566660-71566660 on Build GRCh38
rs957879706 Chr.12: 71566667-71566667 on Build GRCh38
rs778139099 Chr.12: 71566609-71566609 on Build GRCh38
rs368297247 Chr.12: 71566845-71566845 on Build GRCh38
rs373845303 Chr.12: 71566805-71566805 on Build GRCh38
rs761988407 Chr.12: 71566869-71566869 on Build GRCh38
rs762605462 Chr.12: 71566650-71566650 on Build GRCh38
rs765844467 Chr.12: 71566523-71566523 on Build GRCh38
rs747446577 Chr.12: 71566817-71566817 on Build GRCh38
rs199507313 Chr.12: 71566707-71566707 on Build GRCh38
rs755344653 Chr.12: 71566819-71566819 on Build GRCh38
rs186786655 Chr.12: 71566941-71566941 on Build GRCh38
rs987518291 Chr.12: 71566754-71566754 on Build GRCh38
rs776605980 Chr.12: 71566917-71566917 on Build GRCh38
rs746328435 Chr.12: 71566492-71566492 on Build GRCh38
rs752507557 Chr.12: 71566592-71566592 on Build GRCh38
rs1797374 Chr.12: 71566753-71566753 on Build GRCh38
rs768958781 Chr.12: 71566787-71566787 on Build GRCh38
rs752607669 Chr.12: 71566716-71566716 on Build GRCh38
rs748923260 Chr.12: 71566826-71566826 on Build GRCh38
rs766384127 Chr.12: 71566909-71566909 on Build GRCh38
rs767450241 Chr.12: 71566587-71566587 on Build GRCh38
rs377329683 Chr.12: 71566501-71566501 on Build GRCh38
rs369404015 Chr.12: 71566487-71566487 on Build GRCh38
rs572674883 Chr.12: 71566511-71566511 on Build GRCh38
rs150655850 Chr.12: 71566906-71566906 on Build GRCh38
rs778037024 Chr.12: 71566469-71566469 on Build GRCh38
rs201779813 Chr.12: 71566907-71566907 on Build GRCh38
rs757297502 Chr.12: 71566613-71566613 on Build GRCh38
rs560308801 Chr.12: 71566880-71566880 on Build GRCh38
rs779005404 Chr.12: 71566615-71566615 on Build GRCh38
rs772740418 Chr.12: 71566645-71566645 on Build GRCh38
rs762363291 Chr.12: 71566517-71566517 on Build GRCh38
rs148862507 Chr.12: 71566881-71566881 on Build GRCh38
rs181911196 Chr.12: 71566864-71566864 on Build GRCh38
rs377304651 Chr.12: 71566579-71566579 on Build GRCh38
rs750497140 Chr.12: 71566747-71566747 on Build GRCh38
rs768224247 Chr.12: 71566526-71566527 on Build GRCh38
rs756452580 Chr.12: 71566593-71566593 on Build GRCh38
rs944193584 Chr.12: 71566774-71566774 on Build GRCh38
rs772539650 Chr.12: 71566630-71566630 on Build GRCh38
rs769349487 Chr.12: 71566935-71566937 on Build GRCh38
rs200467075 Chr.12: 71566847-71566847 on Build GRCh38
rs767537930 Chr.12: 71566713-71566713 on Build GRCh38
rs749317425 Chr.12: 71566480-71566480 on Build GRCh38
rs747358426 Chr.12: 71566636-71566636 on Build GRCh38
rs1726469 Chr.12: 71566804-71566804 on Build GRCh38
rs372139572 Chr.12: 71566916-71566916 on Build GRCh38
rs11178849 Chr.12: 71566497-71566497 on Build GRCh38
rs376379225 Chr.12: 71566641-71566641 on Build GRCh38
rs778902758 Chr.12: 71566745-71566745 on Build GRCh38
rs751460484 Chr.12: 71566582-71566582 on Build GRCh38
rs759469110 Chr.12: 71566584-71566584 on Build GRCh38
rs761496077 Chr.12: 71566508-71566508 on Build GRCh38
rs370233161 Chr.12: 71566661-71566661 on Build GRCh38
rs376618532 Chr.12: 71566720-71566720 on Build GRCh38
rs755436607 Chr.12: 71566920-71566920 on Build GRCh38
rs780448269 Chr.12: 71566635-71566635 on Build GRCh38

View Full Product Details