Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9263376-9263890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H6PD
Entrez Gene ID
9563
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases
CORTRD1, G6PDH, GDH
UniGene
Hs.463511
Gene Chromosome Location
Chr.1: 9234767-9271337 on Build GRCh38
Transcripts
NM_001282587.1 NM_004285.3 XM_005263540.4 XM_006711052.3 XM_017002865.1 XM_017002866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6342499 c.1110G>T p.R370R Chr.1: 9263603-9263603 on Build GRCh38
1667122 c.1051G>T p.G351C Chr.1: 9263544-9263544 on Build GRCh38
5470962 c.1054G>A p.V352M Chr.1: 9263547-9263547 on Build GRCh38
7103636 c.1213G>T p.G405C Chr.1: 9263706-9263706 on Build GRCh38
6256411 c.1086G>T p.L362F Chr.1: 9263579-9263579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754138358 Chr.1: 9263738-9263738 on Build GRCh38
rs752700691 Chr.1: 9263662-9263662 on Build GRCh38
rs765907217 Chr.1: 9263601-9263601 on Build GRCh38
rs769224273 Chr.1: 9263587-9263587 on Build GRCh38
rs113556335 Chr.1: 9263432-9263432 on Build GRCh38
rs775189836 Chr.1: 9263765-9263765 on Build GRCh38
rs778974717 Chr.1: 9263604-9263604 on Build GRCh38
rs77405831 Chr.1: 9263412-9263413 on Build GRCh38
rs767745240 Chr.1: 9263595-9263595 on Build GRCh38
rs544218562 Chr.1: 9263436-9263436 on Build GRCh38
rs757161663 Chr.1: 9263729-9263729 on Build GRCh38
rs34580447 Chr.1: 9263835-9263835 on Build GRCh38
rs772831999 Chr.1: 9263427-9263428 on Build GRCh38
rs768856168 Chr.1: 9263640-9263640 on Build GRCh38
rs200623006 Chr.1: 9263862-9263862 on Build GRCh38
rs139280233 Chr.1: 9263657-9263657 on Build GRCh38
rs773962051 Chr.1: 9263829-9263829 on Build GRCh38
rs763502217 Chr.1: 9263524-9263524 on Build GRCh38
rs879771370 Chr.1: 9263850-9263851 on Build GRCh38
rs776704214 Chr.1: 9263514-9263514 on Build GRCh38
rs754871100 Chr.1: 9263552-9263552 on Build GRCh38
rs148202091 Chr.1: 9263857-9263857 on Build GRCh38
rs776657727 Chr.1: 9263523-9263523 on Build GRCh38
rs780493906 Chr.1: 9263679-9263679 on Build GRCh38
rs1045557565 Chr.1: 9263664-9263664 on Build GRCh38
rs747491947 Chr.1: 9263751-9263751 on Build GRCh38
rs751190442 Chr.1: 9263458-9263458 on Build GRCh38
rs763555076 Chr.1: 9263584-9263584 on Build GRCh38
rs201655325 Chr.1: 9263519-9263519 on Build GRCh38
rs759320700 Chr.1: 9263589-9263590 on Build GRCh38
rs149231638 Chr.1: 9263656-9263656 on Build GRCh38
rs746053638 Chr.1: 9263577-9263577 on Build GRCh38
rs199782560 Chr.1: 9263547-9263547 on Build GRCh38
rs937286131 Chr.1: 9263672-9263672 on Build GRCh38
rs1019398484 Chr.1: 9263616-9263616 on Build GRCh38
rs750176902 Chr.1: 9263787-9263787 on Build GRCh38
rs886258579 Chr.1: 9263419-9263419 on Build GRCh38
rs369125646 Chr.1: 9263688-9263688 on Build GRCh38
rs761795776 Chr.1: 9263780-9263780 on Build GRCh38
rs763669347 Chr.1: 9263768-9263768 on Build GRCh38
rs762433179 Chr.1: 9263766-9263766 on Build GRCh38
rs748229239 Chr.1: 9263459-9263459 on Build GRCh38
rs200192570 Chr.1: 9263527-9263527 on Build GRCh38
rs199663762 Chr.1: 9263660-9263660 on Build GRCh38
rs367704323 Chr.1: 9263567-9263567 on Build GRCh38
rs562470687 Chr.1: 9263717-9263717 on Build GRCh38
rs749535728 Chr.1: 9263698-9263698 on Build GRCh38
rs757102104 Chr.1: 9263677-9263677 on Build GRCh38
rs150510233 Chr.1: 9263691-9263691 on Build GRCh38
rs906913496 Chr.1: 9263431-9263431 on Build GRCh38
rs146475150 Chr.1: 9263803-9263803 on Build GRCh38
rs767402836 Chr.1: 9263734-9263734 on Build GRCh38
rs745535415 Chr.1: 9263755-9263755 on Build GRCh38
rs765290945 Chr.1: 9263455-9263455 on Build GRCh38
rs201017402 Chr.1: 9263727-9263727 on Build GRCh38
rs1053056559 Chr.1: 9263430-9263430 on Build GRCh38
rs140374783 Chr.1: 9263490-9263490 on Build GRCh38
rs774711646 Chr.1: 9263588-9263588 on Build GRCh38
rs966297321 Chr.1: 9263641-9263641 on Build GRCh38
rs139332011 Chr.1: 9263726-9263726 on Build GRCh38
rs148535734 Chr.1: 9263826-9263826 on Build GRCh38
rs902836606 Chr.1: 9263858-9263858 on Build GRCh38
rs754923262 Chr.1: 9263603-9263603 on Build GRCh38
rs774793481 Chr.1: 9263723-9263723 on Build GRCh38
rs377461550 Chr.1: 9263700-9263700 on Build GRCh38
rs112859991 Chr.1: 9263687-9263687 on Build GRCh38
rs768490485 Chr.1: 9263499-9263499 on Build GRCh38
rs1048900717 Chr.1: 9263412-9263412 on Build GRCh38
rs779056308 Chr.1: 9263741-9263741 on Build GRCh38
rs768308841 Chr.1: 9263828-9263828 on Build GRCh38
rs759138892 Chr.1: 9263714-9263714 on Build GRCh38
rs201211727 Chr.1: 9263834-9263834 on Build GRCh38
rs367995382 Chr.1: 9263612-9263612 on Build GRCh38
rs371234309 Chr.1: 9263856-9263856 on Build GRCh38
rs373155135 Chr.1: 9263850-9263850 on Build GRCh38
rs1024176043 Chr.1: 9263423-9263423 on Build GRCh38
rs749195458 Chr.1: 9263497-9263497 on Build GRCh38
rs746274075 Chr.1: 9263819-9263819 on Build GRCh38
rs370333038 Chr.1: 9263861-9263861 on Build GRCh38
rs750142418 Chr.1: 9263597-9263597 on Build GRCh38
rs142773575 Chr.1: 9263511-9263511 on Build GRCh38
rs372714313 Chr.1: 9263696-9263696 on Build GRCh38
rs34274595 Chr.1: 9263811-9263811 on Build GRCh38
rs771358643 Chr.1: 9263707-9263707 on Build GRCh38
rs748119651 Chr.1: 9263742-9263742 on Build GRCh38
rs762309411 Chr.1: 9263594-9263594 on Build GRCh38
rs770012834 Chr.1: 9263582-9263582 on Build GRCh38
rs996470142 Chr.1: 9263759-9263759 on Build GRCh38
rs199919142 Chr.1: 9263639-9263639 on Build GRCh38
rs1021351055 Chr.1: 9263437-9263437 on Build GRCh38
rs756870094 Chr.1: 9263532-9263532 on Build GRCh38
rs966937656 Chr.1: 9263609-9263609 on Build GRCh38
rs766394409 Chr.1: 9263479-9263480 on Build GRCh38
rs376261710 Chr.1: 9263753-9263753 on Build GRCh38
rs770072769 Chr.1: 9263629-9263629 on Build GRCh38
rs754695706 Chr.1: 9263846-9263846 on Build GRCh38
rs371007806 Chr.1: 9263489-9263489 on Build GRCh38
rs765376175 Chr.1: 9263517-9263517 on Build GRCh38
rs774085640 Chr.1: 9263510-9263510 on Build GRCh38
rs115109857 Chr.1: 9263654-9263654 on Build GRCh38
rs767262518 Chr.1: 9263535-9263535 on Build GRCh38
rs778291064 Chr.1: 9263748-9263748 on Build GRCh38
rs201429712 Chr.1: 9263655-9263655 on Build GRCh38
rs146809075 Chr.1: 9263583-9263583 on Build GRCh38
rs141143158 Chr.1: 9263404-9263405 on Build GRCh38
rs200035545 Chr.1: 9263810-9263810 on Build GRCh38
rs748032955 Chr.1: 9263561-9263561 on Build GRCh38
rs139459848 Chr.1: 9263699-9263699 on Build GRCh38
rs574364535 Chr.1: 9263708-9263708 on Build GRCh38
rs148693050 Chr.1: 9263545-9263545 on Build GRCh38
rs765140953 Chr.1: 9263606-9263609 on Build GRCh38
rs780289540 Chr.1: 9263638-9263638 on Build GRCh38
rs755118190 Chr.1: 9263739-9263739 on Build GRCh38
rs900592496 Chr.1: 9263397-9263397 on Build GRCh38
rs573705148 Chr.1: 9263456-9263456 on Build GRCh38
rs938499645 Chr.1: 9263559-9263559 on Build GRCh38
rs774483435 Chr.1: 9263777-9263777 on Build GRCh38
rs1001382455 Chr.1: 9263579-9263579 on Build GRCh38
rs914480831 Chr.1: 9263840-9263840 on Build GRCh38
rs774966768 Chr.1: 9263646-9263646 on Build GRCh38
rs551302514 Chr.1: 9263530-9263530 on Build GRCh38
rs756889821 Chr.1: 9263460-9263460 on Build GRCh38
rs201340516 Chr.1: 9263709-9263709 on Build GRCh38
rs767454234 Chr.1: 9263786-9263786 on Build GRCh38
rs974979153 Chr.1: 9263649-9263649 on Build GRCh38
rs149463138 Chr.1: 9263536-9263536 on Build GRCh38
rs752434362 Chr.1: 9263676-9263676 on Build GRCh38
rs200294863 Chr.1: 9263818-9263818 on Build GRCh38
rs371176376 Chr.1: 9263462-9263462 on Build GRCh38
rs553867100 Chr.1: 9263697-9263697 on Build GRCh38
rs201960157 Chr.1: 9263756-9263756 on Build GRCh38
rs771118768 Chr.1: 9263513-9263513 on Build GRCh38
rs762206316 Chr.1: 9263647-9263647 on Build GRCh38
rs182877860 Chr.1: 9263602-9263602 on Build GRCh38
rs948124664 Chr.1: 9263543-9263543 on Build GRCh38
rs752960171 Chr.1: 9263795-9263795 on Build GRCh38
rs6688832 Chr.1: 9263851-9263851 on Build GRCh38
rs1004828382 Chr.1: 9263420-9263420 on Build GRCh38
rs759086163 Chr.1: 9263659-9263659 on Build GRCh38
rs561670704 Chr.1: 9263418-9263418 on Build GRCh38
rs755243643 Chr.1: 9263790-9263790 on Build GRCh38
rs778577434 Chr.1: 9263553-9263553 on Build GRCh38
rs387907167 Chr.1: 9263569-9263569 on Build GRCh38
rs201145062 Chr.1: 9263598-9263598 on Build GRCh38
rs745329356 Chr.1: 9263480-9263480 on Build GRCh38
rs746120861 Chr.1: 9263628-9263628 on Build GRCh38
rs758491842 Chr.1: 9263747-9263747 on Build GRCh38
rs758260308 Chr.1: 9263610-9263610 on Build GRCh38
rs147080717 Chr.1: 9263671-9263671 on Build GRCh38
rs772512315 Chr.1: 9263650-9263650 on Build GRCh38
rs770186803 Chr.1: 9263825-9263825 on Build GRCh38
rs147962990 Chr.1: 9263836-9263836 on Build GRCh38
rs751338036 Chr.1: 9263676-9263676 on Build GRCh38
rs731183 Chr.1: 9263398-9263398 on Build GRCh38
rs35511555 Chr.1: 9263680-9263680 on Build GRCh38
rs34448374 Chr.1: 9263546-9263546 on Build GRCh38

View Full Product Details