Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11059881-11060399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRM
Entrez Gene ID
6723
Gene Name
spermidine synthase
Gene Aliases
PAPT, SPDSY, SPS1, SRML1
UniGene
Hs.76244
Gene Chromosome Location
Chr.1: 11054592-11060053 on Build GRCh38
Transcripts
NM_003132.2 XM_017002179.1

SNP Detail

SNP ID Chromosome Location
rs1003232507 Chr.1: 11059998-11059998 on Build GRCh38
rs1056184792 Chr.1: 11060171-11060171 on Build GRCh38
rs901290445 Chr.1: 11059946-11059946 on Build GRCh38
rs556818195 Chr.1: 11060380-11060380 on Build GRCh38
rs930067970 Chr.1: 11059955-11059955 on Build GRCh38
rs762880971 Chr.1: 11060138-11060147 on Build GRCh38
rs986037136 Chr.1: 11060353-11060353 on Build GRCh38
rs911493375 Chr.1: 11060059-11060059 on Build GRCh38
rs892136075 Chr.1: 11060180-11060180 on Build GRCh38
rs985720517 Chr.1: 11060190-11060190 on Build GRCh38
rs1047149859 Chr.1: 11060093-11060093 on Build GRCh38
rs763990256 Chr.1: 11060146-11060155 on Build GRCh38
rs548344472 Chr.1: 11060296-11060296 on Build GRCh38
rs1045014546 Chr.1: 11059989-11059989 on Build GRCh38
rs569752199 Chr.1: 11060351-11060351 on Build GRCh38
rs956696878 Chr.1: 11060016-11060016 on Build GRCh38
rs78102922 Chr.1: 11060199-11060199 on Build GRCh38
rs745582057 Chr.1: 11059899-11059913 on Build GRCh38
rs747837409 Chr.1: 11059913-11059913 on Build GRCh38
rs1034673115 Chr.1: 11060008-11060008 on Build GRCh38
rs532318680 Chr.1: 11060223-11060223 on Build GRCh38
rs180929432 Chr.1: 11060161-11060161 on Build GRCh38
rs564707100 Chr.1: 11060200-11060200 on Build GRCh38
rs565542323 Chr.1: 11060242-11060242 on Build GRCh38
rs768958464 Chr.1: 11059910-11059910 on Build GRCh38
rs1009616373 Chr.1: 11060181-11060181 on Build GRCh38
rs756647343 Chr.1: 11059902-11059902 on Build GRCh38
rs900018033 Chr.1: 11060374-11060374 on Build GRCh38
rs772944133 Chr.1: 11059923-11059923 on Build GRCh38
rs547627804 Chr.1: 11060227-11060227 on Build GRCh38
rs922810498 Chr.1: 11060207-11060207 on Build GRCh38
rs926442367 Chr.1: 11060258-11060258 on Build GRCh38
rs976753873 Chr.1: 11060073-11060073 on Build GRCh38
rs956110814 Chr.1: 11060297-11060297 on Build GRCh38
rs976974276 Chr.1: 11060206-11060206 on Build GRCh38
rs1009318659 Chr.1: 11060025-11060025 on Build GRCh38
rs1022058050 Chr.1: 11060196-11060196 on Build GRCh38
rs780258826 Chr.1: 11059906-11059906 on Build GRCh38
rs1011999945 Chr.1: 11060299-11060299 on Build GRCh38
rs538469132 Chr.1: 11060371-11060371 on Build GRCh38
rs1005763650 Chr.1: 11059976-11059976 on Build GRCh38
rs1003455393 Chr.1: 11060164-11060164 on Build GRCh38
rs750929877 Chr.1: 11059900-11059900 on Build GRCh38
rs1044900842 Chr.1: 11060133-11060133 on Build GRCh38
rs771883659 Chr.1: 11059918-11059918 on Build GRCh38
rs951426790 Chr.1: 11060222-11060223 on Build GRCh38
rs978871514 Chr.1: 11060054-11060054 on Build GRCh38
rs749400920 Chr.1: 11059907-11059907 on Build GRCh38
rs966303136 Chr.1: 11060372-11060372 on Build GRCh38
rs964246884 Chr.1: 11060073-11060074 on Build GRCh38
rs903631610 Chr.1: 11060320-11060320 on Build GRCh38
rs531444785 Chr.1: 11060050-11060050 on Build GRCh38
rs941220541 Chr.1: 11060120-11060120 on Build GRCh38
rs888288381 Chr.1: 11059964-11059964 on Build GRCh38
rs1043768061 Chr.1: 11060309-11060309 on Build GRCh38
rs922602213 Chr.1: 11060074-11060074 on Build GRCh38
rs905085509 Chr.1: 11059996-11059996 on Build GRCh38
rs1056405082 Chr.1: 11060264-11060264 on Build GRCh38
rs942716985 Chr.1: 11059915-11059916 on Build GRCh38
rs371968296 Chr.1: 11060284-11060284 on Build GRCh38
rs909926123 Chr.1: 11060103-11060103 on Build GRCh38
rs781495048 Chr.1: 11059899-11059899 on Build GRCh38
rs1021554371 Chr.1: 11060303-11060303 on Build GRCh38
rs779279835 Chr.1: 11059912-11059912 on Build GRCh38
rs980635696 Chr.1: 11060253-11060253 on Build GRCh38
rs760625688 Chr.1: 11059925-11059925 on Build GRCh38
rs910458275 Chr.1: 11060361-11060361 on Build GRCh38
rs929927016 Chr.1: 11060083-11060083 on Build GRCh38
rs968921621 Chr.1: 11060053-11060053 on Build GRCh38
rs909780574 Chr.1: 11059961-11059988 on Build GRCh38
rs911440914 Chr.1: 11059914-11059914 on Build GRCh38
rs939130339 Chr.1: 11060261-11060261 on Build GRCh38
rs905137881 Chr.1: 11060148-11060148 on Build GRCh38

View Full Product Details