Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 99648004-99648546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
NM_152788.4 XM_005269028.4 XM_005269029.4 XM_006719504.3 XM_006719505.3 XM_006719506.3 XM_006719507.3 XM_006719508.3 XM_006719509.3 XM_006719510.3 XM_006719512.3 XM_006719513.3 XM_006719514.3 XM_011538571.2 XM_017019651.1 XM_017019652.1 XM_017019653.1 XM_017019654.1 XM_017019655.1 XM_017019656.1 XM_017019657.1 XM_017019658.1 NM_153364.3

Gene Symbol
FAM71C
Entrez Gene ID
196472
Gene Name
family with sequence similarity 71 member C
Gene Aliases
-
UniGene
Hs.591011
Gene Chromosome Location
Chr.12: 99647750-99650114 on Build GRCh38
Transcripts
NM_152788.4 XM_005269028.4 XM_005269029.4 XM_006719504.3 XM_006719505.3 XM_006719506.3 XM_006719507.3 XM_006719508.3 XM_006719509.3 XM_006719510.3 XM_006719512.3 XM_006719513.3 XM_006719514.3 XM_011538571.2 XM_017019651.1 XM_017019652.1 XM_017019653.1 XM_017019654.1 XM_017019655.1 XM_017019656.1 XM_017019657.1 XM_017019658.1 NM_153364.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
355018 c.133A>G p.M45V Chr.12: 99648307-99648307 on Build GRCh38
1158002 c.24G>A p.P8P Chr.12: 99648198-99648198 on Build GRCh38
3700175 c.86A>T p.Q29L Chr.12: 99648260-99648260 on Build GRCh38
4911254 c.266C>T p.A89V Chr.12: 99648440-99648440 on Build GRCh38
1206438 c.370A>G p.M124V Chr.12: 99648544-99648544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775448296 Chr.12: 99648255-99648255 on Build GRCh38
rs369353550 Chr.12: 99648187-99648187 on Build GRCh38
rs11109969 Chr.12: 99648385-99648385 on Build GRCh38
rs746189188 Chr.12: 99648329-99648329 on Build GRCh38
rs149716372 Chr.12: 99648136-99648136 on Build GRCh38
rs772273782 Chr.12: 99648227-99648227 on Build GRCh38
rs1006508574 Chr.12: 99648034-99648034 on Build GRCh38
rs141205945 Chr.12: 99648357-99648357 on Build GRCh38
rs188419765 Chr.12: 99648071-99648071 on Build GRCh38
rs901063107 Chr.12: 99648052-99648052 on Build GRCh38
rs759123630 Chr.12: 99648267-99648267 on Build GRCh38
rs200408405 Chr.12: 99648358-99648358 on Build GRCh38
rs780699403 Chr.12: 99648148-99648148 on Build GRCh38
rs74343597 Chr.12: 99648509-99648509 on Build GRCh38
rs756669117 Chr.12: 99648214-99648214 on Build GRCh38
rs772068989 Chr.12: 99648178-99648178 on Build GRCh38
rs769922789 Chr.12: 99648260-99648260 on Build GRCh38
rs181913768 Chr.12: 99648442-99648442 on Build GRCh38
rs200084539 Chr.12: 99648163-99648163 on Build GRCh38
rs781431969 Chr.12: 99648433-99648433 on Build GRCh38
rs754365772 Chr.12: 99648310-99648310 on Build GRCh38
rs771264905 Chr.12: 99648221-99648221 on Build GRCh38
rs759710522 Chr.12: 99648176-99648176 on Build GRCh38
rs757719965 Chr.12: 99648400-99648400 on Build GRCh38
rs192871611 Chr.12: 99648197-99648197 on Build GRCh38
rs755589291 Chr.12: 99648282-99648282 on Build GRCh38
rs779288295 Chr.12: 99648222-99648222 on Build GRCh38
rs751298344 Chr.12: 99648263-99648263 on Build GRCh38
rs562310463 Chr.12: 99648198-99648198 on Build GRCh38
rs540572362 Chr.12: 99648083-99648083 on Build GRCh38
rs922290339 Chr.12: 99648508-99648508 on Build GRCh38
rs779843101 Chr.12: 99648132-99648132 on Build GRCh38
rs200401824 Chr.12: 99648288-99648288 on Build GRCh38
rs751188669 Chr.12: 99648200-99648200 on Build GRCh38
rs386765697 Chr.12: 99648163-99648164 on Build GRCh38
rs371174339 Chr.12: 99648289-99648289 on Build GRCh38
rs147841911 Chr.12: 99648414-99648414 on Build GRCh38
rs760326691 Chr.12: 99648487-99648487 on Build GRCh38
rs201447865 Chr.12: 99648353-99648353 on Build GRCh38
rs758140403 Chr.12: 99648124-99648124 on Build GRCh38
rs529377642 Chr.12: 99648154-99648154 on Build GRCh38
rs984747632 Chr.12: 99648294-99648294 on Build GRCh38
rs534181485 Chr.12: 99648404-99648405 on Build GRCh38
rs146105601 Chr.12: 99648397-99648397 on Build GRCh38
rs758762072 Chr.12: 99648415-99648415 on Build GRCh38
rs35471478 Chr.12: 99648368-99648368 on Build GRCh38
rs771924298 Chr.12: 99648466-99648466 on Build GRCh38
rs80068786 Chr.12: 99648060-99648060 on Build GRCh38
rs35601898 Chr.12: 99648044-99648044 on Build GRCh38
rs775600923 Chr.12: 99648184-99648184 on Build GRCh38
rs776179544 Chr.12: 99648499-99648499 on Build GRCh38
rs754421919 Chr.12: 99648145-99648145 on Build GRCh38
rs760828488 Chr.12: 99648258-99648258 on Build GRCh38
rs765707994 Chr.12: 99648504-99648504 on Build GRCh38
rs375580075 Chr.12: 99648207-99648207 on Build GRCh38
rs143631956 Chr.12: 99648503-99648503 on Build GRCh38
rs753279689 Chr.12: 99648398-99648398 on Build GRCh38
rs949547743 Chr.12: 99648153-99648153 on Build GRCh38
rs375821466 Chr.12: 99648315-99648315 on Build GRCh38
rs942377709 Chr.12: 99648217-99648217 on Build GRCh38
rs774406383 Chr.12: 99648172-99648172 on Build GRCh38
rs370044784 Chr.12: 99648505-99648505 on Build GRCh38
rs112162917 Chr.12: 99648394-99648394 on Build GRCh38
rs990788899 Chr.12: 99648343-99648343 on Build GRCh38
rs776291685 Chr.12: 99648521-99648524 on Build GRCh38
rs919130423 Chr.12: 99648028-99648028 on Build GRCh38
rs976319306 Chr.12: 99648237-99648237 on Build GRCh38
rs757795041 Chr.12: 99648162-99648163 on Build GRCh38
rs755181903 Chr.12: 99648425-99648425 on Build GRCh38
rs11109967 Chr.12: 99648074-99648074 on Build GRCh38
rs533644132 Chr.12: 99648379-99648379 on Build GRCh38
rs770218593 Chr.12: 99648155-99648155 on Build GRCh38
rs918777996 Chr.12: 99648170-99648170 on Build GRCh38
rs367749248 Chr.12: 99648416-99648416 on Build GRCh38
rs745345294 Chr.12: 99648515-99648516 on Build GRCh38
rs746231620 Chr.12: 99648493-99648494 on Build GRCh38
rs774070920 Chr.12: 99648373-99648373 on Build GRCh38
rs139770941 Chr.12: 99648219-99648219 on Build GRCh38
rs763680646 Chr.12: 99648497-99648497 on Build GRCh38
rs558756838 Chr.12: 99648439-99648439 on Build GRCh38
rs767840757 Chr.12: 99648486-99648486 on Build GRCh38
rs749638352 Chr.12: 99648171-99648171 on Build GRCh38
rs1026181901 Chr.12: 99648248-99648248 on Build GRCh38
rs1018149581 Chr.12: 99648051-99648051 on Build GRCh38
rs753956423 Chr.12: 99648300-99648300 on Build GRCh38
rs752215626 Chr.12: 99648281-99648281 on Build GRCh38
rs778302965 Chr.12: 99648298-99648298 on Build GRCh38
rs767184534 Chr.12: 99648277-99648277 on Build GRCh38
rs765210814 Chr.12: 99648185-99648185 on Build GRCh38
rs747261860 Chr.12: 99648418-99648418 on Build GRCh38
rs372408915 Chr.12: 99648204-99648204 on Build GRCh38
rs745698238 Chr.12: 99648445-99648445 on Build GRCh38
rs115856728 Chr.12: 99648393-99648393 on Build GRCh38
rs879054947 Chr.12: 99648094-99648094 on Build GRCh38
rs771542531 Chr.12: 99648341-99648343 on Build GRCh38
rs34319076 Chr.12: 99648162-99648162 on Build GRCh38
rs749851760 Chr.12: 99648166-99648169 on Build GRCh38
rs199610950 Chr.12: 99648424-99648424 on Build GRCh38
rs758609015 Chr.12: 99648342-99648342 on Build GRCh38
rs145530655 Chr.12: 99648218-99648218 on Build GRCh38
rs11109968 Chr.12: 99648262-99648262 on Build GRCh38
rs141451941 Chr.12: 99648443-99648443 on Build GRCh38
rs775101098 Chr.12: 99648387-99648387 on Build GRCh38
rs79915291 Chr.12: 99648164-99648164 on Build GRCh38
rs868589073 Chr.12: 99648232-99648232 on Build GRCh38
rs938078020 Chr.12: 99648112-99648112 on Build GRCh38
rs760259280 Chr.12: 99648223-99648223 on Build GRCh38
rs151145389 Chr.12: 99648318-99648318 on Build GRCh38
rs750305493 Chr.12: 99648190-99648190 on Build GRCh38
rs573069920 Chr.12: 99648082-99648082 on Build GRCh38
rs184425136 Chr.12: 99648374-99648374 on Build GRCh38
rs770712483 Chr.12: 99648163-99648163 on Build GRCh38
rs146401531 Chr.12: 99648206-99648206 on Build GRCh38

View Full Product Details