Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105111033-105111532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1033
Entrez Gene ID
23325
Gene Name
KIAA1033
Gene Aliases
MRT43, SWIP
UniGene
Hs.12144
Gene Chromosome Location
Chr.12: 105107714-105169134 on Build GRCh38
Transcripts
NM_001293640.1 NM_015275.2 XM_011538073.2 XM_011538074.2 XM_011538077.2 XM_017019082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312280 c.196T>G p.L66V Chr.12: 105111259-105111259 on Build GRCh38
3810847 c.88A>C p.N30H Chr.12: 105111151-105111151 on Build GRCh38
4186638 c.151G>T p.D51Y Chr.12: 105111214-105111214 on Build GRCh38
4838840 c.73G>A p.E25K Chr.12: 105111136-105111136 on Build GRCh38
7373552 c.112T>C p.Y38H Chr.12: 105111175-105111175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747921897 Chr.12: 105111248-105111248 on Build GRCh38
rs749936552 Chr.12: 105111223-105111223 on Build GRCh38
rs200700259 Chr.12: 105111086-105111086 on Build GRCh38
rs536953726 Chr.12: 105111220-105111220 on Build GRCh38
rs913340203 Chr.12: 105111357-105111357 on Build GRCh38
rs961797226 Chr.12: 105111258-105111258 on Build GRCh38
rs774589525 Chr.12: 105111189-105111189 on Build GRCh38
rs751867701 Chr.12: 105111085-105111085 on Build GRCh38
rs759625987 Chr.12: 105111198-105111198 on Build GRCh38
rs376604258 Chr.12: 105111217-105111217 on Build GRCh38
rs372395687 Chr.12: 105111216-105111216 on Build GRCh38
rs367807155 Chr.12: 105111141-105111141 on Build GRCh38
rs532409419 Chr.12: 105111145-105111145 on Build GRCh38
rs947605594 Chr.12: 105111432-105111432 on Build GRCh38
rs914844624 Chr.12: 105111422-105111422 on Build GRCh38
rs545281016 Chr.12: 105111349-105111349 on Build GRCh38
rs374857462 Chr.12: 105111256-105111256 on Build GRCh38
rs955034316 Chr.12: 105111199-105111199 on Build GRCh38
rs776428689 Chr.12: 105111074-105111074 on Build GRCh38
rs373669216 Chr.12: 105111103-105111103 on Build GRCh38
rs767793363 Chr.12: 105111092-105111092 on Build GRCh38
rs745332143 Chr.12: 105111134-105111134 on Build GRCh38
rs35294333 Chr.12: 105111175-105111175 on Build GRCh38
rs867813832 Chr.12: 105111263-105111263 on Build GRCh38
rs764330483 Chr.12: 105111282-105111282 on Build GRCh38
rs1043056859 Chr.12: 105111159-105111159 on Build GRCh38
rs147988679 Chr.12: 105111180-105111180 on Build GRCh38
rs766343767 Chr.12: 105111082-105111082 on Build GRCh38
rs1042083862 Chr.12: 105111073-105111073 on Build GRCh38
rs114844466 Chr.12: 105111272-105111272 on Build GRCh38
rs777596923 Chr.12: 105111221-105111221 on Build GRCh38
rs920228760 Chr.12: 105111250-105111250 on Build GRCh38
rs779734652 Chr.12: 105111230-105111230 on Build GRCh38
rs754194473 Chr.12: 105111219-105111219 on Build GRCh38
rs746872657 Chr.12: 105111156-105111156 on Build GRCh38
rs992033000 Chr.12: 105111414-105111414 on Build GRCh38
rs1043079557 Chr.12: 105111435-105111435 on Build GRCh38
rs779613567 Chr.12: 105111144-105111144 on Build GRCh38
rs866295026 Chr.12: 105111154-105111154 on Build GRCh38
rs996473754 Chr.12: 105111501-105111501 on Build GRCh38
rs186069955 Chr.12: 105111367-105111367 on Build GRCh38
rs372211839 Chr.12: 105111251-105111251 on Build GRCh38
rs762168208 Chr.12: 105111295-105111295 on Build GRCh38
rs746641611 Chr.12: 105111236-105111236 on Build GRCh38
rs753120367 Chr.12: 105111095-105111095 on Build GRCh38
rs778402246 Chr.12: 105111102-105111102 on Build GRCh38
rs771724807 Chr.12: 105111135-105111135 on Build GRCh38
rs768318313 Chr.12: 105111170-105111170 on Build GRCh38
rs924596300 Chr.12: 105111507-105111507 on Build GRCh38
rs772187070 Chr.12: 105111275-105111276 on Build GRCh38
rs200170294 Chr.12: 105111226-105111226 on Build GRCh38
rs770842529 Chr.12: 105111499-105111499 on Build GRCh38
rs763147384 Chr.12: 105111075-105111075 on Build GRCh38
rs34099762 Chr.12: 105111311-105111312 on Build GRCh38
rs772474232 Chr.12: 105111484-105111485 on Build GRCh38
rs756546800 Chr.12: 105111098-105111098 on Build GRCh38
rs780878662 Chr.12: 105111244-105111244 on Build GRCh38
rs773279430 Chr.12: 105111253-105111253 on Build GRCh38
rs368178226 Chr.12: 105111204-105111204 on Build GRCh38
rs776870349 Chr.12: 105111289-105111289 on Build GRCh38
rs775845456 Chr.12: 105111207-105111207 on Build GRCh38
rs141620718 Chr.12: 105111242-105111242 on Build GRCh38
rs904518052 Chr.12: 105111471-105111471 on Build GRCh38
rs977474456 Chr.12: 105111446-105111446 on Build GRCh38
rs182772377 Chr.12: 105111281-105111281 on Build GRCh38

View Full Product Details