Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109277899-109278400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN4
Entrez Gene ID
121643
Gene Name
forkhead box N4
Gene Aliases
-
UniGene
Hs.528316
Gene Chromosome Location
Chr.12: 109277978-109309406 on Build GRCh38
Transcripts
NM_213596.2 XM_011537922.2 XM_017018818.1

SNP Detail

SNP ID Chromosome Location
rs370005369 Chr.12: 109278360-109278360 on Build GRCh38
rs577250526 Chr.12: 109278076-109278076 on Build GRCh38
rs1043408462 Chr.12: 109278053-109278053 on Build GRCh38
rs886474221 Chr.12: 109278203-109278203 on Build GRCh38
rs889723102 Chr.12: 109277965-109277965 on Build GRCh38
rs1049650657 Chr.12: 109277935-109277935 on Build GRCh38
rs1052730083 Chr.12: 109278022-109278022 on Build GRCh38
rs1010736961 Chr.12: 109278027-109278027 on Build GRCh38
rs971576473 Chr.12: 109278050-109278050 on Build GRCh38
rs570638927 Chr.12: 109278352-109278352 on Build GRCh38
rs772038022 Chr.12: 109278077-109278081 on Build GRCh38
rs34408675 Chr.12: 109277972-109277973 on Build GRCh38
rs959741631 Chr.12: 109278282-109278282 on Build GRCh38
rs1006709380 Chr.12: 109278068-109278068 on Build GRCh38
rs560265343 Chr.12: 109278204-109278204 on Build GRCh38
rs901020576 Chr.12: 109277919-109277922 on Build GRCh38
rs146853631 Chr.12: 109278063-109278063 on Build GRCh38
rs767704096 Chr.12: 109278128-109278128 on Build GRCh38
rs889690516 Chr.12: 109278304-109278304 on Build GRCh38
rs998029516 Chr.12: 109277934-109277934 on Build GRCh38
rs531528802 Chr.12: 109278250-109278250 on Build GRCh38
rs181115971 Chr.12: 109277996-109277996 on Build GRCh38
rs979833723 Chr.12: 109277958-109277958 on Build GRCh38
rs1037201847 Chr.12: 109278136-109278136 on Build GRCh38
rs1016956574 Chr.12: 109278205-109278205 on Build GRCh38
rs59226497 Chr.12: 109278127-109278127 on Build GRCh38
rs970348112 Chr.12: 109278216-109278216 on Build GRCh38
rs372383774 Chr.12: 109278017-109278017 on Build GRCh38
rs980241712 Chr.12: 109278218-109278218 on Build GRCh38
rs947066924 Chr.12: 109278303-109278303 on Build GRCh38
rs952308342 Chr.12: 109278112-109278112 on Build GRCh38
rs528100229 Chr.12: 109277976-109277976 on Build GRCh38
rs912894781 Chr.12: 109278030-109278030 on Build GRCh38
rs543833073 Chr.12: 109278049-109278049 on Build GRCh38
rs564745661 Chr.12: 109278344-109278344 on Build GRCh38
rs1049576693 Chr.12: 109278215-109278215 on Build GRCh38
rs747527608 Chr.12: 109278229-109278229 on Build GRCh38
rs1035861693 Chr.12: 109278249-109278249 on Build GRCh38
rs778978388 Chr.12: 109277938-109277938 on Build GRCh38
rs35629179 Chr.12: 109278215-109278216 on Build GRCh38
rs571912501 Chr.12: 109278241-109278241 on Build GRCh38
rs942639941 Chr.12: 109278374-109278374 on Build GRCh38
rs1040870888 Chr.12: 109278079-109278079 on Build GRCh38
rs574768598 Chr.12: 109278005-109278006 on Build GRCh38
rs542471648 Chr.12: 109278246-109278246 on Build GRCh38
rs939752960 Chr.12: 109278135-109278135 on Build GRCh38
rs988591878 Chr.12: 109278295-109278295 on Build GRCh38
rs911102007 Chr.12: 109278363-109278363 on Build GRCh38
rs560839428 Chr.12: 109278248-109278248 on Build GRCh38
rs1033460089 Chr.12: 109278001-109278001 on Build GRCh38
rs771206245 Chr.12: 109278137-109278138 on Build GRCh38
rs150331860 Chr.12: 109278299-109278299 on Build GRCh38

View Full Product Details