Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113158450-113158984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP73
Entrez Gene ID
387885
Gene Name
cilia and flagella associated protein 73
Gene Aliases
CCDC42B, MIA2
UniGene
Hs.486856
Gene Chromosome Location
Chr.12: 113149162-113159276 on Build GRCh38
Transcripts
NM_001144872.1 NM_001111322.1 NM_024072.3

Gene Symbol
DDX54
Entrez Gene ID
79039
Gene Name
DEAD-box helicase 54
Gene Aliases
DP97
UniGene
Hs.506861
Gene Chromosome Location
Chr.12: 113157173-113185479 on Build GRCh38
Transcripts
NM_001144872.1 NM_001111322.1 NM_024072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5823584 c.2593G>A p.A865T Chr.12: 113158930-113158930 on Build GRCh38
6641958 c.2570G>A p.R857H Chr.12: 113158953-113158953 on Build GRCh38
6641955 c.2611C>T p.R871C Chr.12: 113158912-113158912 on Build GRCh38
2172330 c.2593G>T p.A865S Chr.12: 113158930-113158930 on Build GRCh38
1298943 c.2605G>C p.G869R Chr.12: 113158918-113158918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920493194 Chr.12: 113158639-113158639 on Build GRCh38
rs766193820 Chr.12: 113158836-113158836 on Build GRCh38
rs540550540 Chr.12: 113158822-113158822 on Build GRCh38
rs374031625 Chr.12: 113158904-113158904 on Build GRCh38
rs994081942 Chr.12: 113158694-113158694 on Build GRCh38
rs922432968 Chr.12: 113158538-113158538 on Build GRCh38
rs1029099234 Chr.12: 113158545-113158545 on Build GRCh38
rs376972962 Chr.12: 113158830-113158830 on Build GRCh38
rs781605282 Chr.12: 113158891-113158891 on Build GRCh38
rs746084777 Chr.12: 113158956-113158956 on Build GRCh38
rs904640918 Chr.12: 113158553-113158553 on Build GRCh38
rs755747930 Chr.12: 113158862-113158862 on Build GRCh38
rs577667023 Chr.12: 113158706-113158706 on Build GRCh38
rs779064090 Chr.12: 113158954-113158954 on Build GRCh38
rs1005230821 Chr.12: 113158949-113158949 on Build GRCh38
rs1041446835 Chr.12: 113158735-113158735 on Build GRCh38
rs767334221 Chr.12: 113158911-113158911 on Build GRCh38
rs750074972 Chr.12: 113158928-113158928 on Build GRCh38
rs763131577 Chr.12: 113158835-113158835 on Build GRCh38
rs760850225 Chr.12: 113158960-113158960 on Build GRCh38
rs375126560 Chr.12: 113158858-113158858 on Build GRCh38
rs756971909 Chr.12: 113158921-113158921 on Build GRCh38
rs370671347 Chr.12: 113158816-113158816 on Build GRCh38
rs768645747 Chr.12: 113158965-113158965 on Build GRCh38
rs555344760 Chr.12: 113158925-113158925 on Build GRCh38
rs573553617 Chr.12: 113158563-113158563 on Build GRCh38
rs201483723 Chr.12: 113158930-113158930 on Build GRCh38
rs942644788 Chr.12: 113158514-113158514 on Build GRCh38
rs565984923 Chr.12: 113158621-113158621 on Build GRCh38
rs760646463 Chr.12: 113158913-113158913 on Build GRCh38
rs911137557 Chr.12: 113158503-113158503 on Build GRCh38
rs999772499 Chr.12: 113158474-113158474 on Build GRCh38
rs564380412 Chr.12: 113158708-113158708 on Build GRCh38
rs973352365 Chr.12: 113158626-113158626 on Build GRCh38
rs371716079 Chr.12: 113158899-113158899 on Build GRCh38
rs769901613 Chr.12: 113158893-113158893 on Build GRCh38
rs370380296 Chr.12: 113158932-113158932 on Build GRCh38
rs958420938 Chr.12: 113158650-113158650 on Build GRCh38
rs555380008 Chr.12: 113158490-113158490 on Build GRCh38
rs747267485 Chr.12: 113158884-113158884 on Build GRCh38
rs146882826 Chr.12: 113158920-113158920 on Build GRCh38
rs534000886 Chr.12: 113158957-113158957 on Build GRCh38
rs2305901 Chr.12: 113158931-113158931 on Build GRCh38
rs749299982 Chr.12: 113158952-113158952 on Build GRCh38
rs976674067 Chr.12: 113158840-113158840 on Build GRCh38
rs935490753 Chr.12: 113158820-113158820 on Build GRCh38
rs775351819 Chr.12: 113158939-113158939 on Build GRCh38
rs1024995231 Chr.12: 113158641-113158641 on Build GRCh38
rs780478325 Chr.12: 113158873-113158873 on Build GRCh38
rs773970806 Chr.12: 113158843-113158843 on Build GRCh38
rs759532674 Chr.12: 113158851-113158851 on Build GRCh38
rs766502456 Chr.12: 113158859-113158859 on Build GRCh38
rs1050822280 Chr.12: 113158841-113158841 on Build GRCh38
rs567816792 Chr.12: 113158940-113158940 on Build GRCh38
rs760698052 Chr.12: 113158962-113158962 on Build GRCh38
rs151166999 Chr.12: 113158953-113158953 on Build GRCh38
rs556181766 Chr.12: 113158637-113158637 on Build GRCh38
rs748632245 Chr.12: 113158892-113158892 on Build GRCh38
rs753571951 Chr.12: 113158871-113158871 on Build GRCh38
rs551820098 Chr.12: 113158752-113158752 on Build GRCh38
rs528741840 Chr.12: 113158814-113158814 on Build GRCh38
rs763715983 Chr.12: 113158915-113158915 on Build GRCh38
rs775373047 Chr.12: 113158959-113158959 on Build GRCh38
rs367983164 Chr.12: 113158890-113158890 on Build GRCh38
rs987309600 Chr.12: 113158554-113158554 on Build GRCh38
rs954927257 Chr.12: 113158550-113158550 on Build GRCh38
rs760074241 Chr.12: 113158952-113158954 on Build GRCh38
rs370739347 Chr.12: 113158859-113158859 on Build GRCh38
rs767293812 Chr.12: 113158854-113158854 on Build GRCh38
rs551589253 Chr.12: 113158951-113158951 on Build GRCh38
rs546375251 Chr.12: 113158707-113158707 on Build GRCh38
rs1055826657 Chr.12: 113158595-113158595 on Build GRCh38
rs769454189 Chr.12: 113158530-113158530 on Build GRCh38
rs773237388 Chr.12: 113158897-113158897 on Build GRCh38
rs143051939 Chr.12: 113158933-113158933 on Build GRCh38
rs893837846 Chr.12: 113158517-113158517 on Build GRCh38
rs528474548 Chr.12: 113158680-113158680 on Build GRCh38
rs762939741 Chr.12: 113158835-113158835 on Build GRCh38
rs35865215 Chr.12: 113158516-113158517 on Build GRCh38
rs759318593 Chr.12: 113158908-113158908 on Build GRCh38
rs763679173 Chr.12: 113158866-113158866 on Build GRCh38
rs774364625 Chr.12: 113158917-113158917 on Build GRCh38
rs374403009 Chr.12: 113158872-113158872 on Build GRCh38
rs771002131 Chr.12: 113158901-113158901 on Build GRCh38
rs529251320 Chr.12: 113158912-113158912 on Build GRCh38
rs775014742 Chr.12: 113158901-113158903 on Build GRCh38
rs1032550150 Chr.12: 113158502-113158502 on Build GRCh38

View Full Product Details