Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132621152-132621649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RX2
Entrez Gene ID
22953
Gene Name
purinergic receptor P2X 2
Gene Aliases
DFNA41, P2X2
UniGene
Hs.258580
Gene Chromosome Location
Chr.12: 132618780-132623678 on Build GRCh38
Transcripts
NM_001282164.1 NM_001282165.1 NM_012226.4 NM_016318.3 NM_170682.3 NM_170683.3 NM_174872.2 NM_174873.2 XM_005266154.3 XM_005266155.4 XM_005266156.4 XM_011534786.2 XM_017019035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736420 c.969C>T p.I323I Chr.12: 132621318-132621318 on Build GRCh38
5592345 c.991G>A p.G331R Chr.12: 132621340-132621340 on Build GRCh38
163222 c.1138C>G p.Q380E Chr.12: 132621616-132621616 on Build GRCh38
5658166 c.1116A>G p.L372L Chr.12: 132621594-132621594 on Build GRCh38
3980930 c.970C>T p.R324C Chr.12: 132621319-132621319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970612038 Chr.12: 132621325-132621325 on Build GRCh38
rs199514327 Chr.12: 132621574-132621574 on Build GRCh38
rs779057259 Chr.12: 132621347-132621347 on Build GRCh38
rs770350190 Chr.12: 132621573-132621574 on Build GRCh38
rs568178443 Chr.12: 132621213-132621213 on Build GRCh38
rs757748091 Chr.12: 132621242-132621242 on Build GRCh38
rs1046081833 Chr.12: 132621485-132621485 on Build GRCh38
rs760899409 Chr.12: 132621478-132621478 on Build GRCh38
rs751840679 Chr.12: 132621534-132621534 on Build GRCh38
rs761180947 Chr.12: 132621292-132621292 on Build GRCh38
rs749968596 Chr.12: 132621345-132621363 on Build GRCh38
rs749478120 Chr.12: 132621214-132621214 on Build GRCh38
rs375236332 Chr.12: 132621248-132621248 on Build GRCh38
rs774082228 Chr.12: 132621398-132621398 on Build GRCh38
rs377522604 Chr.12: 132621426-132621426 on Build GRCh38
rs1006401732 Chr.12: 132621306-132621306 on Build GRCh38
rs150293706 Chr.12: 132621296-132621296 on Build GRCh38
rs34916191 Chr.12: 132621477-132621478 on Build GRCh38
rs755384404 Chr.12: 132621435-132621439 on Build GRCh38
rs772965391 Chr.12: 132621469-132621469 on Build GRCh38
rs184957114 Chr.12: 132621405-132621405 on Build GRCh38
rs576255022 Chr.12: 132621428-132621428 on Build GRCh38
rs775460016 Chr.12: 132621532-132621532 on Build GRCh38
rs371375307 Chr.12: 132621446-132621446 on Build GRCh38
rs769286408 Chr.12: 132621312-132621312 on Build GRCh38
rs1009088993 Chr.12: 132621364-132621364 on Build GRCh38
rs561894191 Chr.12: 132621487-132621487 on Build GRCh38
rs746677628 Chr.12: 132621578-132621578 on Build GRCh38
rs745794550 Chr.12: 132621297-132621297 on Build GRCh38
rs142799729 Chr.12: 132621573-132621573 on Build GRCh38
rs546130643 Chr.12: 132621415-132621415 on Build GRCh38
rs768316971 Chr.12: 132621397-132621397 on Build GRCh38
rs957011050 Chr.12: 132621288-132621288 on Build GRCh38
rs760530921 Chr.12: 132621221-132621221 on Build GRCh38
rs145966209 Chr.12: 132621558-132621558 on Build GRCh38
rs755133605 Chr.12: 132621343-132621343 on Build GRCh38
rs564207157 Chr.12: 132621588-132621588 on Build GRCh38
rs879120695 Chr.12: 132621457-132621457 on Build GRCh38
rs377665585 Chr.12: 132621442-132621442 on Build GRCh38
rs1018815433 Chr.12: 132621579-132621579 on Build GRCh38
rs747238517 Chr.12: 132621209-132621209 on Build GRCh38
rs769591887 Chr.12: 132621378-132621378 on Build GRCh38
rs201474543 Chr.12: 132621313-132621313 on Build GRCh38
rs771140743 Chr.12: 132621580-132621580 on Build GRCh38
rs771820471 Chr.12: 132621456-132621456 on Build GRCh38
rs139357505 Chr.12: 132621332-132621332 on Build GRCh38
rs775380392 Chr.12: 132621270-132621270 on Build GRCh38
rs1019230662 Chr.12: 132621263-132621265 on Build GRCh38
rs532793633 Chr.12: 132621601-132621601 on Build GRCh38
rs779818525 Chr.12: 132621424-132621425 on Build GRCh38
rs755150600 Chr.12: 132621278-132621278 on Build GRCh38
rs372363857 Chr.12: 132621293-132621293 on Build GRCh38
rs1045973450 Chr.12: 132621303-132621303 on Build GRCh38
rs752766258 Chr.12: 132621527-132621527 on Build GRCh38
rs533763078 Chr.12: 132621438-132621438 on Build GRCh38
rs1024862893 Chr.12: 132621191-132621191 on Build GRCh38
rs780975481 Chr.12: 132621368-132621368 on Build GRCh38
rs959046161 Chr.12: 132621188-132621188 on Build GRCh38
rs746712149 Chr.12: 132621430-132621439 on Build GRCh38
rs754390680 Chr.12: 132621424-132621424 on Build GRCh38
rs765216938 Chr.12: 132621618-132621618 on Build GRCh38
rs199942142 Chr.12: 132621372-132621372 on Build GRCh38
rs369983878 Chr.12: 132621279-132621279 on Build GRCh38
rs796319023 Chr.12: 132621471-132621471 on Build GRCh38
rs964426587 Chr.12: 132621404-132621404 on Build GRCh38
rs142034219 Chr.12: 132621342-132621342 on Build GRCh38
rs764681884 Chr.12: 132621230-132621230 on Build GRCh38
rs115331525 Chr.12: 132621271-132621271 on Build GRCh38
rs762568711 Chr.12: 132621319-132621319 on Build GRCh38
rs762412709 Chr.12: 132621568-132621568 on Build GRCh38
rs756566127 Chr.12: 132621335-132621335 on Build GRCh38
rs762984068 Chr.12: 132621595-132621595 on Build GRCh38
rs71452530 Chr.12: 132621222-132621222 on Build GRCh38
rs4883632 Chr.12: 132621435-132621435 on Build GRCh38
rs766978115 Chr.12: 132621338-132621338 on Build GRCh38
rs747727554 Chr.12: 132621453-132621453 on Build GRCh38
rs970580953 Chr.12: 132621195-132621195 on Build GRCh38
rs753713380 Chr.12: 132621214-132621214 on Build GRCh38
rs562797244 Chr.12: 132621185-132621185 on Build GRCh38
rs1002249416 Chr.12: 132621212-132621212 on Build GRCh38
rs765786338 Chr.12: 132621516-132621516 on Build GRCh38
rs545816836 Chr.12: 132621184-132621184 on Build GRCh38
rs764240633 Chr.12: 132621599-132621599 on Build GRCh38
rs141195683 Chr.12: 132621246-132621246 on Build GRCh38
rs202138002 Chr.12: 132621535-132621535 on Build GRCh38
rs754870259 Chr.12: 132621614-132621614 on Build GRCh38
rs757221359 Chr.12: 132621271-132621272 on Build GRCh38
rs766773574 Chr.12: 132621414-132621414 on Build GRCh38
rs371976818 Chr.12: 132621179-132621179 on Build GRCh38
rs772978182 Chr.12: 132621217-132621217 on Build GRCh38
rs116655914 Chr.12: 132621326-132621326 on Build GRCh38
rs147592928 Chr.12: 132621320-132621320 on Build GRCh38
rs374044513 Chr.12: 132621436-132621436 on Build GRCh38
rs930667299 Chr.12: 132621519-132621519 on Build GRCh38
rs756977093 Chr.12: 132621367-132621367 on Build GRCh38
rs76780477 Chr.12: 132621331-132621331 on Build GRCh38
rs757839919 Chr.12: 132621174-132621174 on Build GRCh38
rs763910590 Chr.12: 132621241-132621241 on Build GRCh38
rs760046758 Chr.12: 132621513-132621513 on Build GRCh38
rs771844396 Chr.12: 132621216-132621216 on Build GRCh38
rs746294218 Chr.12: 132621581-132621581 on Build GRCh38
rs758081130 Chr.12: 132621361-132621361 on Build GRCh38
rs777248624 Chr.12: 132621576-132621576 on Build GRCh38
rs1019516064 Chr.12: 132621365-132621365 on Build GRCh38
rs368580008 Chr.12: 132621411-132621411 on Build GRCh38
rs755964152 Chr.12: 132621439-132621439 on Build GRCh38
rs994936349 Chr.12: 132621231-132621231 on Build GRCh38
rs368169495 Chr.12: 132621224-132621224 on Build GRCh38
rs762231212 Chr.12: 132621327-132621327 on Build GRCh38
rs768000181 Chr.12: 132621328-132621328 on Build GRCh38
rs756154743 Chr.12: 132621262-132621262 on Build GRCh38
rs771356090 Chr.12: 132621479-132621479 on Build GRCh38
rs770251313 Chr.12: 132621584-132621584 on Build GRCh38
rs778226010 Chr.12: 132621330-132621336 on Build GRCh38
rs748397443 Chr.12: 132621353-132621353 on Build GRCh38
rs779913347 Chr.12: 132621549-132621549 on Build GRCh38
rs748700412 Chr.12: 132621384-132621384 on Build GRCh38
rs547555477 Chr.12: 132621575-132621575 on Build GRCh38
rs762522914 Chr.12: 132621240-132621240 on Build GRCh38
rs906167465 Chr.12: 132621497-132621497 on Build GRCh38
rs1012579569 Chr.12: 132621190-132621190 on Build GRCh38
rs191630133 Chr.12: 132621203-132621203 on Build GRCh38
rs769788937 Chr.12: 132621211-132621211 on Build GRCh38
rs373455554 Chr.12: 132621600-132621600 on Build GRCh38
rs556199359 Chr.12: 132621285-132621285 on Build GRCh38
rs775551691 Chr.12: 132621380-132621380 on Build GRCh38
rs763073673 Chr.12: 132621531-132621531 on Build GRCh38
rs373994917 Chr.12: 132621204-132621204 on Build GRCh38
rs886516991 Chr.12: 132621361-132621361 on Build GRCh38
rs199877925 Chr.12: 132621228-132621228 on Build GRCh38
rs761573086 Chr.12: 132621403-132621403 on Build GRCh38
rs577212217 Chr.12: 132621406-132621406 on Build GRCh38
rs776597290 Chr.12: 132621227-132621227 on Build GRCh38
rs138785836 Chr.12: 132621567-132621567 on Build GRCh38
rs775138313 Chr.12: 132621233-132621233 on Build GRCh38
rs781670792 Chr.12: 132621441-132621445 on Build GRCh38
rs976750532 Chr.12: 132621606-132621606 on Build GRCh38
rs1030854398 Chr.12: 132621366-132621366 on Build GRCh38
rs989975869 Chr.12: 132621392-132621392 on Build GRCh38
rs748539033 Chr.12: 132621441-132621444 on Build GRCh38
rs116100744 Chr.12: 132621323-132621323 on Build GRCh38

View Full Product Details