Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 133057651-133058154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF84
Entrez Gene ID
7637
Gene Name
zinc finger protein 84
Gene Aliases
HPF2
UniGene
Hs.445019
Gene Chromosome Location
Chr.12: 133037286-133063304 on Build GRCh38
Transcripts
NM_001127372.2 NM_001289971.1 NM_001289972.1 NM_003428.5 XM_005266185.1 XM_005266186.1 XM_011534832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639083 c.1161G>T p.K387N Chr.12: 133057876-133057876 on Build GRCh38
5625614 c.1171A>T p.I391F Chr.12: 133057886-133057886 on Build GRCh38
6478634 c.1249A>T p.S417C Chr.12: 133057964-133057964 on Build GRCh38
3954460 c.1208A>T p.Y403F Chr.12: 133057923-133057923 on Build GRCh38
6264876 c.936A>T p.T312T Chr.12: 133057651-133057651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929102729 Chr.12: 133057901-133057901 on Build GRCh38
rs623100 Chr.12: 133057864-133057864 on Build GRCh38
rs76518979 Chr.12: 133057717-133057717 on Build GRCh38
rs1032350692 Chr.12: 133057741-133057741 on Build GRCh38
rs1032465252 Chr.12: 133058106-133058106 on Build GRCh38
rs896126487 Chr.12: 133057987-133057987 on Build GRCh38
rs1000955828 Chr.12: 133058100-133058100 on Build GRCh38
rs113644344 Chr.12: 133057680-133057680 on Build GRCh38
rs200729061 Chr.12: 133057973-133057973 on Build GRCh38
rs937753611 Chr.12: 133057963-133057963 on Build GRCh38
rs954137006 Chr.12: 133057742-133057742 on Build GRCh38
rs919190463 Chr.12: 133057900-133057900 on Build GRCh38
rs909077526 Chr.12: 133057933-133057933 on Build GRCh38
rs200987966 Chr.12: 133058092-133058092 on Build GRCh38
rs796855951 Chr.12: 133057843-133057843 on Build GRCh38
rs912550798 Chr.12: 133057836-133057836 on Build GRCh38
rs984628987 Chr.12: 133057932-133057932 on Build GRCh38
rs370116710 Chr.12: 133057934-133057934 on Build GRCh38
rs902777149 Chr.12: 133058095-133058095 on Build GRCh38
rs949092833 Chr.12: 133058047-133058047 on Build GRCh38
rs61735098 Chr.12: 133057801-133057801 on Build GRCh38
rs1042593584 Chr.12: 133058089-133058089 on Build GRCh38

View Full Product Details