Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11675739-11676229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAD2L2
Entrez Gene ID
10459
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Gene Aliases
MAD2B, POLZ2, REV7
UniGene
Hs.19400
Gene Chromosome Location
Chr.1: 11674480-11691788 on Build GRCh38
Transcripts
NM_001127325.1 NM_006341.3 XM_011540507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4866281 c.394G>A p.V132M Chr.1: 11676118-11676118 on Build GRCh38
6006486 c.319A>T p.S107C Chr.1: 11676082-11676082 on Build GRCh38
2178621 c.370C>T p.R124W Chr.1: 11676103-11676103 on Build GRCh38
5377341 c.514_515CC>TT p.P172L Chr.1: 11676048-11676049 on Build GRCh38
1333017 c.425delC p.P142fs*45 Chr.1: 11676048-11676048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771133569 Chr.1: 11675932-11675932 on Build GRCh38
rs1020003612 Chr.1: 11676109-11676109 on Build GRCh38
rs764536092 Chr.1: 11676080-11676080 on Build GRCh38
rs553279720 Chr.1: 11676068-11676068 on Build GRCh38
rs753066223 Chr.1: 11675896-11675896 on Build GRCh38
rs766981615 Chr.1: 11675978-11675978 on Build GRCh38
rs200543059 Chr.1: 11675995-11675995 on Build GRCh38
rs771958137 Chr.1: 11676044-11676044 on Build GRCh38
rs762740385 Chr.1: 11675954-11675954 on Build GRCh38
rs569783386 Chr.1: 11675832-11675832 on Build GRCh38
rs773139319 Chr.1: 11676048-11676048 on Build GRCh38
rs769764556 Chr.1: 11676029-11676029 on Build GRCh38
rs375899265 Chr.1: 11675834-11675834 on Build GRCh38
rs952968051 Chr.1: 11675874-11675874 on Build GRCh38
rs548061357 Chr.1: 11675798-11675798 on Build GRCh38
rs765865207 Chr.1: 11676059-11676059 on Build GRCh38
rs771992289 Chr.1: 11676067-11676067 on Build GRCh38
rs377271043 Chr.1: 11676188-11676188 on Build GRCh38
rs374864885 Chr.1: 11675997-11675997 on Build GRCh38
rs768348660 Chr.1: 11675967-11675967 on Build GRCh38
rs747020613 Chr.1: 11675776-11675776 on Build GRCh38
rs765889245 Chr.1: 11675906-11675907 on Build GRCh38
rs758780314 Chr.1: 11675996-11675996 on Build GRCh38
rs537477828 Chr.1: 11675933-11675933 on Build GRCh38
rs750068072 Chr.1: 11675894-11675895 on Build GRCh38
rs2233021 Chr.1: 11676006-11676006 on Build GRCh38
rs558634063 Chr.1: 11675974-11675974 on Build GRCh38
rs536427641 Chr.1: 11675815-11675815 on Build GRCh38
rs775281295 Chr.1: 11676187-11676187 on Build GRCh38
rs369306059 Chr.1: 11676033-11676033 on Build GRCh38
rs755830715 Chr.1: 11675893-11675893 on Build GRCh38
rs771152011 Chr.1: 11675780-11675780 on Build GRCh38
rs2233023 Chr.1: 11675805-11675805 on Build GRCh38
rs1008256560 Chr.1: 11676109-11676111 on Build GRCh38
rs143857885 Chr.1: 11675767-11675767 on Build GRCh38
rs1044217666 Chr.1: 11675897-11675897 on Build GRCh38
rs757153898 Chr.1: 11675985-11675985 on Build GRCh38
rs774257384 Chr.1: 11675970-11675970 on Build GRCh38
rs760176047 Chr.1: 11676052-11676052 on Build GRCh38
rs971376147 Chr.1: 11676196-11676196 on Build GRCh38
rs757723800 Chr.1: 11676102-11676102 on Build GRCh38
rs952916219 Chr.1: 11676186-11676186 on Build GRCh38
rs775603631 Chr.1: 11675943-11675943 on Build GRCh38
rs765270200 Chr.1: 11675945-11675945 on Build GRCh38
rs778368042 Chr.1: 11675902-11675902 on Build GRCh38
rs779596380 Chr.1: 11675895-11675895 on Build GRCh38
rs751500020 Chr.1: 11675840-11675840 on Build GRCh38
rs755242044 Chr.1: 11676143-11676143 on Build GRCh38
rs750071876 Chr.1: 11675892-11675892 on Build GRCh38
rs373373788 Chr.1: 11676163-11676163 on Build GRCh38
rs746989382 Chr.1: 11676167-11676167 on Build GRCh38
rs905278384 Chr.1: 11675905-11675905 on Build GRCh38
rs577078082 Chr.1: 11675984-11675984 on Build GRCh38
rs377766985 Chr.1: 11676168-11676168 on Build GRCh38
rs761716384 Chr.1: 11676039-11676039 on Build GRCh38
rs1019718343 Chr.1: 11675831-11675831 on Build GRCh38
rs747580606 Chr.1: 11675926-11675926 on Build GRCh38
rs774171698 Chr.1: 11676038-11676038 on Build GRCh38
rs747871567 Chr.1: 11676155-11676155 on Build GRCh38
rs920071698 Chr.1: 11675851-11675851 on Build GRCh38
rs770005189 Chr.1: 11675942-11675942 on Build GRCh38
rs981145683 Chr.1: 11676201-11676201 on Build GRCh38
rs368583657 Chr.1: 11675835-11675835 on Build GRCh38
rs976260508 Chr.1: 11676105-11676105 on Build GRCh38
rs1004076478 Chr.1: 11675907-11675907 on Build GRCh38
rs55785094 Chr.1: 11675789-11675789 on Build GRCh38
rs755660515 Chr.1: 11675901-11675903 on Build GRCh38
rs757891395 Chr.1: 11676007-11676007 on Build GRCh38
rs759111242 Chr.1: 11676175-11676175 on Build GRCh38
rs780062910 Chr.1: 11676131-11676131 on Build GRCh38
rs542007676 Chr.1: 11676119-11676119 on Build GRCh38
rs779137723 Chr.1: 11676153-11676153 on Build GRCh38
rs571911139 Chr.1: 11675768-11675768 on Build GRCh38
rs371504576 Chr.1: 11676028-11676028 on Build GRCh38
rs767069540 Chr.1: 11675858-11675858 on Build GRCh38
rs2233020 Chr.1: 11676074-11676074 on Build GRCh38
rs745917324 Chr.1: 11676027-11676027 on Build GRCh38
rs764049519 Chr.1: 11675836-11675836 on Build GRCh38
rs749494169 Chr.1: 11676139-11676139 on Build GRCh38
rs2233022 Chr.1: 11675820-11675820 on Build GRCh38
rs372152375 Chr.1: 11676176-11676176 on Build GRCh38
rs752413643 Chr.1: 11675863-11675863 on Build GRCh38
rs781769370 Chr.1: 11676013-11676013 on Build GRCh38
rs756203983 Chr.1: 11676130-11676130 on Build GRCh38
rs200815563 Chr.1: 11676169-11676169 on Build GRCh38
rs934380933 Chr.1: 11676034-11676034 on Build GRCh38
rs372997698 Chr.1: 11676036-11676036 on Build GRCh38
rs772582068 Chr.1: 11675763-11675763 on Build GRCh38
rs35692795 Chr.1: 11675759-11675760 on Build GRCh38
rs751926997 Chr.1: 11676099-11676099 on Build GRCh38
rs753237679 Chr.1: 11676031-11676031 on Build GRCh38
rs759908670 Chr.1: 11675828-11675828 on Build GRCh38
rs2233024 Chr.1: 11675784-11675784 on Build GRCh38
rs746069242 Chr.1: 11675941-11675941 on Build GRCh38
rs756697500 Chr.1: 11675843-11675843 on Build GRCh38
rs182391652 Chr.1: 11676191-11676191 on Build GRCh38

View Full Product Details