Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11858620-11859129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPPB
Entrez Gene ID
4879
Gene Name
natriuretic peptide B
Gene Aliases
BNP
UniGene
Hs.219140
Gene Chromosome Location
Chr.1: 11857464-11858935 on Build GRCh38
Transcripts
NM_002521.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747670 c.78C>T p.S26S Chr.1: 11858756-11858756 on Build GRCh38
6728814 c.41T>C p.L14P Chr.1: 11858793-11858793 on Build GRCh38
5377382 c.26G>A p.R9Q Chr.1: 11858808-11858808 on Build GRCh38
384176 c.87G>T p.L29L Chr.1: 11858747-11858747 on Build GRCh38
5418018 c.33C>T p.L11L Chr.1: 11858801-11858801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61761992 Chr.1: 11858669-11858669 on Build GRCh38
rs761041289 Chr.1: 11858690-11858690 on Build GRCh38
rs767024987 Chr.1: 11858693-11858693 on Build GRCh38
rs78453499 Chr.1: 11858861-11858861 on Build GRCh38
rs374516242 Chr.1: 11858989-11858989 on Build GRCh38
rs763718693 Chr.1: 11858804-11858804 on Build GRCh38
rs779624438 Chr.1: 11858738-11858738 on Build GRCh38
rs13305989 Chr.1: 11858884-11858884 on Build GRCh38
rs767435696 Chr.1: 11858871-11858873 on Build GRCh38
rs960137954 Chr.1: 11859077-11859077 on Build GRCh38
rs975843890 Chr.1: 11858932-11858932 on Build GRCh38
rs534905983 Chr.1: 11859038-11859038 on Build GRCh38
rs372837034 Chr.1: 11858851-11858851 on Build GRCh38
rs987052872 Chr.1: 11858978-11858978 on Build GRCh38
rs200478951 Chr.1: 11858659-11858659 on Build GRCh38
rs908759708 Chr.1: 11858979-11858979 on Build GRCh38
rs781699099 Chr.1: 11858858-11858858 on Build GRCh38
rs5227 Chr.1: 11858760-11858760 on Build GRCh38
rs746326008 Chr.1: 11858859-11858859 on Build GRCh38
rs760123248 Chr.1: 11858767-11858767 on Build GRCh38
rs765544521 Chr.1: 11858709-11858709 on Build GRCh38
rs751994081 Chr.1: 11858702-11858703 on Build GRCh38
rs541881461 Chr.1: 11858674-11858674 on Build GRCh38
rs779385648 Chr.1: 11858821-11858821 on Build GRCh38
rs992787386 Chr.1: 11859078-11859078 on Build GRCh38
rs745418315 Chr.1: 11858666-11858666 on Build GRCh38
rs977496484 Chr.1: 11858665-11858665 on Build GRCh38
rs1016474512 Chr.1: 11859056-11859056 on Build GRCh38
rs777336949 Chr.1: 11858766-11858766 on Build GRCh38
rs750602581 Chr.1: 11858854-11858855 on Build GRCh38
rs899018888 Chr.1: 11859060-11859060 on Build GRCh38
rs1020806775 Chr.1: 11858855-11858855 on Build GRCh38
rs751286475 Chr.1: 11858797-11858797 on Build GRCh38
rs759066860 Chr.1: 11858871-11858879 on Build GRCh38
rs185200240 Chr.1: 11858882-11858882 on Build GRCh38
rs764412274 Chr.1: 11858792-11858792 on Build GRCh38
rs151222918 Chr.1: 11858737-11858737 on Build GRCh38
rs757672495 Chr.1: 11858794-11858795 on Build GRCh38
rs946206842 Chr.1: 11858679-11858679 on Build GRCh38
rs1039076006 Chr.1: 11858726-11858726 on Build GRCh38
rs1013515063 Chr.1: 11858834-11858834 on Build GRCh38
rs779881008 Chr.1: 11858698-11858698 on Build GRCh38
rs148748522 Chr.1: 11859063-11859063 on Build GRCh38
rs904914597 Chr.1: 11859012-11859012 on Build GRCh38
rs35628673 Chr.1: 11858780-11858780 on Build GRCh38
rs192354362 Chr.1: 11858944-11858944 on Build GRCh38
rs748837558 Chr.1: 11858739-11858739 on Build GRCh38
rs370520733 Chr.1: 11858654-11858654 on Build GRCh38
rs879235282 Chr.1: 11858876-11858877 on Build GRCh38
rs781514352 Chr.1: 11858853-11858854 on Build GRCh38
rs763044821 Chr.1: 11858789-11858789 on Build GRCh38
rs199794109 Chr.1: 11858720-11858720 on Build GRCh38
rs781153609 Chr.1: 11858717-11858717 on Build GRCh38
rs932958093 Chr.1: 11858970-11858970 on Build GRCh38
rs201091040 Chr.1: 11858860-11858860 on Build GRCh38
rs570748462 Chr.1: 11858961-11858961 on Build GRCh38
rs1028312184 Chr.1: 11858965-11858965 on Build GRCh38
rs754556720 Chr.1: 11858836-11858836 on Build GRCh38
rs35587340 Chr.1: 11858754-11858755 on Build GRCh38
rs1042294176 Chr.1: 11859087-11859087 on Build GRCh38
rs749330564 Chr.1: 11858855-11858872 on Build GRCh38
rs1054778790 Chr.1: 11858815-11858815 on Build GRCh38
rs764122778 Chr.1: 11858873-11858874 on Build GRCh38
rs543699023 Chr.1: 11858806-11858806 on Build GRCh38
rs35519114 Chr.1: 11858870-11858871 on Build GRCh38
rs768607285 Chr.1: 11858855-11858878 on Build GRCh38
rs756271597 Chr.1: 11858855-11858857 on Build GRCh38
rs767245197 Chr.1: 11858885-11858885 on Build GRCh38
rs772261102 Chr.1: 11858680-11858680 on Build GRCh38
rs1038800387 Chr.1: 11858909-11858909 on Build GRCh38
rs778676397 Chr.1: 11858850-11858850 on Build GRCh38
rs921552050 Chr.1: 11858725-11858725 on Build GRCh38
rs902432808 Chr.1: 11859101-11859101 on Build GRCh38
rs61761994 Chr.1: 11858869-11858870 on Build GRCh38
rs919697302 Chr.1: 11859033-11859033 on Build GRCh38
rs936378921 Chr.1: 11858903-11858903 on Build GRCh38
rs562497263 Chr.1: 11858871-11858871 on Build GRCh38
rs574844256 Chr.1: 11858655-11858655 on Build GRCh38
rs564298750 Chr.1: 11858753-11858753 on Build GRCh38
rs770550563 Chr.1: 11858871-11858876 on Build GRCh38
rs900137081 Chr.1: 11858954-11858954 on Build GRCh38
rs144265843 Chr.1: 11858805-11858805 on Build GRCh38
rs927326575 Chr.1: 11858644-11858644 on Build GRCh38
rs879124621 Chr.1: 11859002-11859002 on Build GRCh38
rs949034616 Chr.1: 11859085-11859085 on Build GRCh38
rs60943484 Chr.1: 11858892-11858893 on Build GRCh38
rs940204476 Chr.1: 11858988-11858988 on Build GRCh38
rs1034427690 Chr.1: 11859075-11859075 on Build GRCh38
rs759741420 Chr.1: 11858695-11858695 on Build GRCh38
rs967570338 Chr.1: 11859103-11859103 on Build GRCh38
rs768381227 Chr.1: 11858748-11858748 on Build GRCh38
rs571539855 Chr.1: 11858858-11858859 on Build GRCh38
rs1000622449 Chr.1: 11858870-11858870 on Build GRCh38
rs955527979 Chr.1: 11858972-11858972 on Build GRCh38
rs753423994 Chr.1: 11858830-11858830 on Build GRCh38
rs148807468 Chr.1: 11858809-11858809 on Build GRCh38
rs1055252012 Chr.1: 11859059-11859059 on Build GRCh38
rs140816420 Chr.1: 11858714-11858714 on Build GRCh38
rs771762194 Chr.1: 11858861-11858862 on Build GRCh38
rs771544495 Chr.1: 11858762-11858762 on Build GRCh38
rs768745177 Chr.1: 11858875-11858875 on Build GRCh38
rs1020276610 Chr.1: 11859081-11859081 on Build GRCh38
rs765349605 Chr.1: 11858771-11858771 on Build GRCh38
rs61761993 Chr.1: 11858757-11858757 on Build GRCh38
rs150360910 Chr.1: 11858759-11858759 on Build GRCh38
rs926150796 Chr.1: 11858902-11858902 on Build GRCh38
rs763747066 Chr.1: 11858711-11858711 on Build GRCh38
rs778803776 Chr.1: 11858855-11858881 on Build GRCh38
rs748544214 Chr.1: 11858676-11858676 on Build GRCh38
rs758657926 Chr.1: 11858710-11858710 on Build GRCh38
rs528934604 Chr.1: 11858857-11858857 on Build GRCh38
rs957474156 Chr.1: 11858886-11858886 on Build GRCh38

View Full Product Details