Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26045750-26046290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHISA2
Entrez Gene ID
387914
Gene Name
shisa family member 2
Gene Aliases
C13orf13, PRO28631, TMEM46, WGAR9166, bA398O19.2, hShisa
UniGene
Hs.433791
Gene Chromosome Location
Chr.13: 26044597-26051060 on Build GRCh38
Transcripts
NM_001007538.1

SNP Detail

SNP ID Chromosome Location
rs549788003 Chr.13: 26046029-26046029 on Build GRCh38
rs57376936 Chr.13: 26046107-26046108 on Build GRCh38
rs778786754 Chr.13: 26046021-26046021 on Build GRCh38
rs916485303 Chr.13: 26045942-26045942 on Build GRCh38
rs371916687 Chr.13: 26046224-26046224 on Build GRCh38
rs866432716 Chr.13: 26045834-26045834 on Build GRCh38
rs983319776 Chr.13: 26046148-26046148 on Build GRCh38
rs9507640 Chr.13: 26045821-26045821 on Build GRCh38
rs138913454 Chr.13: 26046034-26046034 on Build GRCh38
rs757165434 Chr.13: 26045925-26045925 on Build GRCh38
rs763520217 Chr.13: 26046032-26046032 on Build GRCh38
rs561140334 Chr.13: 26045939-26045939 on Build GRCh38
rs56664482 Chr.13: 26046107-26046107 on Build GRCh38
rs60205166 Chr.13: 26046176-26046176 on Build GRCh38
rs904730784 Chr.13: 26045976-26045976 on Build GRCh38
rs113185350 Chr.13: 26045911-26045911 on Build GRCh38
rs1008884322 Chr.13: 26045831-26045831 on Build GRCh38
rs12871243 Chr.13: 26046083-26046083 on Build GRCh38
rs12874694 Chr.13: 26045865-26045865 on Build GRCh38
rs951916727 Chr.13: 26046145-26046145 on Build GRCh38
rs976796924 Chr.13: 26045946-26045946 on Build GRCh38
rs891499114 Chr.13: 26045820-26045820 on Build GRCh38
rs1000450572 Chr.13: 26045791-26045791 on Build GRCh38
rs904740876 Chr.13: 26045784-26045784 on Build GRCh38
rs936228768 Chr.13: 26045986-26045986 on Build GRCh38
rs951243502 Chr.13: 26046109-26046109 on Build GRCh38
rs192088754 Chr.13: 26046156-26046156 on Build GRCh38
rs932155741 Chr.13: 26046168-26046170 on Build GRCh38
rs745749374 Chr.13: 26046223-26046223 on Build GRCh38
rs899290857 Chr.13: 26045776-26045776 on Build GRCh38
rs1041995821 Chr.13: 26046248-26046248 on Build GRCh38
rs1001295225 Chr.13: 26046091-26046091 on Build GRCh38
rs576301337 Chr.13: 26045809-26045809 on Build GRCh38
rs1050311224 Chr.13: 26045971-26045971 on Build GRCh38
rs905577857 Chr.13: 26046087-26046087 on Build GRCh38
rs199527456 Chr.13: 26046082-26046082 on Build GRCh38
rs12870924 Chr.13: 26045918-26045918 on Build GRCh38
rs932758971 Chr.13: 26045970-26045970 on Build GRCh38
rs940054513 Chr.13: 26045941-26045941 on Build GRCh38
rs1023988576 Chr.13: 26046086-26046086 on Build GRCh38
rs559797006 Chr.13: 26045840-26045840 on Build GRCh38
rs969854404 Chr.13: 26045879-26045879 on Build GRCh38
rs9507641 Chr.13: 26046077-26046077 on Build GRCh38
rs12864686 Chr.13: 26045847-26045847 on Build GRCh38
rs980316181 Chr.13: 26045889-26045889 on Build GRCh38
rs981737272 Chr.13: 26046028-26046028 on Build GRCh38
rs985595166 Chr.13: 26046227-26046227 on Build GRCh38
rs1031337430 Chr.13: 26045794-26045794 on Build GRCh38
rs371756701 Chr.13: 26046063-26046063 on Build GRCh38
rs920350549 Chr.13: 26046165-26046165 on Build GRCh38
rs922630715 Chr.13: 26046183-26046183 on Build GRCh38
rs891447120 Chr.13: 26046046-26046046 on Build GRCh38
rs1027498532 Chr.13: 26045894-26045894 on Build GRCh38
rs879163753 Chr.13: 26045802-26045802 on Build GRCh38
rs144570734 Chr.13: 26046082-26046086 on Build GRCh38
rs1027443666 Chr.13: 26046104-26046104 on Build GRCh38
rs951926685 Chr.13: 26045900-26045900 on Build GRCh38
rs150656885 Chr.13: 26045968-26045968 on Build GRCh38
rs966001715 Chr.13: 26046166-26046166 on Build GRCh38
rs543448272 Chr.13: 26045808-26045808 on Build GRCh38
rs879606585 Chr.13: 26045794-26045795 on Build GRCh38
rs549236923 Chr.13: 26045783-26045784 on Build GRCh38
rs865984408 Chr.13: 26045843-26045843 on Build GRCh38
rs1053391997 Chr.13: 26046008-26046008 on Build GRCh38
rs200852086 Chr.13: 26046082-26046085 on Build GRCh38
rs1024496348 Chr.13: 26045869-26045869 on Build GRCh38

View Full Product Details