Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30952528-30953033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX26
Entrez Gene ID
122046
Gene Name
testis expressed 26
Gene Aliases
C13orf26
UniGene
Hs.124463
Gene Chromosome Location
Chr.13: 30932633-30975502 on Build GRCh38
Transcripts
NM_152325.1 XM_011534919.2 XM_011534921.2 XM_011534924.2 XM_011534925.2 XM_011534926.2 XM_011534927.2 XM_011534928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327459 c.164G>A p.R55K Chr.13: 30952677-30952677 on Build GRCh38
1578205 c.228C>G p.Y76* Chr.13: 30952741-30952741 on Build GRCh38
6377242 c.199C>A p.L67I Chr.13: 30952712-30952712 on Build GRCh38
5640642 c.301C>T p.H101Y Chr.13: 30952814-30952814 on Build GRCh38
5640975 c.223G>T p.E75* Chr.13: 30952736-30952736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748226294 Chr.13: 30952703-30952703 on Build GRCh38
rs781713482 Chr.13: 30952616-30952616 on Build GRCh38
rs368283305 Chr.13: 30952675-30952675 on Build GRCh38
rs539228649 Chr.13: 30952937-30952937 on Build GRCh38
rs557224914 Chr.13: 30952946-30952946 on Build GRCh38
rs768869321 Chr.13: 30952640-30952640 on Build GRCh38
rs763700448 Chr.13: 30952723-30952723 on Build GRCh38
rs956776826 Chr.13: 30952661-30952661 on Build GRCh38
rs1036642237 Chr.13: 30952834-30952834 on Build GRCh38
rs571759189 Chr.13: 30952912-30952912 on Build GRCh38
rs752482750 Chr.13: 30952657-30952657 on Build GRCh38
rs573419112 Chr.13: 30952634-30952634 on Build GRCh38
rs768565071 Chr.13: 30952649-30952649 on Build GRCh38
rs926865069 Chr.13: 30952983-30952983 on Build GRCh38
rs980912343 Chr.13: 30952967-30952967 on Build GRCh38
rs774302114 Chr.13: 30952649-30952649 on Build GRCh38
rs370787883 Chr.13: 30952656-30952656 on Build GRCh38
rs1005163209 Chr.13: 30952881-30952886 on Build GRCh38
rs760167234 Chr.13: 30952877-30952877 on Build GRCh38
rs761329101 Chr.13: 30952732-30952732 on Build GRCh38
rs937008436 Chr.13: 30952727-30952727 on Build GRCh38
rs370475342 Chr.13: 30952660-30952660 on Build GRCh38
rs776688539 Chr.13: 30952650-30952650 on Build GRCh38
rs368133357 Chr.13: 30952626-30952626 on Build GRCh38
rs1022909731 Chr.13: 30952948-30952948 on Build GRCh38
rs765186751 Chr.13: 30952715-30952715 on Build GRCh38
rs1018869054 Chr.13: 30952911-30952911 on Build GRCh38
rs142207829 Chr.13: 30952821-30952821 on Build GRCh38
rs951734802 Chr.13: 30952573-30952573 on Build GRCh38
rs7986039 Chr.13: 30952565-30952565 on Build GRCh38
rs769908618 Chr.13: 30952819-30952819 on Build GRCh38
rs757769152 Chr.13: 30952612-30952612 on Build GRCh38
rs772778738 Chr.13: 30952875-30952875 on Build GRCh38
rs772161327 Chr.13: 30952713-30952713 on Build GRCh38
rs779184166 Chr.13: 30952690-30952690 on Build GRCh38
rs1027506321 Chr.13: 30952566-30952566 on Build GRCh38
rs773964568 Chr.13: 30952863-30952863 on Build GRCh38
rs775788352 Chr.13: 30952632-30952632 on Build GRCh38
rs896252456 Chr.13: 30952920-30952920 on Build GRCh38
rs768524001 Chr.13: 30952843-30952843 on Build GRCh38
rs987406535 Chr.13: 30952570-30952570 on Build GRCh38
rs34650841 Chr.13: 30952648-30952649 on Build GRCh38
rs147583572 Chr.13: 30952741-30952741 on Build GRCh38
rs755350000 Chr.13: 30952760-30952760 on Build GRCh38
rs749411105 Chr.13: 30952618-30952621 on Build GRCh38
rs553808345 Chr.13: 30952607-30952607 on Build GRCh38
rs372590685 Chr.13: 30952724-30952724 on Build GRCh38
rs775491521 Chr.13: 30952722-30952722 on Build GRCh38
rs201587722 Chr.13: 30952655-30952655 on Build GRCh38
rs771215526 Chr.13: 30952635-30952639 on Build GRCh38
rs554917849 Chr.13: 30952577-30952577 on Build GRCh38
rs201530985 Chr.13: 30952816-30952816 on Build GRCh38
rs374192558 Chr.13: 30952666-30952666 on Build GRCh38
rs200748405 Chr.13: 30952862-30952862 on Build GRCh38
rs767032519 Chr.13: 30952739-30952739 on Build GRCh38
rs770110015 Chr.13: 30952630-30952630 on Build GRCh38
rs749074470 Chr.13: 30952831-30952831 on Build GRCh38
rs367679816 Chr.13: 30952870-30952870 on Build GRCh38
rs749864889 Chr.13: 30952826-30952826 on Build GRCh38
rs746460307 Chr.13: 30952611-30952611 on Build GRCh38
rs547160789 Chr.13: 30952879-30952879 on Build GRCh38
rs540576930 Chr.13: 30952595-30952595 on Build GRCh38
rs544867356 Chr.13: 30952726-30952726 on Build GRCh38
rs756459265 Chr.13: 30952890-30952890 on Build GRCh38
rs549374306 Chr.13: 30952779-30952779 on Build GRCh38
rs769787584 Chr.13: 30952721-30952721 on Build GRCh38
rs762463132 Chr.13: 30952842-30952843 on Build GRCh38
rs756568820 Chr.13: 30952676-30952676 on Build GRCh38
rs763796853 Chr.13: 30952664-30952664 on Build GRCh38
rs1023261115 Chr.13: 30952993-30952993 on Build GRCh38
rs900394756 Chr.13: 30952868-30952868 on Build GRCh38
rs774730897 Chr.13: 30952645-30952645 on Build GRCh38
rs895562298 Chr.13: 30952761-30952761 on Build GRCh38
rs145168779 Chr.13: 30952740-30952740 on Build GRCh38
rs373675908 Chr.13: 30952718-30952718 on Build GRCh38
rs995188800 Chr.13: 30952845-30952845 on Build GRCh38
rs533201895 Chr.13: 30952684-30952684 on Build GRCh38
rs753018964 Chr.13: 30952841-30952841 on Build GRCh38
rs530687411 Chr.13: 30952746-30952746 on Build GRCh38
rs776408182 Chr.13: 30952655-30952656 on Build GRCh38
rs756766798 Chr.13: 30952667-30952667 on Build GRCh38
rs115685246 Chr.13: 30952554-30952554 on Build GRCh38
rs569558128 Chr.13: 30952947-30952947 on Build GRCh38
rs376300171 Chr.13: 30952806-30952806 on Build GRCh38
rs780538166 Chr.13: 30952744-30952744 on Build GRCh38
rs780413623 Chr.13: 30952682-30952682 on Build GRCh38
rs748435471 Chr.13: 30952801-30952801 on Build GRCh38
rs968231260 Chr.13: 30952958-30952958 on Build GRCh38
rs772136496 Chr.13: 30952802-30952802 on Build GRCh38
rs755506986 Chr.13: 30952738-30952738 on Build GRCh38
rs761757966 Chr.13: 30952662-30952662 on Build GRCh38
rs761545165 Chr.13: 30952866-30952866 on Build GRCh38
rs773385855 Chr.13: 30952716-30952716 on Build GRCh38
rs1004641820 Chr.13: 30952643-30952643 on Build GRCh38

View Full Product Details