Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36283456-36283956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC169
Entrez Gene ID
728591
Gene Name
coiled-coil domain containing 169
Gene Aliases
C13orf38
UniGene
Hs.656745
Gene Chromosome Location
Chr.13: 36227042-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Gene Symbol
CCDC169-SOHLH2
Entrez Gene ID
100526761
Gene Name
CCDC169-SOHLH2 readthrough
Gene Aliases
C13orf38-SOHLH2
UniGene
Hs.124519
Gene Chromosome Location
Chr.13: 36168208-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4530431 c.169G>A p.E57K Chr.13: 36283697-36283697 on Build GRCh38
5924604 c.305G>A p.R102Q Chr.13: 36283479-36283479 on Build GRCh38
2072491 c.221A>T p.K74M Chr.13: 36283645-36283645 on Build GRCh38
5754070 c.293G>A p.R98H Chr.13: 36283491-36283491 on Build GRCh38
4637703 c.197T>G p.L66R Chr.13: 36283669-36283669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184516088 Chr.13: 36283492-36283492 on Build GRCh38
rs768712718 Chr.13: 36283684-36283684 on Build GRCh38
rs371059870 Chr.13: 36283491-36283491 on Build GRCh38
rs117239043 Chr.13: 36283869-36283869 on Build GRCh38
rs775652620 Chr.13: 36283593-36283593 on Build GRCh38
rs553108055 Chr.13: 36283680-36283680 on Build GRCh38
rs188966399 Chr.13: 36283903-36283903 on Build GRCh38
rs762625720 Chr.13: 36283569-36283570 on Build GRCh38
rs770333814 Chr.13: 36283665-36283665 on Build GRCh38
rs116384062 Chr.13: 36283575-36283575 on Build GRCh38
rs1998800 Chr.13: 36283502-36283502 on Build GRCh38
rs766682832 Chr.13: 36283687-36283706 on Build GRCh38
rs948989285 Chr.13: 36283602-36283602 on Build GRCh38
rs576651881 Chr.13: 36283787-36283787 on Build GRCh38
rs774666137 Chr.13: 36283741-36283741 on Build GRCh38
rs372487398 Chr.13: 36283606-36283606 on Build GRCh38
rs907441138 Chr.13: 36283673-36283673 on Build GRCh38
rs1011246773 Chr.13: 36283728-36283728 on Build GRCh38
rs563174047 Chr.13: 36283753-36283753 on Build GRCh38
rs1030274878 Chr.13: 36283791-36283791 on Build GRCh38
rs748477869 Chr.13: 36283564-36283564 on Build GRCh38
rs555569600 Chr.13: 36283565-36283565 on Build GRCh38
rs577712251 Chr.13: 36283685-36283685 on Build GRCh38
rs935165902 Chr.13: 36283810-36283810 on Build GRCh38
rs543728976 Chr.13: 36283824-36283824 on Build GRCh38
rs954771544 Chr.13: 36283797-36283797 on Build GRCh38
rs752313537 Chr.13: 36283697-36283697 on Build GRCh38
rs866575663 Chr.13: 36283618-36283618 on Build GRCh38
rs1031272799 Chr.13: 36283713-36283713 on Build GRCh38
rs373013864 Chr.13: 36283898-36283898 on Build GRCh38
rs767691232 Chr.13: 36283829-36283829 on Build GRCh38
rs974114673 Chr.13: 36283909-36283909 on Build GRCh38
rs1044716264 Chr.13: 36283838-36283838 on Build GRCh38
rs80273011 Chr.13: 36283483-36283483 on Build GRCh38
rs544981680 Chr.13: 36283711-36283711 on Build GRCh38
rs954800740 Chr.13: 36283744-36283744 on Build GRCh38
rs144027143 Chr.13: 36283493-36283493 on Build GRCh38
rs866129160 Chr.13: 36283598-36283598 on Build GRCh38
rs779037553 Chr.13: 36283527-36283527 on Build GRCh38
rs377181574 Chr.13: 36283580-36283580 on Build GRCh38
rs1011863216 Chr.13: 36283481-36283481 on Build GRCh38
rs762711678 Chr.13: 36283699-36283699 on Build GRCh38
rs763390270 Chr.13: 36283681-36283681 on Build GRCh38
rs763913888 Chr.13: 36283703-36283703 on Build GRCh38
rs751490178 Chr.13: 36283723-36283723 on Build GRCh38
rs547630832 Chr.13: 36283913-36283913 on Build GRCh38
rs1053585568 Chr.13: 36283814-36283814 on Build GRCh38
rs3818459 Chr.13: 36283879-36283879 on Build GRCh38
rs772616109 Chr.13: 36283573-36283573 on Build GRCh38
rs908517809 Chr.13: 36283840-36283840 on Build GRCh38
rs974764393 Chr.13: 36283737-36283737 on Build GRCh38
rs963664835 Chr.13: 36283715-36283715 on Build GRCh38
rs940065456 Chr.13: 36283540-36283540 on Build GRCh38
rs76335557 Chr.13: 36283657-36283657 on Build GRCh38

View Full Product Details