Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36329243-36329767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPG20
Entrez Gene ID
23111
Gene Name
spastic paraplegia 20 (Troyer syndrome)
Gene Aliases
SPARTIN, TAHCCP1
UniGene
Hs.440414
Gene Chromosome Location
Chr.13: 36301638-36370180 on Build GRCh38
Transcripts
NM_001142294.1 NM_001142295.1 NM_001142296.1 NM_015087.4 XM_005266313.4 XM_005266314.3 XM_005266315.3 XM_005266317.3 XM_011535012.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813764 c.1064G>C p.R355T Chr.13: 36329462-36329462 on Build GRCh38
2072519 c.1162delA p.R388fs*12 Chr.13: 36329364-36329364 on Build GRCh38
5842462 c.1058G>A p.R353K Chr.13: 36329468-36329468 on Build GRCh38
1366673 c.1069T>G p.S357A Chr.13: 36329457-36329457 on Build GRCh38
6340474 c.1010C>A p.A337D Chr.13: 36329516-36329516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371489379 Chr.13: 36329549-36329549 on Build GRCh38
rs145026626 Chr.13: 36329414-36329414 on Build GRCh38
rs774429552 Chr.13: 36329557-36329557 on Build GRCh38
rs925200824 Chr.13: 36329688-36329688 on Build GRCh38
rs761054221 Chr.13: 36329429-36329429 on Build GRCh38
rs566134472 Chr.13: 36329292-36329292 on Build GRCh38
rs762149978 Chr.13: 36329406-36329406 on Build GRCh38
rs926596702 Chr.13: 36329599-36329599 on Build GRCh38
rs567567114 Chr.13: 36329636-36329636 on Build GRCh38
rs368305530 Chr.13: 36329399-36329399 on Build GRCh38
rs746750857 Chr.13: 36329498-36329498 on Build GRCh38
rs774428673 Chr.13: 36329402-36329402 on Build GRCh38
rs1004712501 Chr.13: 36329722-36329722 on Build GRCh38
rs913700897 Chr.13: 36329667-36329667 on Build GRCh38
rs753299865 Chr.13: 36329341-36329341 on Build GRCh38
rs1002220389 Chr.13: 36329279-36329279 on Build GRCh38
rs778399570 Chr.13: 36329351-36329351 on Build GRCh38
rs772236125 Chr.13: 36329568-36329568 on Build GRCh38
rs754442567 Chr.13: 36329438-36329438 on Build GRCh38
rs962623619 Chr.13: 36329340-36329340 on Build GRCh38
rs757165845 Chr.13: 36329501-36329501 on Build GRCh38
rs748210013 Chr.13: 36329562-36329562 on Build GRCh38
rs368153772 Chr.13: 36329370-36329370 on Build GRCh38
rs956021462 Chr.13: 36329387-36329387 on Build GRCh38
rs878854993 Chr.13: 36329473-36329473 on Build GRCh38
rs746802252 Chr.13: 36329388-36329388 on Build GRCh38
rs148672324 Chr.13: 36329396-36329396 on Build GRCh38
rs1056415136 Chr.13: 36329668-36329668 on Build GRCh38
rs372578966 Chr.13: 36329401-36329401 on Build GRCh38
rs745880782 Chr.13: 36329530-36329530 on Build GRCh38
rs960700751 Chr.13: 36329615-36329615 on Build GRCh38
rs199556543 Chr.13: 36329373-36329373 on Build GRCh38
rs369445563 Chr.13: 36329317-36329317 on Build GRCh38
rs534756891 Chr.13: 36329710-36329710 on Build GRCh38
rs754570040 Chr.13: 36329344-36329344 on Build GRCh38
rs912293884 Chr.13: 36329379-36329379 on Build GRCh38
rs762094058 Chr.13: 36329453-36329455 on Build GRCh38
rs76577621 Chr.13: 36329712-36329712 on Build GRCh38
rs879928327 Chr.13: 36329678-36329678 on Build GRCh38
rs754017704 Chr.13: 36329333-36329334 on Build GRCh38
rs777655586 Chr.13: 36329372-36329372 on Build GRCh38
rs140222511 Chr.13: 36329371-36329371 on Build GRCh38
rs938000999 Chr.13: 36329649-36329649 on Build GRCh38
rs764862087 Chr.13: 36329446-36329446 on Build GRCh38
rs750913212 Chr.13: 36329326-36329326 on Build GRCh38
rs777598423 Chr.13: 36329489-36329489 on Build GRCh38
rs570717299 Chr.13: 36329601-36329601 on Build GRCh38
rs1036826863 Chr.13: 36329742-36329742 on Build GRCh38
rs756548480 Chr.13: 36329330-36329330 on Build GRCh38
rs200205969 Chr.13: 36329447-36329447 on Build GRCh38
rs758054950 Chr.13: 36329451-36329451 on Build GRCh38
rs780573003 Chr.13: 36329335-36329335 on Build GRCh38
rs781240294 Chr.13: 36329523-36329523 on Build GRCh38
rs995424795 Chr.13: 36329352-36329352 on Build GRCh38
rs142213331 Chr.13: 36329579-36329579 on Build GRCh38
rs754409265 Chr.13: 36329435-36329435 on Build GRCh38
rs1050353760 Chr.13: 36329734-36329734 on Build GRCh38
rs770949994 Chr.13: 36329395-36329395 on Build GRCh38
rs992007347 Chr.13: 36329654-36329654 on Build GRCh38
rs60018272 Chr.13: 36329715-36329715 on Build GRCh38
rs537481996 Chr.13: 36329425-36329425 on Build GRCh38
rs762048880 Chr.13: 36329314-36329314 on Build GRCh38
rs369101005 Chr.13: 36329433-36329433 on Build GRCh38

View Full Product Details