Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48045911-48046414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT15
Entrez Gene ID
55270
Gene Name
nudix hydrolase 15
Gene Aliases
MTH2, NUDT15D
UniGene
Hs.144407
Gene Chromosome Location
Chr.13: 48037567-48052742 on Build GRCh38
Transcripts
NM_018283.3 NR_136687.1 NR_136688.1

SNP Detail

SNP ID Chromosome Location
rs369717493 Chr.13: 48046257-48046257 on Build GRCh38
rs530757995 Chr.13: 48046261-48046265 on Build GRCh38
rs933757262 Chr.13: 48046297-48046297 on Build GRCh38
rs1042913569 Chr.13: 48046100-48046100 on Build GRCh38
rs1015099142 Chr.13: 48046181-48046184 on Build GRCh38
rs1035545900 Chr.13: 48046161-48046161 on Build GRCh38
rs985654509 Chr.13: 48046287-48046287 on Build GRCh38
rs539516008 Chr.13: 48046316-48046316 on Build GRCh38
rs371242733 Chr.13: 48045948-48045948 on Build GRCh38
rs964904544 Chr.13: 48046237-48046237 on Build GRCh38
rs564507484 Chr.13: 48046132-48046132 on Build GRCh38
rs947180078 Chr.13: 48046089-48046089 on Build GRCh38
rs561637446 Chr.13: 48046034-48046034 on Build GRCh38
rs908212734 Chr.13: 48046347-48046347 on Build GRCh38
rs953813582 Chr.13: 48046281-48046281 on Build GRCh38
rs369112228 Chr.13: 48046103-48046103 on Build GRCh38
rs1016775428 Chr.13: 48046064-48046064 on Build GRCh38
rs933684356 Chr.13: 48045971-48045971 on Build GRCh38
rs750049506 Chr.13: 48046311-48046311 on Build GRCh38
rs943934716 Chr.13: 48046028-48046028 on Build GRCh38
rs1002058647 Chr.13: 48046135-48046135 on Build GRCh38
rs905107990 Chr.13: 48046380-48046380 on Build GRCh38
rs1035690431 Chr.13: 48046140-48046140 on Build GRCh38
rs770129191 Chr.13: 48046190-48046190 on Build GRCh38
rs979694454 Chr.13: 48046226-48046226 on Build GRCh38
rs1020958472 Chr.13: 48046201-48046201 on Build GRCh38
rs764788766 Chr.13: 48046303-48046303 on Build GRCh38
rs541689297 Chr.13: 48046017-48046017 on Build GRCh38
rs891621694 Chr.13: 48046087-48046088 on Build GRCh38
rs1001976527 Chr.13: 48046148-48046148 on Build GRCh38
rs1046723839 Chr.13: 48045972-48045972 on Build GRCh38
rs760706532 Chr.13: 48046163-48046163 on Build GRCh38
rs911165356 Chr.13: 48045967-48045967 on Build GRCh38
rs17071169 Chr.13: 48046216-48046216 on Build GRCh38
rs976239586 Chr.13: 48046249-48046249 on Build GRCh38
rs922383347 Chr.13: 48046267-48046267 on Build GRCh38
rs146168097 Chr.13: 48046374-48046374 on Build GRCh38
rs886734408 Chr.13: 48046009-48046009 on Build GRCh38
rs941005447 Chr.13: 48046351-48046351 on Build GRCh38
rs764176405 Chr.13: 48046213-48046213 on Build GRCh38
rs373338414 Chr.13: 48046162-48046162 on Build GRCh38
rs905911800 Chr.13: 48046127-48046127 on Build GRCh38
rs41284207 Chr.13: 48046248-48046248 on Build GRCh38
rs374987487 Chr.13: 48046098-48046098 on Build GRCh38
rs373493609 Chr.13: 48046296-48046296 on Build GRCh38
rs761480842 Chr.13: 48046227-48046227 on Build GRCh38
rs913114609 Chr.13: 48046373-48046373 on Build GRCh38
rs149679383 Chr.13: 48045964-48045964 on Build GRCh38
rs543038144 Chr.13: 48046120-48046120 on Build GRCh38

View Full Product Details