Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49459972-49460518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETDB2
Entrez Gene ID
83852
Gene Name
SET domain bifurcated 2
Gene Aliases
C13orf4, CLLD8, CLLL8, KMT1F
UniGene
Hs.631789
Gene Chromosome Location
Chr.13: 49444293-49495003 on Build GRCh38
Transcripts
NM_001160308.2 NM_001320699.1 NM_031915.2 NM_001320727.1 NR_135324.1

Gene Symbol
SETDB2-PHF11
Entrez Gene ID
107303344
Gene Name
SETDB2-PHF11 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.13: 49444293-49528992 on Build GRCh38
Transcripts
NM_001160308.2 NM_001320699.1 NM_031915.2 NM_001320727.1 NR_135324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629134 c.99G>C p.Q33H Chr.13: 49460189-49460189 on Build GRCh38
6850765 c.36G>A p.W12* Chr.13: 49460126-49460126 on Build GRCh38
4047964 c.19G>A p.D7N Chr.13: 49460109-49460109 on Build GRCh38
459274 c.52G>A p.D18N Chr.13: 49460142-49460142 on Build GRCh38
6311068 c.82G>T p.V28L Chr.13: 49460172-49460172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980788936 Chr.13: 49460416-49460416 on Build GRCh38
rs752714627 Chr.13: 49460149-49460149 on Build GRCh38
rs527656927 Chr.13: 49460001-49460001 on Build GRCh38
rs992957674 Chr.13: 49460478-49460478 on Build GRCh38
rs34727224 Chr.13: 49460387-49460388 on Build GRCh38
rs747432122 Chr.13: 49460241-49460241 on Build GRCh38
rs370563681 Chr.13: 49460348-49460348 on Build GRCh38
rs768557347 Chr.13: 49460071-49460071 on Build GRCh38
rs371292180 Chr.13: 49460175-49460175 on Build GRCh38
rs780432666 Chr.13: 49460183-49460183 on Build GRCh38
rs1030382066 Chr.13: 49460325-49460325 on Build GRCh38
rs774916000 Chr.13: 49460264-49460264 on Build GRCh38
rs762753430 Chr.13: 49460161-49460161 on Build GRCh38
rs778248069 Chr.13: 49460227-49460227 on Build GRCh38
rs749433487 Chr.13: 49460063-49460063 on Build GRCh38
rs927474961 Chr.13: 49460134-49460134 on Build GRCh38
rs764198840 Chr.13: 49460284-49460284 on Build GRCh38
rs181791362 Chr.13: 49460449-49460449 on Build GRCh38
rs367767717 Chr.13: 49460174-49460174 on Build GRCh38
rs538466910 Chr.13: 49460208-49460208 on Build GRCh38
rs1009825264 Chr.13: 49460172-49460172 on Build GRCh38
rs55800238 Chr.13: 49460303-49460303 on Build GRCh38
rs760996884 Chr.13: 49460090-49460090 on Build GRCh38
rs979464346 Chr.13: 49460394-49460394 on Build GRCh38
rs144284121 Chr.13: 49460186-49460186 on Build GRCh38
rs938093155 Chr.13: 49460432-49460432 on Build GRCh38
rs150577541 Chr.13: 49460115-49460115 on Build GRCh38
rs923364854 Chr.13: 49460051-49460051 on Build GRCh38
rs567453411 Chr.13: 49460359-49460359 on Build GRCh38
rs754150494 Chr.13: 49460249-49460250 on Build GRCh38
rs777344778 Chr.13: 49460310-49460310 on Build GRCh38
rs1013741060 Chr.13: 49460069-49460069 on Build GRCh38
rs200490663 Chr.13: 49460156-49460156 on Build GRCh38
rs891563228 Chr.13: 49460167-49460167 on Build GRCh38
rs758868733 Chr.13: 49460214-49460214 on Build GRCh38
rs764209530 Chr.13: 49460092-49460097 on Build GRCh38
rs771228451 Chr.13: 49460249-49460249 on Build GRCh38
rs186480776 Chr.13: 49460261-49460261 on Build GRCh38
rs1056777600 Chr.13: 49460154-49460154 on Build GRCh38
rs759100860 Chr.13: 49460118-49460118 on Build GRCh38
rs748235391 Chr.13: 49460212-49460212 on Build GRCh38
rs938824642 Chr.13: 49460455-49460455 on Build GRCh38
rs748561237 Chr.13: 49460077-49460077 on Build GRCh38
rs764586518 Chr.13: 49460123-49460123 on Build GRCh38
rs745472536 Chr.13: 49460258-49460258 on Build GRCh38
rs1007245110 Chr.13: 49460232-49460232 on Build GRCh38
rs558350020 Chr.13: 49460084-49460084 on Build GRCh38
rs925242476 Chr.13: 49460414-49460414 on Build GRCh38
rs909251939 Chr.13: 49460345-49460345 on Build GRCh38
rs535298698 Chr.13: 49460036-49460036 on Build GRCh38
rs138722347 Chr.13: 49460179-49460179 on Build GRCh38
rs755113120 Chr.13: 49460192-49460192 on Build GRCh38
rs190136290 Chr.13: 49460320-49460320 on Build GRCh38
rs776253482 Chr.13: 49460109-49460109 on Build GRCh38
rs542586734 Chr.13: 49460267-49460267 on Build GRCh38
rs748651700 Chr.13: 49460357-49460357 on Build GRCh38
rs950784900 Chr.13: 49460332-49460332 on Build GRCh38
rs983442830 Chr.13: 49460333-49460333 on Build GRCh38
rs770637591 Chr.13: 49460108-49460108 on Build GRCh38
rs917991989 Chr.13: 49460450-49460450 on Build GRCh38
rs148731175 Chr.13: 49460256-49460256 on Build GRCh38
rs201168386 Chr.13: 49460087-49460087 on Build GRCh38
rs938429225 Chr.13: 49460138-49460138 on Build GRCh38
rs963643489 Chr.13: 49460380-49460380 on Build GRCh38

View Full Product Details