Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 60509571-60510075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD3
Entrez Gene ID
81550
Gene Name
tudor domain containing 3
Gene Aliases
-
UniGene
Hs.525061
Gene Chromosome Location
Chr.13: 60396457-60573879 on Build GRCh38
Transcripts
NM_001146070.1 NM_001146071.1 NM_030794.2 XM_005266556.4 XM_005266560.2 XM_011535247.1 XM_011535249.2 XM_017020777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448681 c.694A>G p.T232A Chr.13: 60509877-60509877 on Build GRCh38
5827406 c.886A>C p.T296P Chr.13: 60509790-60509790 on Build GRCh38
1646609 c.889G>T p.E297* Chr.13: 60509793-60509793 on Build GRCh38
6042755 c.957A>G p.A319A Chr.13: 60509861-60509861 on Build GRCh38
1367543 c.607A>C p.T203P Chr.13: 60509790-60509790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117941795 Chr.13: 60509753-60509753 on Build GRCh38
rs573953540 Chr.13: 60509619-60509619 on Build GRCh38
rs764914707 Chr.13: 60509961-60509961 on Build GRCh38
rs372916852 Chr.13: 60509881-60509881 on Build GRCh38
rs996734815 Chr.13: 60509691-60509691 on Build GRCh38
rs752281409 Chr.13: 60509906-60509908 on Build GRCh38
rs774819441 Chr.13: 60509923-60509923 on Build GRCh38
rs199860029 Chr.13: 60509888-60509888 on Build GRCh38
rs749044387 Chr.13: 60509763-60509763 on Build GRCh38
rs754835385 Chr.13: 60509780-60509780 on Build GRCh38
rs771615809 Chr.13: 60509668-60509668 on Build GRCh38
rs902981525 Chr.13: 60509969-60509969 on Build GRCh38
rs1006218882 Chr.13: 60509607-60509607 on Build GRCh38
rs754960025 Chr.13: 60509759-60509759 on Build GRCh38
rs762431925 Chr.13: 60509858-60509858 on Build GRCh38
rs368762550 Chr.13: 60509970-60509970 on Build GRCh38
rs368325280 Chr.13: 60509822-60509822 on Build GRCh38
rs774703612 Chr.13: 60509856-60509856 on Build GRCh38
rs1040715120 Chr.13: 60510032-60510032 on Build GRCh38
rs369171180 Chr.13: 60509947-60509947 on Build GRCh38
rs763294981 Chr.13: 60509941-60509941 on Build GRCh38
rs758403754 Chr.13: 60509891-60509891 on Build GRCh38
rs749206104 Chr.13: 60509661-60509661 on Build GRCh38
rs761171662 Chr.13: 60509665-60509665 on Build GRCh38
rs758407063 Chr.13: 60509791-60509791 on Build GRCh38
rs991503068 Chr.13: 60509836-60509836 on Build GRCh38
rs763328062 Chr.13: 60509711-60509711 on Build GRCh38
rs756397959 Chr.13: 60509735-60509735 on Build GRCh38
rs954729439 Chr.13: 60509747-60509747 on Build GRCh38
rs1030145646 Chr.13: 60509667-60509667 on Build GRCh38
rs755304408 Chr.13: 60509954-60509954 on Build GRCh38
rs149669879 Chr.13: 60509819-60509819 on Build GRCh38
rs551906273 Chr.13: 60509690-60509690 on Build GRCh38
rs764505323 Chr.13: 60509833-60509833 on Build GRCh38
rs754118001 Chr.13: 60509880-60509880 on Build GRCh38
rs772831358 Chr.13: 60509800-60509800 on Build GRCh38
rs374369946 Chr.13: 60509740-60509740 on Build GRCh38
rs938264604 Chr.13: 60509832-60509832 on Build GRCh38
rs772740201 Chr.13: 60509671-60509671 on Build GRCh38
rs918079029 Chr.13: 60509868-60509868 on Build GRCh38
rs768985609 Chr.13: 60509917-60509917 on Build GRCh38
rs780943995 Chr.13: 60509837-60509838 on Build GRCh38
rs116156909 Chr.13: 60510019-60510019 on Build GRCh38
rs766651856 Chr.13: 60509878-60509878 on Build GRCh38
rs1016100676 Chr.13: 60509621-60509621 on Build GRCh38
rs767463347 Chr.13: 60509861-60509861 on Build GRCh38
rs559410453 Chr.13: 60509657-60509657 on Build GRCh38
rs780043444 Chr.13: 60509660-60509660 on Build GRCh38
rs145444654 Chr.13: 60509896-60509896 on Build GRCh38
rs541198195 Chr.13: 60509649-60509649 on Build GRCh38
rs746393972 Chr.13: 60509818-60509818 on Build GRCh38
rs760437488 Chr.13: 60509674-60509674 on Build GRCh38
rs771474082 Chr.13: 60509792-60509792 on Build GRCh38
rs535416907 Chr.13: 60510000-60510000 on Build GRCh38
rs925499145 Chr.13: 60509831-60509831 on Build GRCh38
rs754099788 Chr.13: 60509953-60509953 on Build GRCh38
rs753103542 Chr.13: 60509707-60509707 on Build GRCh38
rs756283079 Chr.13: 60509867-60509867 on Build GRCh38
rs148850765 Chr.13: 60509902-60509902 on Build GRCh38
rs746467246 Chr.13: 60509897-60509897 on Build GRCh38
rs762114088 Chr.13: 60509927-60509927 on Build GRCh38
rs776257563 Chr.13: 60509903-60509903 on Build GRCh38
rs201326973 Chr.13: 60509884-60509884 on Build GRCh38
rs549770869 Chr.13: 60509859-60509859 on Build GRCh38
rs757275129 Chr.13: 60509718-60509718 on Build GRCh38
rs375852710 Chr.13: 60509907-60509907 on Build GRCh38
rs764523642 Chr.13: 60509713-60509713 on Build GRCh38
rs527529111 Chr.13: 60509662-60509662 on Build GRCh38
rs768019005 Chr.13: 60509929-60509929 on Build GRCh38
rs760771471 Chr.13: 60509946-60509946 on Build GRCh38
rs750474692 Chr.13: 60509864-60509864 on Build GRCh38
rs778928155 Chr.13: 60509787-60509787 on Build GRCh38
rs781391384 Chr.13: 60509732-60509732 on Build GRCh38
rs900439643 Chr.13: 60509627-60509627 on Build GRCh38
rs745575030 Chr.13: 60509904-60509904 on Build GRCh38

View Full Product Details