Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72710226-72710745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MZT1
Entrez Gene ID
440145
Gene Name
mitotic spindle organizing protein 1
Gene Aliases
C13orf37, MOZART1
UniGene
Hs.28465
Gene Chromosome Location
Chr.13: 72708357-72727800 on Build GRCh38
Transcripts
NM_001071775.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7082841 c.232G>A p.E78K Chr.13: 72710339-72710339 on Build GRCh38
5121031 c.226-9T>A p.? Chr.13: 72710354-72710354 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs147090065 Chr.13: 72710546-72710546 on Build GRCh38
rs1001062856 Chr.13: 72710702-72710702 on Build GRCh38
rs566063843 Chr.13: 72710458-72710458 on Build GRCh38
rs376205342 Chr.13: 72710393-72710393 on Build GRCh38
rs936182207 Chr.13: 72710453-72710453 on Build GRCh38
rs181878410 Chr.13: 72710689-72710689 on Build GRCh38
rs780543891 Chr.13: 72710350-72710350 on Build GRCh38
rs536520029 Chr.13: 72710513-72710513 on Build GRCh38
rs561198916 Chr.13: 72710704-72710704 on Build GRCh38
rs765560555 Chr.13: 72710316-72710316 on Build GRCh38
rs7931 Chr.13: 72710273-72710273 on Build GRCh38
rs759924562 Chr.13: 72710304-72710304 on Build GRCh38
rs912855763 Chr.13: 72710708-72710708 on Build GRCh38
rs188769722 Chr.13: 72710554-72710554 on Build GRCh38
rs1001343427 Chr.13: 72710464-72710464 on Build GRCh38
rs761415885 Chr.13: 72710289-72710289 on Build GRCh38
rs772763025 Chr.13: 72710303-72710303 on Build GRCh38
rs758862559 Chr.13: 72710295-72710296 on Build GRCh38
rs572043076 Chr.13: 72710691-72710691 on Build GRCh38
rs768102320 Chr.13: 72710366-72710366 on Build GRCh38
rs771814228 Chr.13: 72710302-72710302 on Build GRCh38
rs764588981 Chr.13: 72710332-72710332 on Build GRCh38
rs887642262 Chr.13: 72710624-72710624 on Build GRCh38
rs751596847 Chr.13: 72710351-72710351 on Build GRCh38
rs537610408 Chr.13: 72710567-72710567 on Build GRCh38
rs1021069298 Chr.13: 72710690-72710690 on Build GRCh38
rs1002027137 Chr.13: 72710586-72710586 on Build GRCh38
rs781450978 Chr.13: 72710362-72710362 on Build GRCh38
rs1040413944 Chr.13: 72710562-72710562 on Build GRCh38
rs892436841 Chr.13: 72710526-72710526 on Build GRCh38
rs749112768 Chr.13: 72710377-72710377 on Build GRCh38
rs753138677 Chr.13: 72710368-72710368 on Build GRCh38
rs929408688 Chr.13: 72710585-72710585 on Build GRCh38
rs1052469354 Chr.13: 72710480-72710480 on Build GRCh38
rs147994764 Chr.13: 72710645-72710645 on Build GRCh38
rs556295966 Chr.13: 72710612-72710612 on Build GRCh38
rs773943956 Chr.13: 72710282-72710282 on Build GRCh38
rs896427476 Chr.13: 72710662-72710662 on Build GRCh38
rs116353270 Chr.13: 72710699-72710699 on Build GRCh38
rs757349597 Chr.13: 72710359-72710359 on Build GRCh38
rs141712224 Chr.13: 72710417-72710417 on Build GRCh38
rs556722298 Chr.13: 72710544-72710544 on Build GRCh38
rs577808533 Chr.13: 72710642-72710642 on Build GRCh38
rs768264117 Chr.13: 72710280-72710280 on Build GRCh38
rs942804642 Chr.13: 72710349-72710349 on Build GRCh38
rs1002002509 Chr.13: 72710648-72710648 on Build GRCh38
rs779768450 Chr.13: 72710370-72710370 on Build GRCh38
rs750037102 Chr.13: 72710660-72710660 on Build GRCh38
rs963185586 Chr.13: 72710629-72710629 on Build GRCh38
rs139369257 Chr.13: 72710276-72710276 on Build GRCh38
rs966874935 Chr.13: 72710385-72710385 on Build GRCh38
rs753244239 Chr.13: 72710317-72710317 on Build GRCh38
rs903972951 Chr.13: 72710427-72710427 on Build GRCh38
rs756491736 Chr.13: 72710484-72710484 on Build GRCh38
rs747105280 Chr.13: 72710373-72710373 on Build GRCh38

View Full Product Details