Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99307214-99307733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR183
Entrez Gene ID
1880
Gene Name
G protein-coupled receptor 183
Gene Aliases
EBI2
UniGene
Hs.784
Gene Chromosome Location
Chr.13: 99294273-99307495 on Build GRCh38
Transcripts
NM_004951.4 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
UniGene
Hs.508545
Gene Chromosome Location
Chr.13: 99200425-99386499 on Build GRCh38
Transcripts
NM_004951.4 NM_001144072.1 NM_177967.3 NR_026644.1 XM_006719948.3 XM_011521082.2 XM_011521083.2 XM_011521084.2

SNP Detail

SNP ID Chromosome Location
rs559939276 Chr.13: 99307499-99307499 on Build GRCh38
rs886873801 Chr.13: 99307320-99307320 on Build GRCh38
rs140061829 Chr.13: 99307541-99307541 on Build GRCh38
rs200539103 Chr.13: 99307536-99307537 on Build GRCh38
rs904397063 Chr.13: 99307471-99307471 on Build GRCh38
rs977436829 Chr.13: 99307601-99307601 on Build GRCh38
rs944762909 Chr.13: 99307367-99307367 on Build GRCh38
rs191373302 Chr.13: 99307392-99307392 on Build GRCh38
rs767765176 Chr.13: 99307240-99307243 on Build GRCh38
rs975293938 Chr.13: 99307660-99307660 on Build GRCh38
rs1041599014 Chr.13: 99307444-99307444 on Build GRCh38
rs1034103366 Chr.13: 99307483-99307483 on Build GRCh38
rs561044126 Chr.13: 99307605-99307605 on Build GRCh38
rs957360066 Chr.13: 99307627-99307627 on Build GRCh38
rs961709954 Chr.13: 99307521-99307521 on Build GRCh38
rs547172139 Chr.13: 99307696-99307696 on Build GRCh38
rs548316750 Chr.13: 99307514-99307514 on Build GRCh38
rs369443935 Chr.13: 99307359-99307359 on Build GRCh38
rs1024370083 Chr.13: 99307523-99307523 on Build GRCh38
rs34324109 Chr.13: 99307491-99307492 on Build GRCh38
rs188403352 Chr.13: 99307356-99307356 on Build GRCh38
rs778470064 Chr.13: 99307524-99307524 on Build GRCh38
rs933787041 Chr.13: 99307689-99307689 on Build GRCh38
rs894219783 Chr.13: 99307501-99307501 on Build GRCh38
rs1034569814 Chr.13: 99307401-99307401 on Build GRCh38
rs34923467 Chr.13: 99307517-99307517 on Build GRCh38
rs995091636 Chr.13: 99307384-99307384 on Build GRCh38
rs899419078 Chr.13: 99307377-99307377 on Build GRCh38
rs892438874 Chr.13: 99307398-99307398 on Build GRCh38
rs35060502 Chr.13: 99307462-99307463 on Build GRCh38
rs185095069 Chr.13: 99307617-99307617 on Build GRCh38
rs534953541 Chr.13: 99307697-99307697 on Build GRCh38
rs921195179 Chr.13: 99307684-99307684 on Build GRCh38
rs565972791 Chr.13: 99307279-99307279 on Build GRCh38
rs751222136 Chr.13: 99307397-99307397 on Build GRCh38
rs910680642 Chr.13: 99307696-99307698 on Build GRCh38
rs910617885 Chr.13: 99307357-99307357 on Build GRCh38
rs972330217 Chr.13: 99307537-99307537 on Build GRCh38
rs999979436 Chr.13: 99307471-99307472 on Build GRCh38
rs1038471797 Chr.13: 99307376-99307376 on Build GRCh38
rs1923891 Chr.13: 99307691-99307691 on Build GRCh38

View Full Product Details