Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28767812-28768314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXG1
Entrez Gene ID
2290
Gene Name
forkhead box G1
Gene Aliases
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
UniGene
Hs.632336
Gene Chromosome Location
Chr.14: 28767072-28770277 on Build GRCh38
Transcripts
NM_005249.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2033899 c.802G>A p.G268S Chr.14: 28768081-28768081 on Build GRCh38
5065409 c.1003C>G p.P335A Chr.14: 28768282-28768282 on Build GRCh38
6331238 c.943T>C p.S315P Chr.14: 28768222-28768222 on Build GRCh38
2033894 c.696T>C p.N232N Chr.14: 28767975-28767975 on Build GRCh38
4050377 c.947T>C p.L316P Chr.14: 28768226-28768226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759790790 Chr.14: 28768029-28768029 on Build GRCh38
rs570340475 Chr.14: 28768116-28768116 on Build GRCh38
rs786205011 Chr.14: 28768248-28768248 on Build GRCh38
rs781085552 Chr.14: 28768242-28768242 on Build GRCh38
rs768525699 Chr.14: 28767936-28767936 on Build GRCh38
rs786205005 Chr.14: 28767856-28767856 on Build GRCh38
rs796052468 Chr.14: 28767987-28767987 on Build GRCh38
rs537686463 Chr.14: 28768131-28768131 on Build GRCh38
rs587783639 Chr.14: 28767951-28767951 on Build GRCh38
rs121913678 Chr.14: 28768044-28768044 on Build GRCh38
rs267606828 Chr.14: 28767922-28767922 on Build GRCh38
rs796052481 Chr.14: 28767950-28767950 on Build GRCh38
rs869312961 Chr.14: 28768055-28768055 on Build GRCh38
rs930327703 Chr.14: 28767897-28767897 on Build GRCh38
rs866923607 Chr.14: 28768179-28768179 on Build GRCh38
rs922136584 Chr.14: 28768205-28768205 on Build GRCh38
rs745418016 Chr.14: 28768137-28768137 on Build GRCh38
rs756719183 Chr.14: 28767861-28767861 on Build GRCh38
rs142067350 Chr.14: 28768164-28768164 on Build GRCh38
rs786205009 Chr.14: 28768009-28768009 on Build GRCh38
rs796052484 Chr.14: 28768010-28768010 on Build GRCh38
rs1057518165 Chr.14: 28767924-28767924 on Build GRCh38
rs796052483 Chr.14: 28768000-28768000 on Build GRCh38
rs587783640 Chr.14: 28768034-28768034 on Build GRCh38
rs771755778 Chr.14: 28767900-28767900 on Build GRCh38
rs757832735 Chr.14: 28768122-28768122 on Build GRCh38
rs796052466 Chr.14: 28767954-28767954 on Build GRCh38
rs746727507 Chr.14: 28768194-28768194 on Build GRCh38
rs879255530 Chr.14: 28767832-28767832 on Build GRCh38
rs786205008 Chr.14: 28767979-28767979 on Build GRCh38
rs796052462 Chr.14: 28767840-28767840 on Build GRCh38
rs796052465 Chr.14: 28767946-28767946 on Build GRCh38
rs199502880 Chr.14: 28767965-28767965 on Build GRCh38
rs786205486 Chr.14: 28767973-28767973 on Build GRCh38
rs587783641 Chr.14: 28768036-28768036 on Build GRCh38
rs796052463 Chr.14: 28767865-28767865 on Build GRCh38
rs766343459 Chr.14: 28767969-28767969 on Build GRCh38
rs774525510 Chr.14: 28768008-28768008 on Build GRCh38
rs778154529 Chr.14: 28767834-28767834 on Build GRCh38
rs905973556 Chr.14: 28767849-28767849 on Build GRCh38
rs753051078 Chr.14: 28768077-28768077 on Build GRCh38
rs774246700 Chr.14: 28768263-28768263 on Build GRCh38
rs796052469 Chr.14: 28768091-28768091 on Build GRCh38
rs796052470 Chr.14: 28768123-28768123 on Build GRCh38
rs968922416 Chr.14: 28768156-28768156 on Build GRCh38
rs141088742 Chr.14: 28767873-28767873 on Build GRCh38
rs587783643 Chr.14: 28768078-28768078 on Build GRCh38
rs748766934 Chr.14: 28767846-28767846 on Build GRCh38
rs869312700 Chr.14: 28768100-28768100 on Build GRCh38
rs727503935 Chr.14: 28767949-28767949 on Build GRCh38
rs368707795 Chr.14: 28768284-28768284 on Build GRCh38
rs772352099 Chr.14: 28768272-28768272 on Build GRCh38
rs1057516138 Chr.14: 28767833-28767833 on Build GRCh38
rs1001642335 Chr.14: 28767863-28767863 on Build GRCh38
rs796052467 Chr.14: 28767980-28767980 on Build GRCh38
rs796052482 Chr.14: 28767952-28767952 on Build GRCh38
rs748001255 Chr.14: 28768256-28768256 on Build GRCh38
rs150277632 Chr.14: 28768268-28768268 on Build GRCh38
rs267606826 Chr.14: 28767903-28767903 on Build GRCh38
rs768320307 Chr.14: 28768235-28768235 on Build GRCh38
rs201191801 Chr.14: 28767885-28767885 on Build GRCh38
rs786205010 Chr.14: 28768067-28768071 on Build GRCh38
rs979367991 Chr.14: 28768182-28768182 on Build GRCh38
rs786205006 Chr.14: 28767889-28767889 on Build GRCh38
rs786205012 Chr.14: 28767960-28767960 on Build GRCh38
rs988740176 Chr.14: 28768129-28768129 on Build GRCh38
rs374673901 Chr.14: 28768089-28768089 on Build GRCh38
rs796934848 Chr.14: 28768062-28768062 on Build GRCh38
rs865783570 Chr.14: 28768118-28768118 on Build GRCh38
rs786204998 Chr.14: 28767923-28767924 on Build GRCh38
rs761037576 Chr.14: 28768278-28768278 on Build GRCh38
rs786205007 Chr.14: 28767968-28767968 on Build GRCh38
rs767873754 Chr.14: 28768038-28768038 on Build GRCh38
rs796052471 Chr.14: 28768159-28768159 on Build GRCh38
rs727503934 Chr.14: 28767927-28767934 on Build GRCh38
rs531378284 Chr.14: 28768282-28768282 on Build GRCh38
rs775851487 Chr.14: 28768273-28768273 on Build GRCh38
rs761866084 Chr.14: 28767963-28767963 on Build GRCh38
rs764587853 Chr.14: 28768113-28768113 on Build GRCh38
rs587783642 Chr.14: 28768041-28768041 on Build GRCh38
rs267606827 Chr.14: 28768203-28768203 on Build GRCh38
rs587783638 Chr.14: 28767842-28767842 on Build GRCh38
rs796052464 Chr.14: 28767930-28767930 on Build GRCh38
rs886041744 Chr.14: 28768076-28768076 on Build GRCh38
rs960034268 Chr.14: 28768068-28768068 on Build GRCh38
rs998702635 Chr.14: 28768195-28768195 on Build GRCh38
rs745486814 Chr.14: 28767880-28767880 on Build GRCh38
rs759699805 Chr.14: 28768270-28768270 on Build GRCh38
rs548935590 Chr.14: 28767925-28767925 on Build GRCh38
rs779697567 Chr.14: 28768186-28768186 on Build GRCh38

View Full Product Details