Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36676496-36677000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX9
Entrez Gene ID
5083
Gene Name
paired box 9
Gene Aliases
STHAG3
UniGene
Hs.132576
Gene Chromosome Location
Chr.14: 36657568-36677807 on Build GRCh38
Transcripts
NM_006194.3

SNP Detail

SNP ID Chromosome Location
rs532783450 Chr.14: 36676847-36676847 on Build GRCh38
rs886050493 Chr.14: 36676780-36676780 on Build GRCh38
rs765038650 Chr.14: 36676813-36676813 on Build GRCh38
rs939779785 Chr.14: 36676537-36676537 on Build GRCh38
rs958141057 Chr.14: 36676610-36676610 on Build GRCh38
rs1000397475 Chr.14: 36676659-36676659 on Build GRCh38
rs17104962 Chr.14: 36676658-36676658 on Build GRCh38
rs770675967 Chr.14: 36676647-36676647 on Build GRCh38
rs536017500 Chr.14: 36676518-36676518 on Build GRCh38
rs17104965 Chr.14: 36676711-36676711 on Build GRCh38
rs998102585 Chr.14: 36676617-36676617 on Build GRCh38
rs796334795 Chr.14: 36676601-36676601 on Build GRCh38
rs773998473 Chr.14: 36676702-36676702 on Build GRCh38
rs1036654489 Chr.14: 36676565-36676565 on Build GRCh38
rs555952749 Chr.14: 36676520-36676520 on Build GRCh38
rs1036773148 Chr.14: 36676855-36676855 on Build GRCh38
rs530578050 Chr.14: 36676805-36676805 on Build GRCh38
rs538324536 Chr.14: 36676527-36676527 on Build GRCh38
rs1028631915 Chr.14: 36676630-36676630 on Build GRCh38
rs1049408933 Chr.14: 36676882-36676882 on Build GRCh38
rs748540311 Chr.14: 36676796-36676797 on Build GRCh38
rs1013448114 Chr.14: 36676842-36676842 on Build GRCh38
rs889668597 Chr.14: 36676889-36676889 on Build GRCh38
rs980790916 Chr.14: 36676556-36676556 on Build GRCh38
rs908091277 Chr.14: 36676841-36676841 on Build GRCh38
rs571697845 Chr.14: 36676573-36676573 on Build GRCh38
rs879716873 Chr.14: 36676724-36676728 on Build GRCh38
rs559930165 Chr.14: 36676902-36676902 on Build GRCh38
rs558264939 Chr.14: 36676555-36676555 on Build GRCh38
rs888155128 Chr.14: 36676536-36676536 on Build GRCh38
rs908890974 Chr.14: 36676745-36676745 on Build GRCh38
rs977397238 Chr.14: 36676697-36676697 on Build GRCh38
rs564902020 Chr.14: 36676967-36676970 on Build GRCh38
rs541916401 Chr.14: 36676683-36676683 on Build GRCh38
rs900936536 Chr.14: 36676866-36676866 on Build GRCh38
rs1050087400 Chr.14: 36676743-36676743 on Build GRCh38
rs866681495 Chr.14: 36676860-36676860 on Build GRCh38
rs186877662 Chr.14: 36676648-36676648 on Build GRCh38
rs760143460 Chr.14: 36676677-36676677 on Build GRCh38
rs563894653 Chr.14: 36676715-36676715 on Build GRCh38
rs767260010 Chr.14: 36676740-36676740 on Build GRCh38
rs17766200 Chr.14: 36676881-36676881 on Build GRCh38
rs113372817 Chr.14: 36676971-36676971 on Build GRCh38
rs567411185 Chr.14: 36676963-36676963 on Build GRCh38
rs1021069794 Chr.14: 36676966-36676966 on Build GRCh38
rs112616603 Chr.14: 36676712-36676712 on Build GRCh38
rs930911666 Chr.14: 36676800-36676800 on Build GRCh38
rs528475626 Chr.14: 36676926-36676926 on Build GRCh38
rs191674536 Chr.14: 36676958-36676958 on Build GRCh38
rs886050494 Chr.14: 36676970-36676973 on Build GRCh38
rs750143933 Chr.14: 36676769-36676769 on Build GRCh38
rs575701715 Chr.14: 36676522-36676522 on Build GRCh38

View Full Product Details