Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50587725-50588238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
UniGene
Hs.584905
Gene Chromosome Location
Chr.14: 50533082-50633068 on Build GRCh38
Transcripts
NM_001127713.1 NM_015915.4 NM_181598.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369998 c.181G>A p.A61T Chr.14: 50587977-50587977 on Build GRCh38
3753981 c.84A>G p.P28P Chr.14: 50587880-50587880 on Build GRCh38
328934 c.95C>T p.A32V Chr.14: 50587891-50587891 on Build GRCh38
3419842 c.282+2T>C p.? Chr.14: 50588080-50588080 on Build GRCh38
459160 c.120C>G p.V40V Chr.14: 50587916-50587916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955468184 Chr.14: 50587875-50587875 on Build GRCh38
rs757670224 Chr.14: 50588099-50588099 on Build GRCh38
rs35014209 Chr.14: 50587880-50587880 on Build GRCh38
rs771689808 Chr.14: 50587979-50587979 on Build GRCh38
rs928533445 Chr.14: 50588180-50588180 on Build GRCh38
rs773339310 Chr.14: 50587864-50587864 on Build GRCh38
rs955736784 Chr.14: 50587971-50587971 on Build GRCh38
rs778072986 Chr.14: 50588186-50588186 on Build GRCh38
rs564832738 Chr.14: 50587960-50587960 on Build GRCh38
rs772206990 Chr.14: 50587840-50587840 on Build GRCh38
rs912921540 Chr.14: 50588145-50588145 on Build GRCh38
rs760438659 Chr.14: 50587850-50587850 on Build GRCh38
rs77224287 Chr.14: 50587757-50587757 on Build GRCh38
rs756464141 Chr.14: 50587911-50587911 on Build GRCh38
rs768342546 Chr.14: 50587996-50587996 on Build GRCh38
rs892309522 Chr.14: 50587771-50587771 on Build GRCh38
rs1000830932 Chr.14: 50587763-50587763 on Build GRCh38
rs753197391 Chr.14: 50587894-50587894 on Build GRCh38
rs149901427 Chr.14: 50587959-50587959 on Build GRCh38
rs750050049 Chr.14: 50588107-50588107 on Build GRCh38
rs1018074376 Chr.14: 50588085-50588085 on Build GRCh38
rs775540772 Chr.14: 50587841-50587841 on Build GRCh38
rs945703082 Chr.14: 50588147-50588147 on Build GRCh38
rs1031214248 Chr.14: 50587770-50587770 on Build GRCh38
rs766461592 Chr.14: 50587871-50587871 on Build GRCh38
rs936552512 Chr.14: 50588140-50588140 on Build GRCh38
rs745354380 Chr.14: 50587831-50587831 on Build GRCh38
rs758015932 Chr.14: 50588116-50588116 on Build GRCh38
rs115722924 Chr.14: 50587817-50587817 on Build GRCh38
rs764492146 Chr.14: 50588093-50588093 on Build GRCh38
rs759633651 Chr.14: 50588057-50588057 on Build GRCh38
rs1055547045 Chr.14: 50588212-50588212 on Build GRCh38
rs778130435 Chr.14: 50587933-50587933 on Build GRCh38
rs763902321 Chr.14: 50587854-50587854 on Build GRCh38
rs753147363 Chr.14: 50588069-50588069 on Build GRCh38
rs776429880 Chr.14: 50588000-50588000 on Build GRCh38
rs895234363 Chr.14: 50587774-50587774 on Build GRCh38
rs17850684 Chr.14: 50587925-50587925 on Build GRCh38
rs766627863 Chr.14: 50588010-50588010 on Build GRCh38
rs985606716 Chr.14: 50588067-50588067 on Build GRCh38
rs746934529 Chr.14: 50588128-50588128 on Build GRCh38
rs200314808 Chr.14: 50587992-50587992 on Build GRCh38
rs778699311 Chr.14: 50587963-50587963 on Build GRCh38
rs201580688 Chr.14: 50587973-50587973 on Build GRCh38
rs182963739 Chr.14: 50588019-50588019 on Build GRCh38
rs974359414 Chr.14: 50587988-50587988 on Build GRCh38
rs761583539 Chr.14: 50588004-50588004 on Build GRCh38
rs981774508 Chr.14: 50588176-50588176 on Build GRCh38
rs777311737 Chr.14: 50587783-50587783 on Build GRCh38
rs192302048 Chr.14: 50587810-50587810 on Build GRCh38
rs763169900 Chr.14: 50587866-50587866 on Build GRCh38
rs951568761 Chr.14: 50588014-50588014 on Build GRCh38
rs200452381 Chr.14: 50587886-50587886 on Build GRCh38
rs937213675 Chr.14: 50588209-50588209 on Build GRCh38
rs779551213 Chr.14: 50588125-50588125 on Build GRCh38
rs755879849 Chr.14: 50587945-50587951 on Build GRCh38
rs142314314 Chr.14: 50587883-50587883 on Build GRCh38
rs755019483 Chr.14: 50587881-50587881 on Build GRCh38
rs746927118 Chr.14: 50587995-50587995 on Build GRCh38
rs771475691 Chr.14: 50587921-50587926 on Build GRCh38
rs778760639 Chr.14: 50588101-50588101 on Build GRCh38
rs767543400 Chr.14: 50588064-50588064 on Build GRCh38
rs754290939 Chr.14: 50588096-50588096 on Build GRCh38
rs761183484 Chr.14: 50588074-50588074 on Build GRCh38
rs748928335 Chr.14: 50587800-50587800 on Build GRCh38
rs112194265 Chr.14: 50588130-50588130 on Build GRCh38

View Full Product Details