Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50634278-50634795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAV1
Entrez Gene ID
60485
Gene Name
salvador family WW domain containing protein 1
Gene Aliases
SAV, WW45, WWP4
UniGene
Hs.642842
Gene Chromosome Location
Chr.14: 50633106-50668353 on Build GRCh38
Transcripts
NM_021818.3 XM_011537057.2 XM_017021568.1

SNP Detail

SNP ID Chromosome Location
rs1028461540 Chr.14: 50634352-50634352 on Build GRCh38
rs889498765 Chr.14: 50634733-50634733 on Build GRCh38
rs796070125 Chr.14: 50634402-50634402 on Build GRCh38
rs989936958 Chr.14: 50634353-50634355 on Build GRCh38
rs915944367 Chr.14: 50634559-50634559 on Build GRCh38
rs146185414 Chr.14: 50634643-50634643 on Build GRCh38
rs551503920 Chr.14: 50634714-50634714 on Build GRCh38
rs572094151 Chr.14: 50634383-50634383 on Build GRCh38
rs762909407 Chr.14: 50634571-50634571 on Build GRCh38
rs369850398 Chr.14: 50634745-50634745 on Build GRCh38
rs771909105 Chr.14: 50634371-50634371 on Build GRCh38
rs909094017 Chr.14: 50634619-50634619 on Build GRCh38
rs779394766 Chr.14: 50634339-50634346 on Build GRCh38
rs904569372 Chr.14: 50634582-50634582 on Build GRCh38
rs768577880 Chr.14: 50634473-50634473 on Build GRCh38
rs549678489 Chr.14: 50634588-50634588 on Build GRCh38
rs981139795 Chr.14: 50634547-50634547 on Build GRCh38
rs377283905 Chr.14: 50634467-50634467 on Build GRCh38
rs61985480 Chr.14: 50634540-50634540 on Build GRCh38
rs578114555 Chr.14: 50634443-50634443 on Build GRCh38
rs990688129 Chr.14: 50634548-50634548 on Build GRCh38
rs991781386 Chr.14: 50634381-50634381 on Build GRCh38
rs918708625 Chr.14: 50634648-50634648 on Build GRCh38
rs149710149 Chr.14: 50634535-50634540 on Build GRCh38
rs765780953 Chr.14: 50634618-50634618 on Build GRCh38
rs1012147380 Chr.14: 50634531-50634531 on Build GRCh38
rs551056696 Chr.14: 50634617-50634617 on Build GRCh38
rs1007148123 Chr.14: 50634496-50634496 on Build GRCh38
rs997930993 Chr.14: 50634343-50634343 on Build GRCh38
rs998736805 Chr.14: 50634625-50634625 on Build GRCh38
rs181967395 Chr.14: 50634312-50634312 on Build GRCh38
rs930052488 Chr.14: 50634657-50634657 on Build GRCh38
rs548407231 Chr.14: 50634387-50634387 on Build GRCh38
rs967117760 Chr.14: 50634541-50634541 on Build GRCh38
rs568330630 Chr.14: 50634451-50634451 on Build GRCh38
rs893319457 Chr.14: 50634521-50634521 on Build GRCh38
rs912584080 Chr.14: 50634709-50634709 on Build GRCh38
rs186120699 Chr.14: 50634766-50634766 on Build GRCh38
rs563722708 Chr.14: 50634509-50634509 on Build GRCh38
rs939154009 Chr.14: 50634542-50634542 on Build GRCh38
rs928576484 Chr.14: 50634379-50634379 on Build GRCh38
rs1020426573 Chr.14: 50634357-50634357 on Build GRCh38
rs555368421 Chr.14: 50634322-50634322 on Build GRCh38
rs941834817 Chr.14: 50634412-50634412 on Build GRCh38
rs564578877 Chr.14: 50634440-50634440 on Build GRCh38
rs374756173 Chr.14: 50634536-50634541 on Build GRCh38
rs1051093109 Chr.14: 50634687-50634687 on Build GRCh38
rs937234113 Chr.14: 50634380-50634383 on Build GRCh38
rs773923579 Chr.14: 50634753-50634753 on Build GRCh38
rs1053327436 Chr.14: 50634600-50634600 on Build GRCh38
rs745656636 Chr.14: 50634313-50634313 on Build GRCh38
rs528063720 Chr.14: 50634710-50634710 on Build GRCh38
rs1043033126 Chr.14: 50634576-50634576 on Build GRCh38
rs1039986741 Chr.14: 50634732-50634732 on Build GRCh38
rs141442635 Chr.14: 50634336-50634338 on Build GRCh38
rs541145001 Chr.14: 50634422-50634422 on Build GRCh38
rs981086076 Chr.14: 50634379-50634380 on Build GRCh38
rs967065593 Chr.14: 50634358-50634358 on Build GRCh38
rs948841598 Chr.14: 50634560-50634560 on Build GRCh38
rs529662147 Chr.14: 50634510-50634510 on Build GRCh38

View Full Product Details