Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76176535-76177042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH2L
Entrez Gene ID
55668
Gene Name
G-patch domain containing 2 like
Gene Aliases
C14orf118
UniGene
Hs.410231
Gene Chromosome Location
Chr.14: 76151885-76214473 on Build GRCh38
Transcripts
NM_001322026.1 NM_001322028.1 NM_001322029.1 NM_001322030.1 NM_001322032.1 NM_017926.3 NM_017972.3 NR_110314.1 XM_006720191.2 XM_017021427.1 XM_017021428.1 XM_017021429.1 XM_017021430.1 XM_017021431.1 XM_017021432.1 XM_017021433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2138876 c.1014G>C p.E338D Chr.14: 76176652-76176652 on Build GRCh38
2138877 c.1014G>C p.E338D Chr.14: 76176652-76176652 on Build GRCh38
5848575 c.989C>T p.T330I Chr.14: 76176627-76176627 on Build GRCh38
5848576 c.989C>T p.T330I Chr.14: 76176627-76176627 on Build GRCh38
6141230 c.1040C>T p.P347L Chr.14: 76176678-76176678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143220234 Chr.14: 76176681-76176681 on Build GRCh38
rs553284537 Chr.14: 76177000-76177000 on Build GRCh38
rs985723325 Chr.14: 76176981-76176982 on Build GRCh38
rs1030138557 Chr.14: 76176943-76176943 on Build GRCh38
rs749594146 Chr.14: 76176586-76176586 on Build GRCh38
rs199877754 Chr.14: 76176635-76176635 on Build GRCh38
rs917334450 Chr.14: 76176841-76176841 on Build GRCh38
rs979334752 Chr.14: 76176995-76176995 on Build GRCh38
rs891495745 Chr.14: 76176910-76176910 on Build GRCh38
rs372381529 Chr.14: 76176637-76176637 on Build GRCh38
rs535302212 Chr.14: 76176972-76176973 on Build GRCh38
rs370077733 Chr.14: 76176710-76176710 on Build GRCh38
rs761370374 Chr.14: 76177013-76177013 on Build GRCh38
rs149167474 Chr.14: 76176661-76176661 on Build GRCh38
rs745865345 Chr.14: 76176673-76176673 on Build GRCh38
rs773786359 Chr.14: 76176628-76176628 on Build GRCh38
rs561613959 Chr.14: 76176680-76176680 on Build GRCh38
rs771308795 Chr.14: 76176738-76176738 on Build GRCh38
rs17104191 Chr.14: 76176814-76176814 on Build GRCh38
rs1018100407 Chr.14: 76176983-76176983 on Build GRCh38
rs1029277963 Chr.14: 76177016-76177016 on Build GRCh38
rs761131548 Chr.14: 76176629-76176629 on Build GRCh38
rs751112415 Chr.14: 76176663-76176663 on Build GRCh38
rs761329785 Chr.14: 76176732-76176732 on Build GRCh38
rs569166835 Chr.14: 76176972-76176972 on Build GRCh38
rs775135134 Chr.14: 76176574-76176574 on Build GRCh38
rs895987541 Chr.14: 76176849-76176849 on Build GRCh38
rs7144326 Chr.14: 76176855-76176855 on Build GRCh38
rs748238909 Chr.14: 76176701-76176701 on Build GRCh38
rs945705649 Chr.14: 76176940-76176940 on Build GRCh38
rs1014809314 Chr.14: 76176854-76176854 on Build GRCh38
rs768165874 Chr.14: 76176620-76176620 on Build GRCh38
rs71429939 Chr.14: 76176565-76176565 on Build GRCh38
rs781122818 Chr.14: 76176668-76176668 on Build GRCh38
rs773949942 Chr.14: 76176618-76176618 on Build GRCh38
rs531191014 Chr.14: 76176559-76176559 on Build GRCh38
rs927559063 Chr.14: 76176859-76176859 on Build GRCh38
rs1016926292 Chr.14: 76176878-76176878 on Build GRCh38
rs909927082 Chr.14: 76176703-76176703 on Build GRCh38
rs965874468 Chr.14: 76176585-76176585 on Build GRCh38
rs967994010 Chr.14: 76176983-76176984 on Build GRCh38
rs774543493 Chr.14: 76176594-76176594 on Build GRCh38
rs1040122043 Chr.14: 76176967-76176967 on Build GRCh38
rs10143467 Chr.14: 76176901-76176901 on Build GRCh38
rs765563409 Chr.14: 76176700-76176700 on Build GRCh38
rs961425661 Chr.14: 76176813-76176813 on Build GRCh38
rs368107124 Chr.14: 76176595-76176595 on Build GRCh38
rs201242940 Chr.14: 76176973-76176974 on Build GRCh38
rs552507919 Chr.14: 76176911-76176911 on Build GRCh38
rs1044961627 Chr.14: 76176857-76176857 on Build GRCh38
rs946600364 Chr.14: 76176631-76176631 on Build GRCh38
rs375620948 Chr.14: 76176677-76176677 on Build GRCh38
rs375779596 Chr.14: 76176727-76176727 on Build GRCh38
rs373061705 Chr.14: 76176717-76176717 on Build GRCh38
rs372909607 Chr.14: 76176716-76176716 on Build GRCh38
rs768731265 Chr.14: 76176593-76176593 on Build GRCh38
rs756043705 Chr.14: 76176675-76176675 on Build GRCh38
rs1054104946 Chr.14: 76176908-76176908 on Build GRCh38
rs779941979 Chr.14: 76176676-76176676 on Build GRCh38
rs761932574 Chr.14: 76176616-76176616 on Build GRCh38
rs398057261 Chr.14: 76176982-76176983 on Build GRCh38
rs1018773844 Chr.14: 76176570-76176570 on Build GRCh38

View Full Product Details