Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77329026-77329514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTZ1
Entrez Gene ID
2954
Gene Name
glutathione S-transferase zeta 1
Gene Aliases
GSTZ1-1, MAAI, MAI
UniGene
Hs.655292
Gene Chromosome Location
Chr.14: 77320887-77331597 on Build GRCh38
Transcripts
NM_001312660.1 NM_145870.2 NM_145871.2 XM_011536670.2 XM_011536671.2 XM_017021243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6563274 c.375G>A p.E125E Chr.14: 77329155-77329155 on Build GRCh38
6563275 c.210G>A p.E70E Chr.14: 77329155-77329155 on Build GRCh38
5971262 c.406A>C p.T136P Chr.14: 77329186-77329186 on Build GRCh38
5848584 c.256G>A p.A86T Chr.14: 77329201-77329201 on Build GRCh38
244838 c.222C>G p.T74T Chr.14: 77329167-77329167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8177569 Chr.14: 77329325-77329325 on Build GRCh38
rs546296547 Chr.14: 77329201-77329201 on Build GRCh38
rs2270423 Chr.14: 77329295-77329295 on Build GRCh38
rs907917847 Chr.14: 77329258-77329258 on Build GRCh38
rs116741076 Chr.14: 77329395-77329395 on Build GRCh38
rs769304560 Chr.14: 77329146-77329146 on Build GRCh38
rs774911182 Chr.14: 77329160-77329160 on Build GRCh38
rs535555654 Chr.14: 77329383-77329383 on Build GRCh38
rs1056414501 Chr.14: 77329269-77329269 on Build GRCh38
rs1019467707 Chr.14: 77329308-77329308 on Build GRCh38
rs535308592 Chr.14: 77329076-77329076 on Build GRCh38
rs778518806 Chr.14: 77329093-77329093 on Build GRCh38
rs2234955 Chr.14: 77329177-77329177 on Build GRCh38
rs149972480 Chr.14: 77329180-77329180 on Build GRCh38
rs753964107 Chr.14: 77329200-77329200 on Build GRCh38
rs569460981 Chr.14: 77329413-77329413 on Build GRCh38
rs947890665 Chr.14: 77329305-77329305 on Build GRCh38
rs769220098 Chr.14: 77329077-77329077 on Build GRCh38
rs780273592 Chr.14: 77329268-77329268 on Build GRCh38
rs1037188866 Chr.14: 77329137-77329137 on Build GRCh38
rs958190514 Chr.14: 77329376-77329376 on Build GRCh38
rs769429129 Chr.14: 77329244-77329244 on Build GRCh38
rs753325042 Chr.14: 77329064-77329064 on Build GRCh38
rs780287277 Chr.14: 77329190-77329190 on Build GRCh38
rs773354209 Chr.14: 77329482-77329482 on Build GRCh38
rs776120437 Chr.14: 77329097-77329097 on Build GRCh38
rs1000573549 Chr.14: 77329333-77329333 on Build GRCh38
rs531931416 Chr.14: 77329253-77329253 on Build GRCh38
rs542487494 Chr.14: 77329289-77329289 on Build GRCh38
rs8177567 Chr.14: 77329096-77329096 on Build GRCh38
rs8177566 Chr.14: 77329063-77329063 on Build GRCh38
rs149349623 Chr.14: 77329175-77329175 on Build GRCh38
rs199798136 Chr.14: 77329179-77329179 on Build GRCh38
rs778152963 Chr.14: 77329237-77329237 on Build GRCh38
rs916614546 Chr.14: 77329381-77329381 on Build GRCh38
rs535181954 Chr.14: 77329493-77329493 on Build GRCh38
rs940860827 Chr.14: 77329468-77329468 on Build GRCh38
rs751418985 Chr.14: 77329173-77329173 on Build GRCh38
rs756710006 Chr.14: 77329092-77329092 on Build GRCh38
rs758778372 Chr.14: 77329098-77329099 on Build GRCh38
rs8177568 Chr.14: 77329317-77329317 on Build GRCh38
rs762940952 Chr.14: 77329162-77329162 on Build GRCh38
rs928159420 Chr.14: 77329428-77329428 on Build GRCh38
rs780464896 Chr.14: 77329129-77329130 on Build GRCh38
rs201393813 Chr.14: 77329238-77329238 on Build GRCh38
rs747436622 Chr.14: 77329282-77329282 on Build GRCh38
rs758723768 Chr.14: 77329232-77329232 on Build GRCh38
rs867686264 Chr.14: 77329054-77329054 on Build GRCh38
rs779837133 Chr.14: 77329264-77329265 on Build GRCh38
rs903632314 Chr.14: 77329049-77329049 on Build GRCh38
rs892159280 Chr.14: 77329345-77329345 on Build GRCh38
rs748589210 Chr.14: 77329211-77329211 on Build GRCh38
rs183812061 Chr.14: 77329336-77329336 on Build GRCh38
rs572260974 Chr.14: 77329114-77329114 on Build GRCh38
rs187348145 Chr.14: 77329379-77329379 on Build GRCh38

View Full Product Details