Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95157252-95157755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DICER1
Entrez Gene ID
23405
Gene Name
dicer 1, ribonuclease III
Gene Aliases
DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
UniGene
Hs.738957
Gene Chromosome Location
Chr.14: 95086228-95158010 on Build GRCh38
Transcripts
NM_001291628.1 NM_030621.4 NM_177438.2 XM_011536599.1 XM_011536600.2 XM_011536601.2 XM_011536602.2 XM_017021121.1 XM_017021122.1 NR_015415.1

Gene Symbol
DICER1-AS1
Entrez Gene ID
400242
Gene Name
DICER1 antisense RNA 1
Gene Aliases
DICER1-AS
UniGene
Hs.497573
Gene Chromosome Location
Chr.14: 95157688-95179933 on Build GRCh38
Transcripts
NM_001291628.1 NM_030621.4 NM_177438.2 XM_011536599.1 XM_011536600.2 XM_011536601.2 XM_011536602.2 XM_017021121.1 XM_017021122.1 NR_015415.1

SNP Detail

SNP ID Chromosome Location
rs770078914 Chr.14: 95157720-95157720 on Build GRCh38
rs968813382 Chr.14: 95157497-95157497 on Build GRCh38
rs1022323298 Chr.14: 95157642-95157642 on Build GRCh38
rs537358585 Chr.14: 95157470-95157470 on Build GRCh38
rs980738209 Chr.14: 95157676-95157676 on Build GRCh38
rs1049767824 Chr.14: 95157576-95157576 on Build GRCh38
rs1038064876 Chr.14: 95157552-95157552 on Build GRCh38
rs1021038113 Chr.14: 95157318-95157319 on Build GRCh38
rs925857434 Chr.14: 95157625-95157625 on Build GRCh38
rs906058055 Chr.14: 95157536-95157536 on Build GRCh38
rs142780931 Chr.14: 95157454-95157454 on Build GRCh38
rs550985594 Chr.14: 95157608-95157608 on Build GRCh38
rs1033197648 Chr.14: 95157323-95157323 on Build GRCh38
rs912012608 Chr.14: 95157345-95157345 on Build GRCh38
rs1034799127 Chr.14: 95157296-95157296 on Build GRCh38
rs1008321753 Chr.14: 95157616-95157616 on Build GRCh38
rs913298422 Chr.14: 95157515-95157515 on Build GRCh38
rs919258059 Chr.14: 95157366-95157366 on Build GRCh38
rs969011312 Chr.14: 95157643-95157643 on Build GRCh38
rs938452991 Chr.14: 95157513-95157513 on Build GRCh38
rs531286635 Chr.14: 95157475-95157475 on Build GRCh38
rs930600372 Chr.14: 95157374-95157374 on Build GRCh38
rs1029473275 Chr.14: 95157586-95157586 on Build GRCh38
rs989096096 Chr.14: 95157486-95157486 on Build GRCh38
rs913254526 Chr.14: 95157663-95157663 on Build GRCh38
rs929088002 Chr.14: 95157523-95157532 on Build GRCh38
rs897759002 Chr.14: 95157459-95157480 on Build GRCh38
rs931917707 Chr.14: 95157711-95157711 on Build GRCh38
rs1015290810 Chr.14: 95157564-95157564 on Build GRCh38
rs1037591684 Chr.14: 95157449-95157449 on Build GRCh38
rs576280784 Chr.14: 95157465-95157465 on Build GRCh38
rs940521486 Chr.14: 95157546-95157546 on Build GRCh38
rs1033250064 Chr.14: 95157463-95157463 on Build GRCh38
rs907723299 Chr.14: 95157537-95157537 on Build GRCh38
rs1019795538 Chr.14: 95157476-95157476 on Build GRCh38
rs541144434 Chr.14: 95157574-95157574 on Build GRCh38
rs145124430 Chr.14: 95157667-95157668 on Build GRCh38
rs924787385 Chr.14: 95157428-95157428 on Build GRCh38
rs937510435 Chr.14: 95157678-95157678 on Build GRCh38
rs115683360 Chr.14: 95157456-95157456 on Build GRCh38
rs956155937 Chr.14: 95157479-95157479 on Build GRCh38
rs572262415 Chr.14: 95157707-95157707 on Build GRCh38
rs967649056 Chr.14: 95157319-95157319 on Build GRCh38
rs896211480 Chr.14: 95157303-95157303 on Build GRCh38
rs978992253 Chr.14: 95157320-95157321 on Build GRCh38
rs1004760745 Chr.14: 95157438-95157438 on Build GRCh38
rs925909916 Chr.14: 95157490-95157490 on Build GRCh38
rs986688077 Chr.14: 95157340-95157340 on Build GRCh38
rs1010385440 Chr.14: 95157636-95157636 on Build GRCh38
rs547495405 Chr.14: 95157348-95157348 on Build GRCh38
rs770094099 Chr.14: 95157669-95157670 on Build GRCh38
rs896276398 Chr.14: 95157426-95157426 on Build GRCh38
rs940554114 Chr.14: 95157699-95157699 on Build GRCh38
rs532700009 Chr.14: 95157502-95157502 on Build GRCh38
rs574001203 Chr.14: 95157534-95157534 on Build GRCh38
rs886050947 Chr.14: 95157376-95157376 on Build GRCh38
rs1002013373 Chr.14: 95157286-95157286 on Build GRCh38
rs907629799 Chr.14: 95157690-95157690 on Build GRCh38
rs926923405 Chr.14: 95157507-95157507 on Build GRCh38
rs956230884 Chr.14: 95157633-95157633 on Build GRCh38
rs953566859 Chr.14: 95157327-95157327 on Build GRCh38
rs1029420869 Chr.14: 95157710-95157710 on Build GRCh38
rs992573426 Chr.14: 95157682-95157682 on Build GRCh38
rs771616336 Chr.14: 95157494-95157494 on Build GRCh38
rs1041173035 Chr.14: 95157728-95157728 on Build GRCh38
rs1056692589 Chr.14: 95157393-95157393 on Build GRCh38
rs747751993 Chr.14: 95157668-95157669 on Build GRCh38
rs535171242 Chr.14: 95157306-95157306 on Build GRCh38
rs886050946 Chr.14: 95157340-95157340 on Build GRCh38
rs937855901 Chr.14: 95157392-95157392 on Build GRCh38
rs531601246 Chr.14: 95157724-95157724 on Build GRCh38
rs996464861 Chr.14: 95157573-95157573 on Build GRCh38
rs1049024578 Chr.14: 95157376-95157376 on Build GRCh38
rs1019530853 Chr.14: 95157467-95157467 on Build GRCh38
rs950152647 Chr.14: 95157665-95157665 on Build GRCh38
rs1048972576 Chr.14: 95157277-95157277 on Build GRCh38
rs905039570 Chr.14: 95157279-95157279 on Build GRCh38
rs903543920 Chr.14: 95157458-95157458 on Build GRCh38
rs888071515 Chr.14: 95157558-95157558 on Build GRCh38
rs905092027 Chr.14: 95157391-95157391 on Build GRCh38
rs889750174 Chr.14: 95157611-95157611 on Build GRCh38
rs546068564 Chr.14: 95157712-95157712 on Build GRCh38
rs899066266 Chr.14: 95157560-95157560 on Build GRCh38
rs1037529992 Chr.14: 95157702-95157702 on Build GRCh38
rs1019702426 Chr.14: 95157623-95157623 on Build GRCh38
rs1001711212 Chr.14: 95157544-95157544 on Build GRCh38
rs552464480 Chr.14: 95157679-95157679 on Build GRCh38
rs1037082878 Chr.14: 95157693-95157693 on Build GRCh38
rs886250670 Chr.14: 95157430-95157430 on Build GRCh38
rs886050945 Chr.14: 95157328-95157328 on Build GRCh38
rs959665959 Chr.14: 95157681-95157681 on Build GRCh38
rs978828838 Chr.14: 95157326-95157326 on Build GRCh38
rs566126911 Chr.14: 95157360-95157360 on Build GRCh38
rs564278393 Chr.14: 95157714-95157714 on Build GRCh38

View Full Product Details