Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102933446-102933942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654722 c.5108G>A p.G1703D Chr.14: 102933740-102933740 on Build GRCh38
3493932 c.5071C>T p.P1691S Chr.14: 102933777-102933777 on Build GRCh38
4846255 c.5078C>G p.S1693C Chr.14: 102933770-102933770 on Build GRCh38
1707899 c.5065_5066CC>TT p.P1689L Chr.14: 102933782-102933783 on Build GRCh38
3711428 c.5021C>T p.T1674I Chr.14: 102933827-102933827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775493204 Chr.14: 102933891-102933891 on Build GRCh38
rs768331940 Chr.14: 102933816-102933816 on Build GRCh38
rs996655888 Chr.14: 102933481-102933481 on Build GRCh38
rs761881673 Chr.14: 102933700-102933700 on Build GRCh38
rs866840115 Chr.14: 102933509-102933509 on Build GRCh38
rs546321376 Chr.14: 102933623-102933623 on Build GRCh38
rs900497486 Chr.14: 102933564-102933564 on Build GRCh38
rs761440584 Chr.14: 102933900-102933900 on Build GRCh38
rs970445027 Chr.14: 102933704-102933704 on Build GRCh38
rs1051753126 Chr.14: 102933634-102933634 on Build GRCh38
rs765191399 Chr.14: 102933809-102933809 on Build GRCh38
rs192828362 Chr.14: 102933545-102933545 on Build GRCh38
rs932077513 Chr.14: 102933632-102933632 on Build GRCh38
rs371638840 Chr.14: 102933763-102933763 on Build GRCh38
rs535677589 Chr.14: 102933820-102933820 on Build GRCh38
rs745757978 Chr.14: 102933665-102933665 on Build GRCh38
rs758823144 Chr.14: 102933744-102933744 on Build GRCh38
rs768478720 Chr.14: 102933859-102933859 on Build GRCh38
rs546976531 Chr.14: 102933894-102933894 on Build GRCh38
rs540652024 Chr.14: 102933562-102933562 on Build GRCh38
rs183748397 Chr.14: 102933901-102933901 on Build GRCh38
rs935293874 Chr.14: 102933847-102933847 on Build GRCh38
rs924154033 Chr.14: 102933550-102933550 on Build GRCh38
rs201614344 Chr.14: 102933686-102933686 on Build GRCh38
rs376036277 Chr.14: 102933781-102933781 on Build GRCh38
rs534270033 Chr.14: 102933872-102933872 on Build GRCh38
rs781663654 Chr.14: 102933748-102933748 on Build GRCh38
rs147122849 Chr.14: 102933558-102933558 on Build GRCh38
rs539160551 Chr.14: 102933477-102933477 on Build GRCh38
rs57314686 Chr.14: 102933585-102933585 on Build GRCh38
rs867664425 Chr.14: 102933836-102933836 on Build GRCh38
rs1031262602 Chr.14: 102933522-102933522 on Build GRCh38
rs755227527 Chr.14: 102933503-102933503 on Build GRCh38
rs933824918 Chr.14: 102933805-102933805 on Build GRCh38
rs548033579 Chr.14: 102933691-102933691 on Build GRCh38
rs1037751500 Chr.14: 102933491-102933491 on Build GRCh38
rs761685326 Chr.14: 102933775-102933775 on Build GRCh38
rs532515816 Chr.14: 102933597-102933597 on Build GRCh38
rs772966692 Chr.14: 102933799-102933799 on Build GRCh38
rs375495475 Chr.14: 102933879-102933879 on Build GRCh38
rs760303410 Chr.14: 102933765-102933765 on Build GRCh38
rs958421736 Chr.14: 102933528-102933528 on Build GRCh38
rs949501000 Chr.14: 102933547-102933547 on Build GRCh38
rs941158225 Chr.14: 102933610-102933610 on Build GRCh38
rs767627879 Chr.14: 102933789-102933789 on Build GRCh38
rs1030412379 Chr.14: 102933490-102933490 on Build GRCh38
rs1051769678 Chr.14: 102933600-102933600 on Build GRCh38
rs909804402 Chr.14: 102933609-102933609 on Build GRCh38
rs778842513 Chr.14: 102933653-102933653 on Build GRCh38
rs758672847 Chr.14: 102933824-102933824 on Build GRCh38
rs750844426 Chr.14: 102933843-102933843 on Build GRCh38
rs1027061201 Chr.14: 102933526-102933526 on Build GRCh38
rs575837504 Chr.14: 102933515-102933515 on Build GRCh38
rs778703289 Chr.14: 102933862-102933862 on Build GRCh38
rs764290721 Chr.14: 102933825-102933825 on Build GRCh38
rs367568433 Chr.14: 102933678-102933678 on Build GRCh38
rs977864022 Chr.14: 102933727-102933727 on Build GRCh38
rs780343468 Chr.14: 102933764-102933765 on Build GRCh38
rs767539948 Chr.14: 102933707-102933707 on Build GRCh38
rs764049477 Chr.14: 102933790-102933790 on Build GRCh38
rs752816236 Chr.14: 102933729-102933729 on Build GRCh38
rs895079916 Chr.14: 102933899-102933899 on Build GRCh38
rs1035150130 Chr.14: 102933912-102933912 on Build GRCh38
rs751989434 Chr.14: 102933832-102933832 on Build GRCh38
rs755007496 Chr.14: 102933721-102933721 on Build GRCh38
rs369647222 Chr.14: 102933808-102933808 on Build GRCh38
rs35239675 Chr.14: 102933745-102933745 on Build GRCh38
rs865957197 Chr.14: 102933855-102933855 on Build GRCh38
rs376636312 Chr.14: 102933682-102933682 on Build GRCh38
rs572178376 Chr.14: 102933865-102933865 on Build GRCh38
rs774458674 Chr.14: 102933699-102933699 on Build GRCh38
rs774336264 Chr.14: 102933769-102933769 on Build GRCh38
rs35064888 Chr.14: 102933666-102933666 on Build GRCh38
rs978272393 Chr.14: 102933548-102933548 on Build GRCh38
rs780861413 Chr.14: 102933529-102933531 on Build GRCh38
rs181816130 Chr.14: 102933876-102933876 on Build GRCh38
rs1024631415 Chr.14: 102933905-102933905 on Build GRCh38
rs541659344 Chr.14: 102933521-102933521 on Build GRCh38
rs200227249 Chr.14: 102933857-102933857 on Build GRCh38
rs140640502 Chr.14: 102933747-102933747 on Build GRCh38
rs115854678 Chr.14: 102933511-102933511 on Build GRCh38
rs781727740 Chr.14: 102933838-102933838 on Build GRCh38
rs10139783 Chr.14: 102933648-102933648 on Build GRCh38
rs924097013 Chr.14: 102933739-102933739 on Build GRCh38
rs776322004 Chr.14: 102933854-102933854 on Build GRCh38
rs990531350 Chr.14: 102933542-102933542 on Build GRCh38
rs775226335 Chr.14: 102933764-102933764 on Build GRCh38
rs548479650 Chr.14: 102933661-102933661 on Build GRCh38
rs35416106 Chr.14: 102933875-102933875 on Build GRCh38
rs765510912 Chr.14: 102933728-102933728 on Build GRCh38
rs758646022 Chr.14: 102933716-102933717 on Build GRCh38
rs372216187 Chr.14: 102933664-102933664 on Build GRCh38
rs757553589 Chr.14: 102933835-102933835 on Build GRCh38
rs959110113 Chr.14: 102933539-102933539 on Build GRCh38
rs755677490 Chr.14: 102933848-102933848 on Build GRCh38
rs562965814 Chr.14: 102933645-102933645 on Build GRCh38
rs927665463 Chr.14: 102933579-102933580 on Build GRCh38
rs770880009 Chr.14: 102933880-102933880 on Build GRCh38
rs562551309 Chr.14: 102933761-102933761 on Build GRCh38
rs1051697726 Chr.14: 102933788-102933788 on Build GRCh38
rs771945177 Chr.14: 102933866-102933866 on Build GRCh38
rs762818594 Chr.14: 102933692-102933692 on Build GRCh38

View Full Product Details