Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104931880-104932381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD4
Entrez Gene ID
122618
Gene Name
phospholipase D family member 4
Gene Aliases
C14orf175
UniGene
Hs.407101
Gene Chromosome Location
Chr.14: 104924806-104937789 on Build GRCh38
Transcripts
NM_001308174.1 NM_138790.3 XM_011536411.2 XM_017020965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5821720 c.1008-1G>A p.? Chr.14: 104932011-104932011 on Build GRCh38
3885710 c.1186G>A p.A396T Chr.14: 104932139-104932139 on Build GRCh38
6321979 c.1242G>T p.P414P Chr.14: 104932276-104932276 on Build GRCh38
6343337 c.1218G>T p.V406V Chr.14: 104932171-104932171 on Build GRCh38
5718728 c.1037G>A p.R346Q Chr.14: 104932041-104932041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754321469 Chr.14: 104932011-104932011 on Build GRCh38
rs763068874 Chr.14: 104932284-104932284 on Build GRCh38
rs770428272 Chr.14: 104932041-104932041 on Build GRCh38
rs371939654 Chr.14: 104932111-104932111 on Build GRCh38
rs759628235 Chr.14: 104932157-104932157 on Build GRCh38
rs750026647 Chr.14: 104932225-104932225 on Build GRCh38
rs757528734 Chr.14: 104931904-104931904 on Build GRCh38
rs771834884 Chr.14: 104932032-104932032 on Build GRCh38
rs577930915 Chr.14: 104932060-104932060 on Build GRCh38
rs373382635 Chr.14: 104932151-104932151 on Build GRCh38
rs755486070 Chr.14: 104932016-104932016 on Build GRCh38
rs759416582 Chr.14: 104931976-104931976 on Build GRCh38
rs779560333 Chr.14: 104932188-104932188 on Build GRCh38
rs780421637 Chr.14: 104932098-104932103 on Build GRCh38
rs557134091 Chr.14: 104932153-104932153 on Build GRCh38
rs980452281 Chr.14: 104932124-104932124 on Build GRCh38
rs190449169 Chr.14: 104932137-104932137 on Build GRCh38
rs769308299 Chr.14: 104932117-104932117 on Build GRCh38
rs373643392 Chr.14: 104932070-104932070 on Build GRCh38
rs767786871 Chr.14: 104931993-104931993 on Build GRCh38
rs781219285 Chr.14: 104931905-104931905 on Build GRCh38
rs766460978 Chr.14: 104932068-104932068 on Build GRCh38
rs745428855 Chr.14: 104932114-104932114 on Build GRCh38
rs755811970 Chr.14: 104932257-104932257 on Build GRCh38
rs775340893 Chr.14: 104932118-104932118 on Build GRCh38
rs779617815 Chr.14: 104932073-104932073 on Build GRCh38
rs747336001 Chr.14: 104932210-104932241 on Build GRCh38
rs778308503 Chr.14: 104932098-104932098 on Build GRCh38
rs750617640 Chr.14: 104932064-104932064 on Build GRCh38
rs183433704 Chr.14: 104932352-104932352 on Build GRCh38
rs777506608 Chr.14: 104932033-104932033 on Build GRCh38
rs764526194 Chr.14: 104932175-104932175 on Build GRCh38
rs771887856 Chr.14: 104931968-104931968 on Build GRCh38
rs780026398 Chr.14: 104932005-104932005 on Build GRCh38
rs763279228 Chr.14: 104931975-104931975 on Build GRCh38
rs754891880 Chr.14: 104932261-104932261 on Build GRCh38
rs1044245532 Chr.14: 104931922-104931922 on Build GRCh38
rs1007834582 Chr.14: 104932362-104932362 on Build GRCh38
rs749252923 Chr.14: 104932119-104932119 on Build GRCh38
rs117846862 Chr.14: 104932361-104932361 on Build GRCh38
rs753498497 Chr.14: 104932358-104932358 on Build GRCh38
rs756568076 Chr.14: 104931911-104931911 on Build GRCh38
rs762407353 Chr.14: 104932316-104932316 on Build GRCh38
rs779507202 Chr.14: 104932258-104932258 on Build GRCh38
rs762019106 Chr.14: 104931992-104931992 on Build GRCh38
rs34261826 Chr.14: 104932050-104932050 on Build GRCh38
rs750659366 Chr.14: 104932003-104932003 on Build GRCh38
rs774682950 Chr.14: 104932310-104932310 on Build GRCh38
rs890343032 Chr.14: 104932281-104932281 on Build GRCh38
rs760918977 Chr.14: 104932146-104932146 on Build GRCh38
rs776057514 Chr.14: 104932044-104932044 on Build GRCh38
rs756150793 Chr.14: 104932004-104932004 on Build GRCh38
rs752531584 Chr.14: 104931990-104931990 on Build GRCh38
rs61745186 Chr.14: 104932207-104932207 on Build GRCh38
rs768388440 Chr.14: 104932127-104932127 on Build GRCh38
rs779386022 Chr.14: 104932020-104932020 on Build GRCh38
rs545456164 Chr.14: 104932278-104932278 on Build GRCh38
rs928173825 Chr.14: 104932156-104932156 on Build GRCh38
rs778655047 Chr.14: 104932269-104932269 on Build GRCh38
rs768787608 Chr.14: 104932295-104932295 on Build GRCh38
rs756922214 Chr.14: 104932179-104932179 on Build GRCh38
rs1044680437 Chr.14: 104932177-104932177 on Build GRCh38
rs375752994 Chr.14: 104932193-104932193 on Build GRCh38
rs769022908 Chr.14: 104931927-104931927 on Build GRCh38
rs772840836 Chr.14: 104931973-104931973 on Build GRCh38
rs746653468 Chr.14: 104932107-104932107 on Build GRCh38
rs753210276 Chr.14: 104932161-104932161 on Build GRCh38
rs753267679 Chr.14: 104932082-104932082 on Build GRCh38
rs767957768 Chr.14: 104932224-104932224 on Build GRCh38
rs747778753 Chr.14: 104931965-104931965 on Build GRCh38
rs373627327 Chr.14: 104932347-104932347 on Build GRCh38
rs528566264 Chr.14: 104932213-104932213 on Build GRCh38
rs748863521 Chr.14: 104932259-104932259 on Build GRCh38
rs745541259 Chr.14: 104932185-104932185 on Build GRCh38
rs368763697 Chr.14: 104931902-104931902 on Build GRCh38
rs113269427 Chr.14: 104932201-104932201 on Build GRCh38
rs765388718 Chr.14: 104932202-104932202 on Build GRCh38
rs751939506 Chr.14: 104932176-104932176 on Build GRCh38
rs750675759 Chr.14: 104932220-104932220 on Build GRCh38
rs758765691 Chr.14: 104932084-104932084 on Build GRCh38
rs780554033 Chr.14: 104931926-104931926 on Build GRCh38
rs74316182 Chr.14: 104932055-104932055 on Build GRCh38
rs746674903 Chr.14: 104932035-104932035 on Build GRCh38
rs1031258522 Chr.14: 104932074-104932074 on Build GRCh38
rs750901959 Chr.14: 104932061-104932062 on Build GRCh38
rs907056447 Chr.14: 104932178-104932178 on Build GRCh38
rs373902639 Chr.14: 104931910-104931910 on Build GRCh38
rs899838595 Chr.14: 104932017-104932017 on Build GRCh38
rs764571811 Chr.14: 104932210-104932210 on Build GRCh38
rs1011188055 Chr.14: 104932043-104932043 on Build GRCh38
rs773536737 Chr.14: 104931899-104931899 on Build GRCh38
rs755502080 Chr.14: 104932071-104932071 on Build GRCh38
rs1011069335 Chr.14: 104932099-104932099 on Build GRCh38
rs867343562 Chr.14: 104932148-104932148 on Build GRCh38
rs763883111 Chr.14: 104932063-104932063 on Build GRCh38
rs759489682 Chr.14: 104932054-104932054 on Build GRCh38
rs574109751 Chr.14: 104932186-104932186 on Build GRCh38
rs765063111 Chr.14: 104931984-104931984 on Build GRCh38
rs780651605 Chr.14: 104932182-104932182 on Build GRCh38
rs770533954 Chr.14: 104932110-104932110 on Build GRCh38
rs760827962 Chr.14: 104932067-104932067 on Build GRCh38
rs116640235 Chr.14: 104931974-104931974 on Build GRCh38
rs765409152 Chr.14: 104932159-104932159 on Build GRCh38
rs929333550 Chr.14: 104932136-104932136 on Build GRCh38
rs774877978 Chr.14: 104931963-104931963 on Build GRCh38
rs956442773 Chr.14: 104932090-104932090 on Build GRCh38
rs747948069 Chr.14: 104932194-104932194 on Build GRCh38
rs757496975 Chr.14: 104932214-104932214 on Build GRCh38
rs570648047 Chr.14: 104932197-104932197 on Build GRCh38
rs376735811 Chr.14: 104932102-104932102 on Build GRCh38
rs767749330 Chr.14: 104932333-104932333 on Build GRCh38
rs966594082 Chr.14: 104932115-104932115 on Build GRCh38
rs775755238 Chr.14: 104932206-104932206 on Build GRCh38
rs749199589 Chr.14: 104932192-104932192 on Build GRCh38
rs771721622 Chr.14: 104932275-104932275 on Build GRCh38
rs746059278 Chr.14: 104932276-104932276 on Build GRCh38
rs748118744 Chr.14: 104932274-104932274 on Build GRCh38
rs758884819 Chr.14: 104932066-104932067 on Build GRCh38
rs200861056 Chr.14: 104932198-104932198 on Build GRCh38
rs748586346 Chr.14: 104932021-104932021 on Build GRCh38
rs759025986 Chr.14: 104932174-104932174 on Build GRCh38
rs750825198 Chr.14: 104932336-104932336 on Build GRCh38
rs759019260 Chr.14: 104932144-104932144 on Build GRCh38

View Full Product Details