Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105472860-105473364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP2
Entrez Gene ID
1397
Gene Name
cysteine rich protein 2
Gene Aliases
CRIP, CRP2, ESP1
UniGene
Hs.534309
Gene Chromosome Location
Chr.14: 105472938-105480170 on Build GRCh38
Transcripts
NM_001270837.1 NR_073082.1

SNP Detail

SNP ID Chromosome Location
rs587612655 Chr.14: 105473219-105473219 on Build GRCh38
rs782782227 Chr.14: 105473328-105473328 on Build GRCh38
rs910113771 Chr.14: 105473176-105473176 on Build GRCh38
rs980933668 Chr.14: 105473215-105473215 on Build GRCh38
rs377353696 Chr.14: 105472901-105472901 on Build GRCh38
rs1045286855 Chr.14: 105472900-105472900 on Build GRCh38
rs587665362 Chr.14: 105473222-105473222 on Build GRCh38
rs965185539 Chr.14: 105473078-105473078 on Build GRCh38
rs945690007 Chr.14: 105473252-105473252 on Build GRCh38
rs587714708 Chr.14: 105473198-105473198 on Build GRCh38
rs781891130 Chr.14: 105473304-105473304 on Build GRCh38
rs752602932 Chr.14: 105472896-105472896 on Build GRCh38
rs1006668523 Chr.14: 105473019-105473019 on Build GRCh38
rs891426491 Chr.14: 105472949-105472949 on Build GRCh38
rs1057259743 Chr.14: 105473245-105473245 on Build GRCh38
rs369075117 Chr.14: 105473202-105473202 on Build GRCh38
rs782328647 Chr.14: 105473327-105473327 on Build GRCh38
rs1056885256 Chr.14: 105472943-105472943 on Build GRCh38
rs587594527 Chr.14: 105473028-105473028 on Build GRCh38
rs782118836 Chr.14: 105473327-105473328 on Build GRCh38
rs948186369 Chr.14: 105472895-105472895 on Build GRCh38
rs913003994 Chr.14: 105473248-105473248 on Build GRCh38
rs900986337 Chr.14: 105473300-105473300 on Build GRCh38
rs534634716 Chr.14: 105473052-105473067 on Build GRCh38
rs1039602887 Chr.14: 105473271-105473271 on Build GRCh38
rs781920694 Chr.14: 105473335-105473335 on Build GRCh38
rs587602462 Chr.14: 105473168-105473168 on Build GRCh38
rs1043347898 Chr.14: 105472987-105472987 on Build GRCh38
rs587736033 Chr.14: 105472934-105472934 on Build GRCh38
rs782153236 Chr.14: 105473268-105473268 on Build GRCh38
rs587670690 Chr.14: 105473071-105473071 on Build GRCh38
rs951437400 Chr.14: 105473124-105473124 on Build GRCh38
rs1042761451 Chr.14: 105473261-105473261 on Build GRCh38
rs904444889 Chr.14: 105473263-105473263 on Build GRCh38
rs927021592 Chr.14: 105472924-105472924 on Build GRCh38
rs1025741826 Chr.14: 105473094-105473094 on Build GRCh38
rs587671943 Chr.14: 105473196-105473196 on Build GRCh38
rs904477668 Chr.14: 105473010-105473010 on Build GRCh38
rs969662678 Chr.14: 105473203-105473203 on Build GRCh38
rs928079111 Chr.14: 105473216-105473216 on Build GRCh38
rs587699540 Chr.14: 105473086-105473086 on Build GRCh38
rs141146967 Chr.14: 105472907-105472907 on Build GRCh38
rs1009922053 Chr.14: 105472976-105472976 on Build GRCh38
rs782033805 Chr.14: 105473229-105473229 on Build GRCh38

View Full Product Details