Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105479189-105479686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP2
Entrez Gene ID
1397
Gene Name
cysteine rich protein 2
Gene Aliases
CRIP, CRP2, ESP1
UniGene
Hs.534309
Gene Chromosome Location
Chr.14: 105472938-105480170 on Build GRCh38
Transcripts
NM_001270837.1 NM_001270841.1 NM_001312.3 NR_073081.1 NR_073082.1 NR_073083.1 NR_073084.1 NR_073085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554892 c.611G>A p.G204D Chr.14: 105479637-105479637 on Build GRCh38
4049574 c.549C>T p.F183F Chr.14: 105479483-105479483 on Build GRCh38
4049573 c.518A>G p.Y173C Chr.14: 105479452-105479452 on Build GRCh38
5847715 c.485C>T p.P162L Chr.14: 105479203-105479203 on Build GRCh38
697767 c.516C>G p.P172P Chr.14: 105479450-105479450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782128703 Chr.14: 105479484-105479484 on Build GRCh38
rs201353545 Chr.14: 105479596-105479596 on Build GRCh38
rs147969410 Chr.14: 105479615-105479615 on Build GRCh38
rs782005577 Chr.14: 105479551-105479551 on Build GRCh38
rs112961323 Chr.14: 105479441-105479441 on Build GRCh38
rs782481300 Chr.14: 105479489-105479489 on Build GRCh38
rs374234792 Chr.14: 105479439-105479439 on Build GRCh38
rs1020015243 Chr.14: 105479362-105479362 on Build GRCh38
rs942540528 Chr.14: 105479272-105479272 on Build GRCh38
rs782237184 Chr.14: 105479230-105479230 on Build GRCh38
rs782160175 Chr.14: 105479240-105479240 on Build GRCh38
rs587651277 Chr.14: 105479304-105479304 on Build GRCh38
rs781900033 Chr.14: 105479528-105479528 on Build GRCh38
rs781987341 Chr.14: 105479420-105479420 on Build GRCh38
rs782391580 Chr.14: 105479419-105479419 on Build GRCh38
rs782332892 Chr.14: 105479541-105479541 on Build GRCh38
rs372566753 Chr.14: 105479566-105479566 on Build GRCh38
rs782324929 Chr.14: 105479608-105479608 on Build GRCh38
rs371661421 Chr.14: 105479508-105479508 on Build GRCh38
rs190426348 Chr.14: 105479546-105479546 on Build GRCh38
rs782493052 Chr.14: 105479493-105479493 on Build GRCh38
rs782270461 Chr.14: 105479399-105479399 on Build GRCh38
rs781908396 Chr.14: 105479629-105479629 on Build GRCh38
rs1035562263 Chr.14: 105479339-105479339 on Build GRCh38
rs922463911 Chr.14: 105479278-105479278 on Build GRCh38
rs782214229 Chr.14: 105479636-105479636 on Build GRCh38
rs782741903 Chr.14: 105479432-105479432 on Build GRCh38
rs368608563 Chr.14: 105479444-105479444 on Build GRCh38
rs782790533 Chr.14: 105479627-105479627 on Build GRCh38
rs782366208 Chr.14: 105479617-105479617 on Build GRCh38
rs782571495 Chr.14: 105479504-105479504 on Build GRCh38
rs975691685 Chr.14: 105479273-105479273 on Build GRCh38
rs978629484 Chr.14: 105479257-105479257 on Build GRCh38
rs925891188 Chr.14: 105479265-105479265 on Build GRCh38
rs782152210 Chr.14: 105479553-105479553 on Build GRCh38
rs782799685 Chr.14: 105479466-105479466 on Build GRCh38
rs200862313 Chr.14: 105479411-105479411 on Build GRCh38
rs374436573 Chr.14: 105479423-105479423 on Build GRCh38
rs781904647 Chr.14: 105479262-105479262 on Build GRCh38
rs782004249 Chr.14: 105479506-105479506 on Build GRCh38
rs587656148 Chr.14: 105479462-105479462 on Build GRCh38
rs782529567 Chr.14: 105479268-105479268 on Build GRCh38
rs781979606 Chr.14: 105479664-105479664 on Build GRCh38
rs375528137 Chr.14: 105479532-105479532 on Build GRCh38
rs587644713 Chr.14: 105479268-105479268 on Build GRCh38
rs782515020 Chr.14: 105479536-105479536 on Build GRCh38
rs375776764 Chr.14: 105479570-105479570 on Build GRCh38
rs587701482 Chr.14: 105479531-105479531 on Build GRCh38
rs781783153 Chr.14: 105479483-105479483 on Build GRCh38
rs376270982 Chr.14: 105479456-105479456 on Build GRCh38
rs782604391 Chr.14: 105479577-105479577 on Build GRCh38
rs370967523 Chr.14: 105479573-105479573 on Build GRCh38
rs369904386 Chr.14: 105479534-105479534 on Build GRCh38
rs782806169 Chr.14: 105479623-105479623 on Build GRCh38
rs782726914 Chr.14: 105479490-105479490 on Build GRCh38
rs782478387 Chr.14: 105479645-105479645 on Build GRCh38
rs782062453 Chr.14: 105479563-105479563 on Build GRCh38
rs201777436 Chr.14: 105479465-105479465 on Build GRCh38
rs587747584 Chr.14: 105479621-105479621 on Build GRCh38
rs367581179 Chr.14: 105479464-105479464 on Build GRCh38
rs782705976 Chr.14: 105479613-105479613 on Build GRCh38
rs1032926506 Chr.14: 105479405-105479405 on Build GRCh38
rs587602810 Chr.14: 105479246-105479246 on Build GRCh38
rs782326079 Chr.14: 105479207-105479207 on Build GRCh38
rs201991904 Chr.14: 105479632-105479632 on Build GRCh38
rs587679419 Chr.14: 105479293-105479293 on Build GRCh38
rs782114039 Chr.14: 105479569-105479569 on Build GRCh38
rs1002460344 Chr.14: 105479338-105479338 on Build GRCh38
rs782583030 Chr.14: 105479553-105479554 on Build GRCh38
rs111531362 Chr.14: 105479537-105479537 on Build GRCh38
rs782562821 Chr.14: 105479442-105479442 on Build GRCh38
rs781918986 Chr.14: 105479555-105479555 on Build GRCh38
rs587623868 Chr.14: 105479410-105479410 on Build GRCh38
rs782519666 Chr.14: 105479438-105479438 on Build GRCh38
rs782762445 Chr.14: 105479253-105479253 on Build GRCh38
rs1056734105 Chr.14: 105479502-105479502 on Build GRCh38
rs782072966 Chr.14: 105479602-105479602 on Build GRCh38
rs782020936 Chr.14: 105479458-105479458 on Build GRCh38
rs377179054 Chr.14: 105479538-105479538 on Build GRCh38
rs782132634 Chr.14: 105479252-105479252 on Build GRCh38
rs587665278 Chr.14: 105479348-105479348 on Build GRCh38
rs782254966 Chr.14: 105479650-105479650 on Build GRCh38
rs781851248 Chr.14: 105479535-105479535 on Build GRCh38
rs926591312 Chr.14: 105479561-105479561 on Build GRCh38
rs373473162 Chr.14: 105479408-105479408 on Build GRCh38
rs369993075 Chr.14: 105479526-105479526 on Build GRCh38
rs782586339 Chr.14: 105479633-105479633 on Build GRCh38
rs782751060 Chr.14: 105479389-105479389 on Build GRCh38
rs368707395 Chr.14: 105479431-105479431 on Build GRCh38
rs587734882 Chr.14: 105479299-105479299 on Build GRCh38
rs782208576 Chr.14: 105479647-105479648 on Build GRCh38
rs144595865 Chr.14: 105479480-105479480 on Build GRCh38
rs782076047 Chr.14: 105479417-105479417 on Build GRCh38
rs370640751 Chr.14: 105479435-105479435 on Build GRCh38
rs587744349 Chr.14: 105479394-105479394 on Build GRCh38
rs782689186 Chr.14: 105479494-105479494 on Build GRCh38
rs782795301 Chr.14: 105479424-105479424 on Build GRCh38
rs782638170 Chr.14: 105479453-105479453 on Build GRCh38
rs782521512 Chr.14: 105479401-105479401 on Build GRCh38
rs887084802 Chr.14: 105479301-105479301 on Build GRCh38
rs782169252 Chr.14: 105479479-105479479 on Build GRCh38
rs375691223 Chr.14: 105479655-105479655 on Build GRCh38
rs781835354 Chr.14: 105479568-105479568 on Build GRCh38
rs1038735017 Chr.14: 105479467-105479467 on Build GRCh38
rs370433909 Chr.14: 105479389-105479389 on Build GRCh38
rs782600114 Chr.14: 105479542-105479542 on Build GRCh38
rs782639431 Chr.14: 105479649-105479649 on Build GRCh38
rs782351211 Chr.14: 105479236-105479236 on Build GRCh38
rs373998439 Chr.14: 105479529-105479529 on Build GRCh38
rs782741240 Chr.14: 105479517-105479517 on Build GRCh38
rs183771092 Chr.14: 105479661-105479661 on Build GRCh38
rs781844912 Chr.14: 105479396-105479396 on Build GRCh38
rs782044812 Chr.14: 105479514-105479514 on Build GRCh38
rs782607884 Chr.14: 105479395-105479395 on Build GRCh38
rs138027167 Chr.14: 105479662-105479662 on Build GRCh38
rs782595754 Chr.14: 105479386-105479386 on Build GRCh38
rs371698227 Chr.14: 105479447-105479447 on Build GRCh38
rs941367049 Chr.14: 105479308-105479308 on Build GRCh38
rs782457480 Chr.14: 105479503-105479503 on Build GRCh38
rs782484806 Chr.14: 105479578-105479578 on Build GRCh38
rs1019877387 Chr.14: 105479219-105479219 on Build GRCh38
rs782047009 Chr.14: 105479248-105479248 on Build GRCh38
rs181863442 Chr.14: 105479430-105479430 on Build GRCh38
rs587650667 Chr.14: 105479500-105479500 on Build GRCh38
rs782685631 Chr.14: 105479598-105479598 on Build GRCh38
rs781985704 Chr.14: 105479637-105479637 on Build GRCh38
rs11849001 Chr.14: 105479554-105479554 on Build GRCh38
rs905478390 Chr.14: 105479335-105479335 on Build GRCh38
rs587632346 Chr.14: 105479327-105479327 on Build GRCh38
rs781990899 Chr.14: 105479478-105479478 on Build GRCh38
rs782408427 Chr.14: 105479597-105479597 on Build GRCh38
rs370173238 Chr.14: 105479412-105479412 on Build GRCh38
rs782240987 Chr.14: 105479454-105479454 on Build GRCh38
rs781868766 Chr.14: 105479492-105479492 on Build GRCh38
rs1038607506 Chr.14: 105479334-105479334 on Build GRCh38
rs781930160 Chr.14: 105479247-105479247 on Build GRCh38
rs782087383 Chr.14: 105479624-105479624 on Build GRCh38
rs896670743 Chr.14: 105479341-105479341 on Build GRCh38
rs782009863 Chr.14: 105479239-105479239 on Build GRCh38
rs1042494382 Chr.14: 105479607-105479607 on Build GRCh38
rs782379631 Chr.14: 105479549-105479549 on Build GRCh38
rs967003677 Chr.14: 105479374-105479374 on Build GRCh38
rs748461792 Chr.14: 105479294-105479294 on Build GRCh38
rs782434719 Chr.14: 105479541-105479541 on Build GRCh38
rs782349570 Chr.14: 105479505-105479505 on Build GRCh38
rs782184000 Chr.14: 105479603-105479603 on Build GRCh38
rs782437580 Chr.14: 105479631-105479631 on Build GRCh38
rs782583716 Chr.14: 105479226-105479226 on Build GRCh38

View Full Product Details