Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30940819-30941333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 NM_017762.2 XM_005254508.2 XM_011521737.2 XM_017022384.1

Gene Symbol
MTMR10
Entrez Gene ID
54893
Gene Name
myotubularin related protein 10
Gene Aliases
-
UniGene
Hs.30141
Gene Chromosome Location
Chr.15: 30919764-30991604 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 NM_017762.2 XM_005254508.2 XM_011521737.2 XM_017022384.1

SNP Detail

SNP ID Chromosome Location
rs540987910 Chr.15: 30941084-30941084 on Build GRCh38
rs980005123 Chr.15: 30940871-30940871 on Build GRCh38
rs71963314 Chr.15: 30941155-30941158 on Build GRCh38
rs777798776 Chr.15: 30941120-30941120 on Build GRCh38
rs750951368 Chr.15: 30941302-30941302 on Build GRCh38
rs533902091 Chr.15: 30940867-30940867 on Build GRCh38
rs781037634 Chr.15: 30941284-30941284 on Build GRCh38
rs1038237481 Chr.15: 30940901-30940901 on Build GRCh38
rs369926441 Chr.15: 30941267-30941270 on Build GRCh38
rs553751221 Chr.15: 30940893-30940893 on Build GRCh38
rs979712282 Chr.15: 30941154-30941155 on Build GRCh38
rs1007798056 Chr.15: 30941236-30941236 on Build GRCh38
rs775655396 Chr.15: 30941247-30941247 on Build GRCh38
rs772107849 Chr.15: 30941222-30941222 on Build GRCh38
rs992670701 Chr.15: 30940856-30940856 on Build GRCh38
rs754161518 Chr.15: 30941289-30941289 on Build GRCh38
rs890757898 Chr.15: 30940977-30940977 on Build GRCh38
rs750611550 Chr.15: 30940931-30940931 on Build GRCh38
rs1039314896 Chr.15: 30941293-30941293 on Build GRCh38
rs980789604 Chr.15: 30941147-30941147 on Build GRCh38
rs762087391 Chr.15: 30941294-30941294 on Build GRCh38
rs1001536760 Chr.15: 30941048-30941048 on Build GRCh38
rs148053346 Chr.15: 30941097-30941097 on Build GRCh38
rs1044291547 Chr.15: 30941212-30941212 on Build GRCh38
rs539067846 Chr.15: 30941303-30941306 on Build GRCh38
rs945525246 Chr.15: 30941185-30941185 on Build GRCh38
rs776567616 Chr.15: 30941264-30941265 on Build GRCh38
rs778741454 Chr.15: 30940951-30940951 on Build GRCh38
rs185352339 Chr.15: 30941022-30941022 on Build GRCh38
rs764321352 Chr.15: 30941281-30941281 on Build GRCh38
rs1050794279 Chr.15: 30941243-30941243 on Build GRCh38
rs146513200 Chr.15: 30941302-30941303 on Build GRCh38
rs938377558 Chr.15: 30941167-30941167 on Build GRCh38
rs1017637410 Chr.15: 30941013-30941013 on Build GRCh38
rs772793403 Chr.15: 30941298-30941302 on Build GRCh38
rs938451350 Chr.15: 30940880-30940881 on Build GRCh38
rs969887400 Chr.15: 30941306-30941307 on Build GRCh38
rs928282529 Chr.15: 30941158-30941159 on Build GRCh38
rs754779809 Chr.15: 30940945-30940945 on Build GRCh38
rs181476201 Chr.15: 30941298-30941298 on Build GRCh38
rs917884371 Chr.15: 30940863-30940863 on Build GRCh38
rs369368315 Chr.15: 30941264-30941267 on Build GRCh38
rs761545924 Chr.15: 30941291-30941292 on Build GRCh38
rs1026362850 Chr.15: 30941290-30941290 on Build GRCh38
rs771697948 Chr.15: 30941245-30941245 on Build GRCh38
rs149134318 Chr.15: 30940910-30940910 on Build GRCh38
rs770512383 Chr.15: 30941213-30941213 on Build GRCh38
rs957095505 Chr.15: 30941083-30941083 on Build GRCh38
rs1006545717 Chr.15: 30940978-30940978 on Build GRCh38
rs141796338 Chr.15: 30941207-30941207 on Build GRCh38
rs1025096219 Chr.15: 30941098-30941098 on Build GRCh38
rs79843877 Chr.15: 30940897-30940897 on Build GRCh38
rs529973073 Chr.15: 30941168-30941168 on Build GRCh38
rs950580154 Chr.15: 30941095-30941095 on Build GRCh38
rs974418683 Chr.15: 30941199-30941199 on Build GRCh38
rs74918724 Chr.15: 30940985-30940985 on Build GRCh38
rs529748832 Chr.15: 30941259-30941259 on Build GRCh38
rs571620335 Chr.15: 30940855-30940855 on Build GRCh38
rs941261420 Chr.15: 30941182-30941182 on Build GRCh38
rs3840035 Chr.15: 30941150-30941153 on Build GRCh38
rs375604526 Chr.15: 30941101-30941101 on Build GRCh38
rs145985032 Chr.15: 30941272-30941272 on Build GRCh38
rs748917017 Chr.15: 30940950-30940950 on Build GRCh38
rs918389897 Chr.15: 30941211-30941211 on Build GRCh38
rs901837329 Chr.15: 30941279-30941279 on Build GRCh38
rs761535961 Chr.15: 30941225-30941226 on Build GRCh38
rs929807493 Chr.15: 30941237-30941237 on Build GRCh38
rs769585026 Chr.15: 30941297-30941298 on Build GRCh38
rs1022541698 Chr.15: 30940954-30940954 on Build GRCh38
rs950873439 Chr.15: 30941292-30941292 on Build GRCh38
rs190608427 Chr.15: 30941079-30941079 on Build GRCh38
rs909164086 Chr.15: 30940846-30940846 on Build GRCh38
rs143252819 Chr.15: 30940956-30940956 on Build GRCh38
rs2955792 Chr.15: 30941017-30941017 on Build GRCh38
rs867561652 Chr.15: 30940971-30940971 on Build GRCh38

View Full Product Details