Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 36697186-36697694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf41
Entrez Gene ID
84529
Gene Name
chromosome 15 open reading frame 41
Gene Aliases
HH114
UniGene
Hs.48348
Gene Chromosome Location
Chr.15: 36579603-36810260 on Build GRCh38
Transcripts
NM_001130010.2 NM_001290232.1 NM_001290233.1 NM_001321756.1 NM_001321757.1 NM_001321758.1 NM_001321759.1 NM_001321760.1 NM_001321761.1 NM_032499.5 XM_011522111.1 XM_017022676.1 XM_017022677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4915920 c.518T>C p.L173P Chr.15: 36697364-36697364 on Build GRCh38
6614421 c.479C>T p.A160V Chr.15: 36697325-36697325 on Build GRCh38
403685 c.487C>T p.H163Y Chr.15: 36697333-36697333 on Build GRCh38
700012 c.525G>T p.E175D Chr.15: 36697371-36697371 on Build GRCh38
700011 c.541C>A p.L181I Chr.15: 36697387-36697387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766683888 Chr.15: 36697380-36697380 on Build GRCh38
rs117638434 Chr.15: 36697377-36697377 on Build GRCh38
rs756521970 Chr.15: 36697328-36697328 on Build GRCh38
rs758029873 Chr.15: 36697426-36697426 on Build GRCh38
rs1055067649 Chr.15: 36697464-36697464 on Build GRCh38
rs767890960 Chr.15: 36697408-36697408 on Build GRCh38
rs962149816 Chr.15: 36697583-36697583 on Build GRCh38
rs992253368 Chr.15: 36697541-36697541 on Build GRCh38
rs779081239 Chr.15: 36697386-36697386 on Build GRCh38
rs541381891 Chr.15: 36697214-36697214 on Build GRCh38
rs947890706 Chr.15: 36697406-36697406 on Build GRCh38
rs2381887 Chr.15: 36697268-36697268 on Build GRCh38
rs376753770 Chr.15: 36697363-36697363 on Build GRCh38
rs984634929 Chr.15: 36697616-36697616 on Build GRCh38
rs778111140 Chr.15: 36697416-36697416 on Build GRCh38
rs758502812 Chr.15: 36697293-36697293 on Build GRCh38
rs776272707 Chr.15: 36697290-36697290 on Build GRCh38
rs554632617 Chr.15: 36697411-36697411 on Build GRCh38
rs552376925 Chr.15: 36697282-36697282 on Build GRCh38
rs114426703 Chr.15: 36697499-36697499 on Build GRCh38
rs568314973 Chr.15: 36697369-36697369 on Build GRCh38
rs1013607710 Chr.15: 36697477-36697477 on Build GRCh38
rs771142126 Chr.15: 36697306-36697307 on Build GRCh38
rs559143211 Chr.15: 36697505-36697505 on Build GRCh38
rs560206006 Chr.15: 36697228-36697228 on Build GRCh38
rs372408032 Chr.15: 36697657-36697657 on Build GRCh38
rs1007045084 Chr.15: 36697487-36697487 on Build GRCh38
rs750016091 Chr.15: 36697425-36697425 on Build GRCh38
rs779819437 Chr.15: 36697430-36697430 on Build GRCh38
rs999693349 Chr.15: 36697522-36697522 on Build GRCh38
rs755428859 Chr.15: 36697310-36697310 on Build GRCh38
rs370576472 Chr.15: 36697484-36697484 on Build GRCh38
rs779439290 Chr.15: 36697341-36697341 on Build GRCh38
rs768471317 Chr.15: 36697361-36697361 on Build GRCh38
rs74008749 Chr.15: 36697668-36697668 on Build GRCh38
rs570537996 Chr.15: 36697300-36697300 on Build GRCh38
rs1023366751 Chr.15: 36697579-36697579 on Build GRCh38
rs551637332 Chr.15: 36697523-36697523 on Build GRCh38
rs1031221210 Chr.15: 36697534-36697534 on Build GRCh38
rs372427497 Chr.15: 36697340-36697340 on Build GRCh38
rs1014626964 Chr.15: 36697537-36697537 on Build GRCh38
rs756433899 Chr.15: 36697413-36697413 on Build GRCh38
rs529341356 Chr.15: 36697302-36697302 on Build GRCh38
rs894724954 Chr.15: 36697469-36697469 on Build GRCh38
rs748432862 Chr.15: 36697321-36697321 on Build GRCh38
rs765523796 Chr.15: 36697276-36697276 on Build GRCh38
rs778493101 Chr.15: 36697334-36697334 on Build GRCh38
rs771008872 Chr.15: 36697280-36697280 on Build GRCh38
rs762537245 Chr.15: 36697586-36697586 on Build GRCh38
rs373199182 Chr.15: 36697405-36697405 on Build GRCh38
rs375876944 Chr.15: 36697421-36697421 on Build GRCh38
rs759187567 Chr.15: 36697303-36697303 on Build GRCh38
rs746585714 Chr.15: 36697360-36697360 on Build GRCh38
rs1003223024 Chr.15: 36697528-36697528 on Build GRCh38
rs587777100 Chr.15: 36697379-36697379 on Build GRCh38
rs897122174 Chr.15: 36697459-36697459 on Build GRCh38
rs1011907864 Chr.15: 36697226-36697226 on Build GRCh38
rs550097623 Chr.15: 36697368-36697368 on Build GRCh38
rs751587651 Chr.15: 36697394-36697394 on Build GRCh38
rs370084293 Chr.15: 36697314-36697314 on Build GRCh38
rs994670810 Chr.15: 36697602-36697602 on Build GRCh38
rs771857774 Chr.15: 36697446-36697446 on Build GRCh38
rs936191533 Chr.15: 36697420-36697420 on Build GRCh38
rs773360978 Chr.15: 36697570-36697570 on Build GRCh38
rs1051162012 Chr.15: 36697466-36697466 on Build GRCh38

View Full Product Details