Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40251383-40251888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B-PAK6
Entrez Gene ID
106821730
Gene Name
BUB1B-PAK6 readthrough
Gene Aliases
-
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40217428-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
C15orf56
Entrez Gene ID
644809
Gene Name
chromosome 15 open reading frame 56
Gene Aliases
-
UniGene
Hs.631718
Gene Chromosome Location
Chr.15: 40250664-40252909 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
PAK6
Entrez Gene ID
56924
Gene Name
p21 (RAC1) activated kinase 6
Gene Aliases
PAK5
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40239091-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

SNP Detail

SNP ID Chromosome Location
rs183885930 Chr.15: 40251425-40251425 on Build GRCh38
rs763001163 Chr.15: 40251593-40251593 on Build GRCh38
rs964016855 Chr.15: 40251465-40251465 on Build GRCh38
rs200225996 Chr.15: 40251440-40251440 on Build GRCh38
rs926051890 Chr.15: 40251840-40251840 on Build GRCh38
rs188960247 Chr.15: 40251631-40251631 on Build GRCh38
rs761013786 Chr.15: 40251490-40251490 on Build GRCh38
rs948592099 Chr.15: 40251817-40251817 on Build GRCh38
rs937640434 Chr.15: 40251660-40251660 on Build GRCh38
rs776765237 Chr.15: 40251416-40251420 on Build GRCh38
rs920941045 Chr.15: 40251411-40251411 on Build GRCh38
rs981491175 Chr.15: 40251632-40251632 on Build GRCh38
rs74817232 Chr.15: 40251701-40251701 on Build GRCh38
rs920133531 Chr.15: 40251685-40251685 on Build GRCh38
rs961291777 Chr.15: 40251744-40251744 on Build GRCh38
rs965461991 Chr.15: 40251668-40251668 on Build GRCh38
rs1009039801 Chr.15: 40251633-40251633 on Build GRCh38
rs948841702 Chr.15: 40251510-40251510 on Build GRCh38
rs1056914259 Chr.15: 40251466-40251466 on Build GRCh38
rs904512590 Chr.15: 40251595-40251595 on Build GRCh38
rs1032186943 Chr.15: 40251476-40251476 on Build GRCh38
rs886891187 Chr.15: 40251808-40251808 on Build GRCh38
rs983776219 Chr.15: 40251735-40251735 on Build GRCh38
rs1047232987 Chr.15: 40251795-40251799 on Build GRCh38
rs1040753770 Chr.15: 40251854-40251854 on Build GRCh38
rs11634409 Chr.15: 40251413-40251413 on Build GRCh38
rs980195486 Chr.15: 40251834-40251834 on Build GRCh38
rs952524761 Chr.15: 40251412-40251412 on Build GRCh38
rs1044754014 Chr.15: 40251519-40251519 on Build GRCh38
rs1019684513 Chr.15: 40251458-40251458 on Build GRCh38
rs765262786 Chr.15: 40251641-40251641 on Build GRCh38
rs550925603 Chr.15: 40251441-40251441 on Build GRCh38
rs867618498 Chr.15: 40251642-40251642 on Build GRCh38
rs368226860 Chr.15: 40251638-40251638 on Build GRCh38
rs1053345195 Chr.15: 40251609-40251609 on Build GRCh38
rs939840219 Chr.15: 40251464-40251464 on Build GRCh38
rs917093893 Chr.15: 40251767-40251767 on Build GRCh38
rs1000631231 Chr.15: 40251606-40251606 on Build GRCh38
rs908445481 Chr.15: 40251737-40251737 on Build GRCh38
rs917294821 Chr.15: 40251483-40251483 on Build GRCh38
rs987911684 Chr.15: 40251525-40251525 on Build GRCh38
rs751722845 Chr.15: 40251727-40251727 on Build GRCh38
rs1028525415 Chr.15: 40251714-40251714 on Build GRCh38
rs992589724 Chr.15: 40251766-40251766 on Build GRCh38
rs913331412 Chr.15: 40251580-40251580 on Build GRCh38

View Full Product Details