Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52064329-52064827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK6
Entrez Gene ID
5597
Gene Name
mitogen-activated protein kinase 6
Gene Aliases
ERK3, HsT17250, PRKM6, p97MAPK
UniGene
Hs.411847
Gene Chromosome Location
Chr.15: 52004265-52066265 on Build GRCh38
Transcripts
NM_002748.3 XM_005254537.2 XM_005254538.2 XM_005254539.3 XM_011521782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177502 c.1906A>G p.K636E Chr.15: 52064740-52064740 on Build GRCh38
6001833 c.1888G>T p.D630Y Chr.15: 52064722-52064722 on Build GRCh38
5980759 c.1726G>A p.E576K Chr.15: 52064560-52064560 on Build GRCh38
6291783 c.1569G>T p.Q523H Chr.15: 52064403-52064403 on Build GRCh38
6142728 c.1613G>C p.S538T Chr.15: 52064447-52064447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971644977 Chr.15: 52064625-52064633 on Build GRCh38
rs146538100 Chr.15: 52064680-52064680 on Build GRCh38
rs770254975 Chr.15: 52064469-52064471 on Build GRCh38
rs541397063 Chr.15: 52064753-52064753 on Build GRCh38
rs773819403 Chr.15: 52064611-52064611 on Build GRCh38
rs758492531 Chr.15: 52064653-52064653 on Build GRCh38
rs771284250 Chr.15: 52064564-52064567 on Build GRCh38
rs756607646 Chr.15: 52064697-52064697 on Build GRCh38
rs375509015 Chr.15: 52064514-52064514 on Build GRCh38
rs753062718 Chr.15: 52064762-52064762 on Build GRCh38
rs779300139 Chr.15: 52064435-52064435 on Build GRCh38
rs764137758 Chr.15: 52064737-52064737 on Build GRCh38
rs748711903 Chr.15: 52064694-52064694 on Build GRCh38
rs778276565 Chr.15: 52064775-52064775 on Build GRCh38
rs771954953 Chr.15: 52064711-52064711 on Build GRCh38
rs767505442 Chr.15: 52064526-52064526 on Build GRCh38
rs148487587 Chr.15: 52064352-52064352 on Build GRCh38
rs775137132 Chr.15: 52064721-52064721 on Build GRCh38
rs748571787 Chr.15: 52064582-52064582 on Build GRCh38
rs761159355 Chr.15: 52064377-52064377 on Build GRCh38
rs866931478 Chr.15: 52064450-52064450 on Build GRCh38
rs747529399 Chr.15: 52064455-52064455 on Build GRCh38
rs535430008 Chr.15: 52064478-52064478 on Build GRCh38
rs148363441 Chr.15: 52064798-52064798 on Build GRCh38
rs575423229 Chr.15: 52064633-52064633 on Build GRCh38
rs187167606 Chr.15: 52064705-52064705 on Build GRCh38
rs781388541 Chr.15: 52064685-52064685 on Build GRCh38
rs767061120 Chr.15: 52064706-52064714 on Build GRCh38
rs750726475 Chr.15: 52064551-52064551 on Build GRCh38
rs766130172 Chr.15: 52064397-52064397 on Build GRCh38
rs774927895 Chr.15: 52064646-52064648 on Build GRCh38
rs765474727 Chr.15: 52064648-52064648 on Build GRCh38
rs751499153 Chr.15: 52064404-52064404 on Build GRCh38
rs780546950 Chr.15: 52064451-52064451 on Build GRCh38
rs749756922 Chr.15: 52064784-52064784 on Build GRCh38
rs769249333 Chr.15: 52064490-52064490 on Build GRCh38
rs757448967 Chr.15: 52064545-52064545 on Build GRCh38
rs776658192 Chr.15: 52064738-52064738 on Build GRCh38
rs748812099 Chr.15: 52064701-52064701 on Build GRCh38
rs749687244 Chr.15: 52064537-52064540 on Build GRCh38
rs747410314 Chr.15: 52064560-52064560 on Build GRCh38
rs372920091 Chr.15: 52064604-52064604 on Build GRCh38
rs779108097 Chr.15: 52064547-52064547 on Build GRCh38
rs757108172 Chr.15: 52064785-52064785 on Build GRCh38
rs139563932 Chr.15: 52064500-52064500 on Build GRCh38
rs371736330 Chr.15: 52064709-52064709 on Build GRCh38
rs778268878 Chr.15: 52064791-52064791 on Build GRCh38
rs748241660 Chr.15: 52064707-52064708 on Build GRCh38
rs760279697 Chr.15: 52064756-52064757 on Build GRCh38
rs957798218 Chr.15: 52064704-52064707 on Build GRCh38
rs776941865 Chr.15: 52064471-52064471 on Build GRCh38
rs749006261 Chr.15: 52064592-52064592 on Build GRCh38
rs149225460 Chr.15: 52064706-52064706 on Build GRCh38
rs759032674 Chr.15: 52064703-52064704 on Build GRCh38
rs777783622 Chr.15: 52064410-52064410 on Build GRCh38
rs142670170 Chr.15: 52064394-52064394 on Build GRCh38
rs776198020 Chr.15: 52064365-52064365 on Build GRCh38
rs766218454 Chr.15: 52064507-52064507 on Build GRCh38
rs143744875 Chr.15: 52064557-52064557 on Build GRCh38
rs772891162 Chr.15: 52064499-52064499 on Build GRCh38
rs752528392 Chr.15: 52064528-52064528 on Build GRCh38
rs773656764 Chr.15: 52064580-52064580 on Build GRCh38
rs371262771 Chr.15: 52064386-52064386 on Build GRCh38
rs767177059 Chr.15: 52064628-52064628 on Build GRCh38
rs759397866 Chr.15: 52064615-52064615 on Build GRCh38
rs769343002 Chr.15: 52064586-52064586 on Build GRCh38
rs756832443 Chr.15: 52064787-52064787 on Build GRCh38
rs369666891 Chr.15: 52064793-52064793 on Build GRCh38
rs751856795 Chr.15: 52064742-52064742 on Build GRCh38
rs778184504 Chr.15: 52064698-52064698 on Build GRCh38
rs537289987 Chr.15: 52064609-52064609 on Build GRCh38
rs769208660 Chr.15: 52064467-52064467 on Build GRCh38
rs368094632 Chr.15: 52064437-52064437 on Build GRCh38
rs768101670 Chr.15: 52064363-52064363 on Build GRCh38
rs1017123008 Chr.15: 52064519-52064519 on Build GRCh38
rs574779395 Chr.15: 52064795-52064795 on Build GRCh38
rs374841305 Chr.15: 52064392-52064392 on Build GRCh38
rs757227852 Chr.15: 52064430-52064430 on Build GRCh38
rs376041003 Chr.15: 52064521-52064522 on Build GRCh38
rs772694895 Chr.15: 52064590-52064590 on Build GRCh38
rs376638037 Chr.15: 52064558-52064558 on Build GRCh38
rs199988498 Chr.15: 52064421-52064421 on Build GRCh38
rs201436164 Chr.15: 52064704-52064704 on Build GRCh38
rs371987419 Chr.15: 52064472-52064472 on Build GRCh38
rs781736100 Chr.15: 52064578-52064578 on Build GRCh38
rs759188992 Chr.15: 52064522-52064522 on Build GRCh38
rs751900303 Chr.15: 52064440-52064440 on Build GRCh38
rs754927155 Chr.15: 52064405-52064405 on Build GRCh38
rs766643187 Chr.15: 52064660-52064660 on Build GRCh38
rs1049590425 Chr.15: 52064436-52064436 on Build GRCh38
rs750530035 Chr.15: 52064652-52064652 on Build GRCh38
rs559991113 Chr.15: 52064760-52064760 on Build GRCh38
rs368759134 Chr.15: 52064567-52064567 on Build GRCh38
rs144209444 Chr.15: 52064663-52064663 on Build GRCh38
rs143385036 Chr.15: 52064739-52064739 on Build GRCh38
rs901138217 Chr.15: 52064403-52064403 on Build GRCh38
rs751367729 Chr.15: 52064512-52064512 on Build GRCh38
rs760763262 Chr.15: 52064726-52064726 on Build GRCh38
rs772608176 Chr.15: 52064438-52064438 on Build GRCh38
rs756415242 Chr.15: 52064765-52064765 on Build GRCh38

View Full Product Details